Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153811771-153812008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD2B
Entrez Gene ID
57459
Gene Name
GATA zinc finger domain containing 2B
Gene Aliases
MRD18, P66beta, p68
UniGene
Hs.4779
Gene Chromosome Location
Chr.1: 153804725-153922975 on Build GRCh38
Transcripts
XM_005245364.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4651955 c.1604A>G p.Q535R Chr.1: 153811775-153811775 on Build GRCh38
6121114 c.1580G>T p.R527L Chr.1: 153811799-153811799 on Build GRCh38
4403075 c.1532C>T p.A511V Chr.1: 153811847-153811847 on Build GRCh38
5982481 c.1581C>A p.R527R Chr.1: 153811798-153811798 on Build GRCh38
1626463 c.1575T>A p.S525S Chr.1: 153811804-153811804 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888065478 Chr.1: 153811948-153811948 on Build GRCh38
rs771649812 Chr.1: 153811880-153811880 on Build GRCh38
rs907029794 Chr.1: 153811931-153811931 on Build GRCh38
rs752726274 Chr.1: 153811852-153811853 on Build GRCh38
rs779112430 Chr.1: 153811876-153811876 on Build GRCh38
rs1024262134 Chr.1: 153811904-153811904 on Build GRCh38
rs756052339 Chr.1: 153811845-153811845 on Build GRCh38
rs368995332 Chr.1: 153811862-153811862 on Build GRCh38
rs1016061270 Chr.1: 153811946-153811946 on Build GRCh38
rs763844698 Chr.1: 153811943-153811943 on Build GRCh38
rs369291714 Chr.1: 153811889-153811889 on Build GRCh38
rs778858637 Chr.1: 153811868-153811868 on Build GRCh38
rs373092458 Chr.1: 153811869-153811869 on Build GRCh38
rs762200565 Chr.1: 153811876-153811876 on Build GRCh38
rs770999798 Chr.1: 153811870-153811870 on Build GRCh38
rs1005444549 Chr.1: 153811967-153811967 on Build GRCh38
rs779594037 Chr.1: 153811881-153811881 on Build GRCh38
rs751609777 Chr.1: 153811837-153811837 on Build GRCh38
rs763089701 Chr.1: 153811820-153811820 on Build GRCh38
rs749512166 Chr.1: 153811860-153811861 on Build GRCh38
rs374873172 Chr.1: 153811882-153811882 on Build GRCh38
rs371331593 Chr.1: 153811972-153811972 on Build GRCh38
rs868300276 Chr.1: 153811817-153811817 on Build GRCh38
rs201644866 Chr.1: 153811891-153811891 on Build GRCh38
rs766442642 Chr.1: 153811821-153811821 on Build GRCh38
rs183352699 Chr.1: 153811930-153811930 on Build GRCh38
rs777493201 Chr.1: 153811859-153811861 on Build GRCh38
rs753679123 Chr.1: 153811866-153811866 on Build GRCh38
rs777182533 Chr.1: 153811896-153811896 on Build GRCh38
rs995443222 Chr.1: 153811965-153811965 on Build GRCh38
rs757063342 Chr.1: 153811867-153811867 on Build GRCh38
rs573175094 Chr.1: 153811977-153811977 on Build GRCh38
rs774589064 Chr.1: 153811800-153811800 on Build GRCh38
rs879124596 Chr.1: 153811854-153811854 on Build GRCh38
rs755904863 Chr.1: 153811854-153811856 on Build GRCh38
rs1027925567 Chr.1: 153811902-153811902 on Build GRCh38

View Full Product Details