Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1352116-1352369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6586955 c.110+8G>A p.? Chr.16: 1352167-1352167 on Build GRCh38
4769028 c.114G>A p.V38V Chr.16: 1352242-1352242 on Build GRCh38
6522474 c.81G>A p.M27I Chr.16: 1352130-1352130 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879184661 Chr.16: 1352196-1352196 on Build GRCh38
rs771564346 Chr.16: 1352258-1352258 on Build GRCh38
rs774278289 Chr.16: 1352242-1352242 on Build GRCh38
rs935869758 Chr.16: 1352240-1352240 on Build GRCh38
rs760002927 Chr.16: 1352186-1352186 on Build GRCh38
rs894088899 Chr.16: 1352250-1352250 on Build GRCh38
rs367901619 Chr.16: 1352329-1352329 on Build GRCh38
rs1017158084 Chr.16: 1352344-1352344 on Build GRCh38
rs889922749 Chr.16: 1352294-1352294 on Build GRCh38
rs768574619 Chr.16: 1352172-1352172 on Build GRCh38
rs8052503 Chr.16: 1352142-1352142 on Build GRCh38
rs753223909 Chr.16: 1352236-1352236 on Build GRCh38
rs201064884 Chr.16: 1352177-1352177 on Build GRCh38
rs984755560 Chr.16: 1352140-1352140 on Build GRCh38
rs761328581 Chr.16: 1352176-1352176 on Build GRCh38
rs550093407 Chr.16: 1352203-1352203 on Build GRCh38
rs538632263 Chr.16: 1352266-1352266 on Build GRCh38
rs772880608 Chr.16: 1352184-1352184 on Build GRCh38
rs967101407 Chr.16: 1352287-1352287 on Build GRCh38
rs1039291038 Chr.16: 1352245-1352245 on Build GRCh38
rs959739306 Chr.16: 1352345-1352345 on Build GRCh38
rs749067743 Chr.16: 1352229-1352229 on Build GRCh38
rs11554987 Chr.16: 1352243-1352243 on Build GRCh38
rs779889768 Chr.16: 1352216-1352216 on Build GRCh38
rs372379104 Chr.16: 1352241-1352241 on Build GRCh38
rs779788991 Chr.16: 1352146-1352146 on Build GRCh38
rs927145995 Chr.16: 1352234-1352234 on Build GRCh38
rs375508895 Chr.16: 1352254-1352254 on Build GRCh38
rs756294894 Chr.16: 1352213-1352213 on Build GRCh38
rs901250980 Chr.16: 1352288-1352288 on Build GRCh38
rs552514133 Chr.16: 1352275-1352275 on Build GRCh38
rs938573026 Chr.16: 1352148-1352148 on Build GRCh38
rs746148448 Chr.16: 1352212-1352212 on Build GRCh38
rs941874907 Chr.16: 1352189-1352189 on Build GRCh38
rs961662599 Chr.16: 1352349-1352349 on Build GRCh38
rs749262844 Chr.16: 1352151-1352151 on Build GRCh38
rs773791004 Chr.16: 1352174-1352174 on Build GRCh38
rs991432590 Chr.16: 1352150-1352150 on Build GRCh38
rs1035733324 Chr.16: 1352332-1352332 on Build GRCh38
rs764689491 Chr.16: 1352202-1352202 on Build GRCh38
rs753083772 Chr.16: 1352191-1352191 on Build GRCh38
rs1011071248 Chr.16: 1352272-1352272 on Build GRCh38
rs765747805 Chr.16: 1352190-1352190 on Build GRCh38
rs368478807 Chr.16: 1352289-1352289 on Build GRCh38
rs570064313 Chr.16: 1352209-1352209 on Build GRCh38
rs763356168 Chr.16: 1352333-1352333 on Build GRCh38
rs1049855574 Chr.16: 1352292-1352292 on Build GRCh38
rs757317192 Chr.16: 1352208-1352208 on Build GRCh38
rs530066253 Chr.16: 1352201-1352201 on Build GRCh38
rs928343311 Chr.16: 1352145-1352145 on Build GRCh38

View Full Product Details