Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78125728-78125953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC2
Entrez Gene ID
54810
Gene Name
GIPC PDZ domain containing family member 2
Gene Aliases
SEMCAP-2, SEMCAP2
UniGene
Hs.603818
Gene Chromosome Location
Chr.1: 78044962-78138449 on Build GRCh38
Transcripts
NM_001304725.1 NM_017655.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412572 c.751G>A p.D251N Chr.1: 78125917-78125917 on Build GRCh38
6009342 c.775G>A p.G259R Chr.1: 78125941-78125941 on Build GRCh38
6497022 c.771C>A p.Y257* Chr.1: 78125937-78125937 on Build GRCh38
1344437 c.781C>T p.R261* Chr.1: 78125947-78125947 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1018774229 Chr.1: 78125791-78125791 on Build GRCh38
rs917176427 Chr.1: 78125893-78125893 on Build GRCh38
rs546127172 Chr.1: 78125792-78125792 on Build GRCh38
rs949945545 Chr.1: 78125914-78125914 on Build GRCh38
rs749416541 Chr.1: 78125918-78125918 on Build GRCh38
rs369813473 Chr.1: 78125875-78125875 on Build GRCh38
rs368288141 Chr.1: 78125863-78125863 on Build GRCh38
rs982600583 Chr.1: 78125921-78125921 on Build GRCh38
rs758380091 Chr.1: 78125922-78125922 on Build GRCh38
rs750676869 Chr.1: 78125858-78125858 on Build GRCh38
rs113524451 Chr.1: 78125886-78125886 on Build GRCh38
rs370621119 Chr.1: 78125821-78125821 on Build GRCh38
rs989498922 Chr.1: 78125839-78125839 on Build GRCh38
rs767711147 Chr.1: 78125847-78125847 on Build GRCh38
rs543663714 Chr.1: 78125926-78125926 on Build GRCh38
rs757667763 Chr.1: 78125835-78125835 on Build GRCh38
rs978174976 Chr.1: 78125797-78125797 on Build GRCh38
rs201839129 Chr.1: 78125859-78125859 on Build GRCh38
rs754465363 Chr.1: 78125786-78125786 on Build GRCh38
rs779950635 Chr.1: 78125862-78125862 on Build GRCh38

View Full Product Details