Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 21705953-21706192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA2
Entrez Gene ID
2675
Gene Name
GDNF family receptor alpha 2
Gene Aliases
GDNFRB, NRTNR-ALPHA, NTNRA, RETL2, TRNR2
UniGene
Hs.441202
Gene Chromosome Location
Chr.8: 21690403-21789296 on Build GRCh38
Transcripts
NM_001165038.1 NM_001165039.1 NM_001495.4 XM_006716327.3 XM_011544484.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395222 c.800G>A p.R267Q Chr.8: 21706036-21706036 on Build GRCh38
5457392 c.807C>A p.A269A Chr.8: 21706029-21706029 on Build GRCh38
6241889 c.860C>A p.A287E Chr.8: 21705976-21705976 on Build GRCh38
3358250 c.842C>T p.T281M Chr.8: 21705994-21705994 on Build GRCh38
5409175 c.827G>A p.R276Q Chr.8: 21706009-21706009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377524016 Chr.8: 21706029-21706029 on Build GRCh38
rs182661508 Chr.8: 21706151-21706151 on Build GRCh38
rs567654178 Chr.8: 21706133-21706133 on Build GRCh38
rs965430299 Chr.8: 21706088-21706088 on Build GRCh38
rs376318521 Chr.8: 21706107-21706107 on Build GRCh38
rs987336820 Chr.8: 21706137-21706137 on Build GRCh38
rs761005936 Chr.8: 21706075-21706076 on Build GRCh38
rs373522339 Chr.8: 21706000-21706000 on Build GRCh38
rs759890890 Chr.8: 21706111-21706111 on Build GRCh38
rs945527327 Chr.8: 21706172-21706172 on Build GRCh38
rs371123175 Chr.8: 21706067-21706067 on Build GRCh38
rs950660043 Chr.8: 21706124-21706124 on Build GRCh38
rs921236274 Chr.8: 21706117-21706117 on Build GRCh38
rs773188702 Chr.8: 21705993-21705993 on Build GRCh38
rs567874667 Chr.8: 21705976-21705976 on Build GRCh38
rs975397184 Chr.8: 21706101-21706101 on Build GRCh38
rs139085327 Chr.8: 21705985-21705985 on Build GRCh38
rs553428283 Chr.8: 21705987-21705987 on Build GRCh38
rs757402273 Chr.8: 21706041-21706041 on Build GRCh38
rs767024460 Chr.8: 21706115-21706115 on Build GRCh38
rs780751093 Chr.8: 21705986-21705986 on Build GRCh38
rs779222466 Chr.8: 21705973-21705973 on Build GRCh38
rs759366064 Chr.8: 21706010-21706010 on Build GRCh38
rs545406729 Chr.8: 21706060-21706060 on Build GRCh38
rs867649826 Chr.8: 21706030-21706030 on Build GRCh38
rs773986301 Chr.8: 21706004-21706004 on Build GRCh38
rs1002493322 Chr.8: 21705979-21705979 on Build GRCh38
rs1026794849 Chr.8: 21706009-21706009 on Build GRCh38
rs775306784 Chr.8: 21706021-21706021 on Build GRCh38
rs936756906 Chr.8: 21706129-21706129 on Build GRCh38
rs1053829442 Chr.8: 21706136-21706136 on Build GRCh38
rs748771049 Chr.8: 21705994-21705994 on Build GRCh38
rs1018767378 Chr.8: 21706148-21706148 on Build GRCh38
rs764027004 Chr.8: 21706032-21706032 on Build GRCh38
rs755451167 Chr.8: 21706077-21706077 on Build GRCh38
rs578092449 Chr.8: 21706052-21706052 on Build GRCh38
rs767277620 Chr.8: 21706015-21706015 on Build GRCh38
rs753879133 Chr.8: 21706038-21706038 on Build GRCh38
rs1025577226 Chr.8: 21706074-21706074 on Build GRCh38
rs765625029 Chr.8: 21706047-21706047 on Build GRCh38

View Full Product Details