Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30112576-30112849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD3
Entrez Gene ID
79153
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases
GDE7
UniGene
Hs.289015
Gene Chromosome Location
Chr.16: 30104810-30113557 on Build GRCh38
Transcripts
NM_024307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1678992 c.281C>A p.S94* Chr.16: 30112695-30112695 on Build GRCh38
7139545 c.217G>T p.D73Y Chr.16: 30112759-30112759 on Build GRCh38
2156748 c.261_263delTGA p.D87delD Chr.16: 30112713-30112715 on Build GRCh38
703310 c.300G>T p.V100V Chr.16: 30112676-30112676 on Build GRCh38
5567137 c.292A>G p.R98G Chr.16: 30112684-30112684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201511312 Chr.16: 30112744-30112744 on Build GRCh38
rs769777350 Chr.16: 30112734-30112734 on Build GRCh38
rs749615608 Chr.16: 30112678-30112678 on Build GRCh38
rs930633968 Chr.16: 30112831-30112831 on Build GRCh38
rs375485050 Chr.16: 30112751-30112751 on Build GRCh38
rs367918036 Chr.16: 30112701-30112701 on Build GRCh38
rs1018816054 Chr.16: 30112694-30112694 on Build GRCh38
rs750330096 Chr.16: 30112650-30112650 on Build GRCh38
rs764359103 Chr.16: 30112606-30112606 on Build GRCh38
rs761529339 Chr.16: 30112726-30112726 on Build GRCh38
rs768792433 Chr.16: 30112708-30112708 on Build GRCh38
rs61764619 Chr.16: 30112743-30112743 on Build GRCh38
rs762792576 Chr.16: 30112829-30112829 on Build GRCh38
rs928667830 Chr.16: 30112792-30112810 on Build GRCh38
rs945809323 Chr.16: 30112760-30112760 on Build GRCh38
rs774855444 Chr.16: 30112813-30112814 on Build GRCh38
rs775547129 Chr.16: 30112600-30112600 on Build GRCh38
rs775485096 Chr.16: 30112702-30112702 on Build GRCh38
rs780570506 Chr.16: 30112799-30112799 on Build GRCh38
rs950144814 Chr.16: 30112638-30112638 on Build GRCh38
rs752449266 Chr.16: 30112776-30112776 on Build GRCh38
rs781121631 Chr.16: 30112818-30112818 on Build GRCh38
rs746443339 Chr.16: 30112698-30112698 on Build GRCh38
rs748405421 Chr.16: 30112826-30112827 on Build GRCh38
rs751057816 Chr.16: 30112747-30112747 on Build GRCh38
rs780854305 Chr.16: 30112674-30112674 on Build GRCh38
rs773160281 Chr.16: 30112826-30112826 on Build GRCh38
rs776313331 Chr.16: 30112619-30112619 on Build GRCh38
rs767055474 Chr.16: 30112807-30112807 on Build GRCh38
rs758404610 Chr.16: 30112656-30112656 on Build GRCh38
rs1004708839 Chr.16: 30112675-30112675 on Build GRCh38
rs74746402 Chr.16: 30112625-30112625 on Build GRCh38
rs138427877 Chr.16: 30112695-30112695 on Build GRCh38
rs58842311 Chr.16: 30112813-30112813 on Build GRCh38
rs766026177 Chr.16: 30112657-30112657 on Build GRCh38
rs751057196 Chr.16: 30112660-30112660 on Build GRCh38
rs375857248 Chr.16: 30112633-30112633 on Build GRCh38
rs1055263935 Chr.16: 30112800-30112800 on Build GRCh38
rs367999022 Chr.16: 30112782-30112782 on Build GRCh38
rs760850563 Chr.16: 30112603-30112603 on Build GRCh38
rs776846892 Chr.16: 30112709-30112709 on Build GRCh38
rs758578117 Chr.16: 30112629-30112629 on Build GRCh38
rs758930580 Chr.16: 30112783-30112783 on Build GRCh38
rs770115803 Chr.16: 30112820-30112820 on Build GRCh38
rs192236532 Chr.16: 30112775-30112775 on Build GRCh38
rs754609869 Chr.16: 30112661-30112661 on Build GRCh38
rs755761103 Chr.16: 30112779-30112779 on Build GRCh38
rs549898215 Chr.16: 30112780-30112780 on Build GRCh38
rs747062560 Chr.16: 30112703-30112703 on Build GRCh38
rs202089119 Chr.16: 30112623-30112623 on Build GRCh38
rs761629427 Chr.16: 30112628-30112628 on Build GRCh38
rs759914644 Chr.16: 30112815-30112816 on Build GRCh38

View Full Product Details