Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190855181-190855452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMNC
Entrez Gene ID
647309
Gene Name
geminin coiled-coil domain containing
Gene Aliases
GEMC1
UniGene
Hs.518490
Gene Chromosome Location
Chr.3: 190843271-190864200 on Build GRCh38
Transcripts
NM_001146686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275052 c.952C>A p.R318S Chr.3: 190855348-190855348 on Build GRCh38
4599097 c.1000T>A p.S334T Chr.3: 190855300-190855300 on Build GRCh38
6301559 c.879G>A p.M293I Chr.3: 190855421-190855421 on Build GRCh38
6198751 c.967G>A p.G323R Chr.3: 190855333-190855333 on Build GRCh38
1042542 c.905A>G p.H302R Chr.3: 190855395-190855395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142308680 Chr.3: 190855423-190855428 on Build GRCh38
rs530281164 Chr.3: 190855235-190855235 on Build GRCh38
rs553698264 Chr.3: 190855401-190855401 on Build GRCh38
rs532663479 Chr.3: 190855294-190855294 on Build GRCh38
rs754594580 Chr.3: 190855399-190855399 on Build GRCh38
rs766751250 Chr.3: 190855358-190855358 on Build GRCh38
rs1035571254 Chr.3: 190855389-190855389 on Build GRCh38
rs1002782300 Chr.3: 190855383-190855383 on Build GRCh38
rs752267628 Chr.3: 190855329-190855329 on Build GRCh38
rs905693591 Chr.3: 190855356-190855356 on Build GRCh38
rs991154623 Chr.3: 190855368-190855368 on Build GRCh38
rs534689713 Chr.3: 190855400-190855400 on Build GRCh38
rs1051339901 Chr.3: 190855268-190855268 on Build GRCh38
rs776630246 Chr.3: 190855345-190855345 on Build GRCh38
rs561562921 Chr.3: 190855224-190855224 on Build GRCh38
rs749054686 Chr.3: 190855292-190855292 on Build GRCh38
rs750898467 Chr.3: 190855352-190855352 on Build GRCh38
rs950393096 Chr.3: 190855396-190855396 on Build GRCh38
rs550361619 Chr.3: 190855263-190855263 on Build GRCh38
rs774065063 Chr.3: 190855313-190855313 on Build GRCh38
rs552371392 Chr.3: 190855317-190855317 on Build GRCh38
rs780569672 Chr.3: 190855295-190855295 on Build GRCh38
rs368839823 Chr.3: 190855348-190855348 on Build GRCh38
rs971543181 Chr.3: 190855397-190855397 on Build GRCh38
rs148749428 Chr.3: 190855288-190855288 on Build GRCh38
rs941537531 Chr.3: 190855247-190855247 on Build GRCh38
rs114741982 Chr.3: 190855347-190855347 on Build GRCh38
rs763417313 Chr.3: 190855355-190855355 on Build GRCh38
rs570117265 Chr.3: 190855264-190855264 on Build GRCh38
rs908925956 Chr.3: 190855245-190855245 on Build GRCh38
rs760945280 Chr.3: 190855316-190855316 on Build GRCh38
rs186044056 Chr.3: 190855221-190855221 on Build GRCh38
rs370986380 Chr.3: 190855426-190855431 on Build GRCh38
rs962259763 Chr.3: 190855293-190855293 on Build GRCh38
rs768761758 Chr.3: 190855344-190855344 on Build GRCh38
rs751937584 Chr.3: 190855394-190855394 on Build GRCh38

View Full Product Details