Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 162148963-162149194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG2
Entrez Gene ID
2566
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases
CAE2, ECA2, GEFSP3
UniGene
Hs.7195
Gene Chromosome Location
Chr.5: 162067642-162155539 on Build GRCh38
Transcripts
NM_000816.3 NM_198903.2 NM_198904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702767 c.935T>C p.V312A Chr.5: 162149120-162149120 on Build GRCh38
1634118 c.984G>T p.K328N Chr.5: 162149169-162149169 on Build GRCh38
4458662 c.974C>T p.S325L Chr.5: 162149159-162149159 on Build GRCh38
6668682 c.929C>T p.T310I Chr.5: 162149114-162149114 on Build GRCh38
3853402 c.952C>A p.L318I Chr.5: 162149137-162149137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749531013 Chr.5: 162149139-162149139 on Build GRCh38
rs796052510 Chr.5: 162149152-162149152 on Build GRCh38
rs1020493800 Chr.5: 162149043-162149043 on Build GRCh38
rs778784577 Chr.5: 162149063-162149063 on Build GRCh38
rs368940063 Chr.5: 162149057-162149057 on Build GRCh38
rs121909672 Chr.5: 162149168-162149168 on Build GRCh38
rs1012007943 Chr.5: 162149024-162149024 on Build GRCh38
rs893105285 Chr.5: 162149021-162149021 on Build GRCh38
rs745747223 Chr.5: 162149065-162149065 on Build GRCh38
rs763639081 Chr.5: 162149146-162149146 on Build GRCh38
rs768061156 Chr.5: 162149170-162149170 on Build GRCh38
rs369614121 Chr.5: 162149157-162149157 on Build GRCh38
rs753623680 Chr.5: 162149062-162149062 on Build GRCh38
rs774964109 Chr.5: 162149172-162149172 on Build GRCh38
rs763791095 Chr.5: 162149061-162149061 on Build GRCh38
rs893612232 Chr.5: 162149009-162149009 on Build GRCh38
rs113738613 Chr.5: 162149027-162149027 on Build GRCh38
rs77543973 Chr.5: 162149013-162149013 on Build GRCh38
rs1010755279 Chr.5: 162149010-162149010 on Build GRCh38
rs781394954 Chr.5: 162149077-162149077 on Build GRCh38
rs758395626 Chr.5: 162149074-162149074 on Build GRCh38
rs953183818 Chr.5: 162149017-162149017 on Build GRCh38
rs373413302 Chr.5: 162149085-162149085 on Build GRCh38
rs373993626 Chr.5: 162149163-162149163 on Build GRCh38
rs770128179 Chr.5: 162149095-162149095 on Build GRCh38
rs777083068 Chr.5: 162149048-162149048 on Build GRCh38
rs774809218 Chr.5: 162149160-162149160 on Build GRCh38
rs397514737 Chr.5: 162149153-162149153 on Build GRCh38
rs377502016 Chr.5: 162149127-162149127 on Build GRCh38

View Full Product Details