Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 893676-893931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GET4
Entrez Gene ID
51608
Gene Name
golgi to ER traffic protein 4
Gene Aliases
C7orf20, CEE, CGI-20, TRC35
UniGene
Hs.107387
Gene Chromosome Location
Chr.7: 876554-896434 on Build GRCh38
Transcripts
NM_015949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062674 c.844C>T p.L282L Chr.7: 893920-893920 on Build GRCh38
6313567 c.822G>T p.E274D Chr.7: 893815-893815 on Build GRCh38
5408295 c.807C>T p.D269D Chr.7: 893800-893800 on Build GRCh38
5408294 c.799C>T p.R267W Chr.7: 893792-893792 on Build GRCh38
5408296 c.854G>A p.G285D Chr.7: 893930-893930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924611868 Chr.7: 893803-893803 on Build GRCh38
rs773164750 Chr.7: 893892-893892 on Build GRCh38
rs777961497 Chr.7: 893815-893815 on Build GRCh38
rs13237161 Chr.7: 893743-893743 on Build GRCh38
rs199726309 Chr.7: 893895-893895 on Build GRCh38
rs748467156 Chr.7: 893800-893800 on Build GRCh38
rs749832188 Chr.7: 893872-893873 on Build GRCh38
rs367907098 Chr.7: 893833-893833 on Build GRCh38
rs763694700 Chr.7: 893722-893723 on Build GRCh38
rs753175144 Chr.7: 893906-893906 on Build GRCh38
rs376061649 Chr.7: 893733-893733 on Build GRCh38
rs1048488173 Chr.7: 893774-893774 on Build GRCh38
rs774413389 Chr.7: 893838-893838 on Build GRCh38
rs764855932 Chr.7: 893905-893905 on Build GRCh38
rs148309614 Chr.7: 893752-893752 on Build GRCh38
rs951455759 Chr.7: 893871-893872 on Build GRCh38
rs772966227 Chr.7: 893849-893849 on Build GRCh38
rs796391608 Chr.7: 893732-893733 on Build GRCh38
rs765844990 Chr.7: 893791-893791 on Build GRCh38
rs934907700 Chr.7: 893811-893811 on Build GRCh38
rs758832261 Chr.7: 893812-893812 on Build GRCh38
rs761387350 Chr.7: 893702-893702 on Build GRCh38
rs779280125 Chr.7: 893797-893797 on Build GRCh38
rs367686493 Chr.7: 893855-893855 on Build GRCh38
rs540985599 Chr.7: 893742-893742 on Build GRCh38
rs527392694 Chr.7: 893861-893861 on Build GRCh38
rs762827481 Chr.7: 893770-893770 on Build GRCh38
rs983106135 Chr.7: 893765-893765 on Build GRCh38
rs761028399 Chr.7: 893796-893796 on Build GRCh38
rs774818590 Chr.7: 893698-893698 on Build GRCh38
rs756599644 Chr.7: 893843-893844 on Build GRCh38
rs759411039 Chr.7: 893714-893714 on Build GRCh38
rs776411638 Chr.7: 893902-893902 on Build GRCh38
rs772862887 Chr.7: 893779-893779 on Build GRCh38
rs376992179 Chr.7: 893881-893881 on Build GRCh38
rs371133897 Chr.7: 893760-893760 on Build GRCh38
rs975243758 Chr.7: 893795-893795 on Build GRCh38
rs755662174 Chr.7: 893790-893790 on Build GRCh38
rs117948702 Chr.7: 893867-893867 on Build GRCh38
rs376281043 Chr.7: 893761-893761 on Build GRCh38
rs374027022 Chr.7: 893822-893822 on Build GRCh38
rs745927219 Chr.7: 893873-893873 on Build GRCh38
rs903430660 Chr.7: 893699-893699 on Build GRCh38
rs772295368 Chr.7: 893751-893751 on Build GRCh38
rs754331803 Chr.7: 893792-893792 on Build GRCh38
rs746300149 Chr.7: 893833-893834 on Build GRCh38
rs377557727 Chr.7: 893719-893719 on Build GRCh38
rs756878531 Chr.7: 893729-893729 on Build GRCh38
rs1043030979 Chr.7: 893802-893802 on Build GRCh38
rs769035295 Chr.7: 893736-893736 on Build GRCh38
rs780465656 Chr.7: 893867-893867 on Build GRCh38
rs184738143 Chr.7: 893710-893710 on Build GRCh38
rs751181659 Chr.7: 893727-893727 on Build GRCh38
rs552786006 Chr.7: 893850-893850 on Build GRCh38
rs756919659 Chr.7: 893800-893800 on Build GRCh38
rs753954736 Chr.7: 893880-893880 on Build GRCh38
rs200036257 Chr.7: 893845-893845 on Build GRCh38
rs189478048 Chr.7: 893720-893720 on Build GRCh38
rs761983299 Chr.7: 893709-893709 on Build GRCh38
rs77318420 Chr.7: 893732-893732 on Build GRCh38
rs767274141 Chr.7: 893785-893785 on Build GRCh38
rs763485148 Chr.7: 893908-893908 on Build GRCh38
rs764382449 Chr.7: 893909-893909 on Build GRCh38
rs773303014 Chr.7: 893713-893713 on Build GRCh38
rs747182756 Chr.7: 893870-893870 on Build GRCh38
rs374141149 Chr.7: 893854-893854 on Build GRCh38
rs781471641 Chr.7: 893831-893832 on Build GRCh38
rs1013314244 Chr.7: 893721-893721 on Build GRCh38
rs749957924 Chr.7: 893731-893731 on Build GRCh38
rs753302737 Chr.7: 893851-893851 on Build GRCh38
rs975211209 Chr.7: 893883-893883 on Build GRCh38
rs141406197 Chr.7: 893783-893783 on Build GRCh38
rs61747650 Chr.7: 893904-893904 on Build GRCh38
rs140715950 Chr.7: 893793-893793 on Build GRCh38
rs751127066 Chr.7: 893761-893762 on Build GRCh38
rs770495171 Chr.7: 893896-893896 on Build GRCh38
rs779178788 Chr.7: 893737-893737 on Build GRCh38
rs760707922 Chr.7: 893834-893834 on Build GRCh38
rs13237267 Chr.7: 893813-893813 on Build GRCh38
rs771060510 Chr.7: 893758-893758 on Build GRCh38
rs370699463 Chr.7: 893788-893788 on Build GRCh38
rs376367650 Chr.7: 893703-893703 on Build GRCh38
rs757431185 Chr.7: 893871-893871 on Build GRCh38
rs896100877 Chr.7: 893828-893828 on Build GRCh38
rs765168292 Chr.7: 893716-893716 on Build GRCh38
rs763846090 Chr.7: 893723-893723 on Build GRCh38
rs747283069 Chr.7: 893820-893820 on Build GRCh38
rs376984995 Chr.7: 893735-893735 on Build GRCh38

View Full Product Details