Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154891543-154891776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN5
Entrez Gene ID
25929
Gene Name
gem nuclear organelle associated protein 5
Gene Aliases
GEMIN-5
UniGene
Hs.483921
Gene Chromosome Location
Chr.5: 154887416-154938216 on Build GRCh38
Transcripts
NM_001252156.1 NM_015465.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3612911 c.3890C>T p.P1297L Chr.5: 154891613-154891613 on Build GRCh38
5793783 c.3841C>A p.L1281I Chr.5: 154891662-154891662 on Build GRCh38
3919426 c.3946C>T p.Q1316* Chr.5: 154891557-154891557 on Build GRCh38
1644634 c.3838T>A p.F1280I Chr.5: 154891665-154891665 on Build GRCh38
7369373 c.3800C>T p.S1267F Chr.5: 154891703-154891703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774430174 Chr.5: 154891573-154891573 on Build GRCh38
rs748292248 Chr.5: 154891659-154891659 on Build GRCh38
rs779462142 Chr.5: 154891615-154891615 on Build GRCh38
rs368088395 Chr.5: 154891678-154891678 on Build GRCh38
rs138987708 Chr.5: 154891705-154891705 on Build GRCh38
rs61754586 Chr.5: 154891631-154891631 on Build GRCh38
rs371642910 Chr.5: 154891594-154891594 on Build GRCh38
rs974015865 Chr.5: 154891726-154891726 on Build GRCh38
rs766429895 Chr.5: 154891569-154891569 on Build GRCh38
rs771074216 Chr.5: 154891654-154891654 on Build GRCh38
rs761352791 Chr.5: 154891710-154891710 on Build GRCh38
rs758843888 Chr.5: 154891619-154891619 on Build GRCh38
rs767450538 Chr.5: 154891673-154891673 on Build GRCh38
rs200552475 Chr.5: 154891694-154891694 on Build GRCh38
rs746787620 Chr.5: 154891713-154891714 on Build GRCh38
rs35562111 Chr.5: 154891677-154891677 on Build GRCh38
rs751878514 Chr.5: 154891742-154891742 on Build GRCh38
rs755352269 Chr.5: 154891750-154891750 on Build GRCh38
rs780541593 Chr.5: 154891740-154891740 on Build GRCh38
rs200376323 Chr.5: 154891611-154891611 on Build GRCh38
rs759416728 Chr.5: 154891666-154891666 on Build GRCh38
rs749885277 Chr.5: 154891720-154891720 on Build GRCh38
rs142095826 Chr.5: 154891664-154891664 on Build GRCh38
rs779130606 Chr.5: 154891629-154891629 on Build GRCh38
rs752490202 Chr.5: 154891584-154891584 on Build GRCh38
rs981367868 Chr.5: 154891712-154891712 on Build GRCh38
rs763795220 Chr.5: 154891704-154891704 on Build GRCh38
rs746343045 Chr.5: 154891617-154891617 on Build GRCh38
rs758131028 Chr.5: 154891589-154891589 on Build GRCh38
rs777689963 Chr.5: 154891680-154891680 on Build GRCh38
rs149877560 Chr.5: 154891737-154891737 on Build GRCh38
rs776953945 Chr.5: 154891739-154891739 on Build GRCh38
rs527600011 Chr.5: 154891600-154891600 on Build GRCh38
rs756747985 Chr.5: 154891613-154891613 on Build GRCh38
rs780303963 Chr.5: 154891646-154891646 on Build GRCh38
rs761914189 Chr.5: 154891563-154891563 on Build GRCh38
rs764702803 Chr.5: 154891715-154891715 on Build GRCh38
rs146357980 Chr.5: 154891606-154891606 on Build GRCh38
rs1004628057 Chr.5: 154891670-154891670 on Build GRCh38
rs768763545 Chr.5: 154891653-154891653 on Build GRCh38
rs200206273 Chr.5: 154891652-154891652 on Build GRCh38
rs1035721086 Chr.5: 154891634-154891634 on Build GRCh38
rs774483456 Chr.5: 154891665-154891665 on Build GRCh38
rs767197359 Chr.5: 154891574-154891574 on Build GRCh38

View Full Product Details