Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42310130-42310403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANC
Entrez Gene ID
2595
Gene Name
glucosidase alpha, neutral C
Gene Aliases
-
UniGene
Hs.730806
Gene Chromosome Location
Chr.15: 42273658-42353666 on Build GRCh38
Transcripts
NM_001301409.1 NM_198141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1708055 c.728G>A p.G243E Chr.15: 42310288-42310288 on Build GRCh38
5959557 c.769C>A p.Q257K Chr.15: 42310329-42310329 on Build GRCh38
1708056 c.800C>T p.S267L Chr.15: 42310360-42310360 on Build GRCh38
6194954 c.815T>C p.L272P Chr.15: 42310375-42310375 on Build GRCh38
6366336 c.842G>T p.G281V Chr.15: 42310402-42310402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891129983 Chr.15: 42310312-42310312 on Build GRCh38
rs199618794 Chr.15: 42310234-42310234 on Build GRCh38
rs751704578 Chr.15: 42310325-42310325 on Build GRCh38
rs979399384 Chr.15: 42310370-42310370 on Build GRCh38
rs1023518430 Chr.15: 42310291-42310291 on Build GRCh38
rs779906285 Chr.15: 42310366-42310366 on Build GRCh38
rs745862510 Chr.15: 42310273-42310273 on Build GRCh38
rs371777885 Chr.15: 42310281-42310281 on Build GRCh38
rs762115926 Chr.15: 42310300-42310300 on Build GRCh38
rs746543415 Chr.15: 42310373-42310373 on Build GRCh38
rs756710682 Chr.15: 42310334-42310334 on Build GRCh38
rs746447294 Chr.15: 42310289-42310289 on Build GRCh38
rs536624569 Chr.15: 42310335-42310335 on Build GRCh38
rs765674624 Chr.15: 42310302-42310302 on Build GRCh38
rs763584460 Chr.15: 42310314-42310314 on Build GRCh38
rs377313719 Chr.15: 42310336-42310336 on Build GRCh38
rs369899093 Chr.15: 42310184-42310184 on Build GRCh38
rs182875754 Chr.15: 42310354-42310354 on Build GRCh38
rs903015064 Chr.15: 42310374-42310374 on Build GRCh38
rs367848322 Chr.15: 42310265-42310265 on Build GRCh38
rs757040917 Chr.15: 42310257-42310257 on Build GRCh38
rs190042287 Chr.15: 42310179-42310179 on Build GRCh38
rs755063195 Chr.15: 42310332-42310332 on Build GRCh38
rs76522222 Chr.15: 42310249-42310249 on Build GRCh38
rs776296130 Chr.15: 42310299-42310299 on Build GRCh38
rs755257647 Chr.15: 42310245-42310245 on Build GRCh38
rs1038414639 Chr.15: 42310242-42310242 on Build GRCh38
rs745642975 Chr.15: 42310346-42310346 on Build GRCh38
rs143698645 Chr.15: 42310311-42310311 on Build GRCh38
rs766926987 Chr.15: 42310321-42310321 on Build GRCh38
rs778546637 Chr.15: 42310268-42310268 on Build GRCh38
rs768308557 Chr.15: 42310296-42310296 on Build GRCh38
rs778821508 Chr.15: 42310337-42310337 on Build GRCh38
rs780340425 Chr.15: 42310233-42310233 on Build GRCh38
rs749062372 Chr.15: 42310255-42310255 on Build GRCh38
rs778107670 Chr.15: 42310236-42310238 on Build GRCh38
rs559952182 Chr.15: 42310201-42310202 on Build GRCh38
rs775044138 Chr.15: 42310282-42310282 on Build GRCh38
rs534326281 Chr.15: 42310183-42310183 on Build GRCh38
rs768114217 Chr.15: 42310378-42310378 on Build GRCh38
rs1034711702 Chr.15: 42310154-42310154 on Build GRCh38

View Full Product Details