Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45350659-45350886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC2
Entrez Gene ID
2068
Gene Name
ERCC excision repair 2, TFIIH core complex helicase subunit
Gene Aliases
COFS2, EM9, TFIIH, TTD, TTD1, XPD
UniGene
Hs.487294
Gene Chromosome Location
Chr.19: 45349837-45370647 on Build GRCh38
Transcripts
XM_011526611.2 XM_017026467.1 NM_177417.2

Gene Symbol
KLC3
Entrez Gene ID
147700
Gene Name
kinesin light chain 3
Gene Aliases
KLC2, KLC2L, KLCt, KNS2B
UniGene
Hs.298079
Gene Chromosome Location
Chr.19: 45340740-45351520 on Build GRCh38
Transcripts
XM_011526611.2 XM_017026467.1 NM_177417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928495 c.1311G>A p.E437E Chr.19: 45350679-45350679 on Build GRCh38
6990685 c.1341C>G p.V447V Chr.19: 45350709-45350709 on Build GRCh38
6501114 c.1346G>A p.R449Q Chr.19: 45350714-45350714 on Build GRCh38
5390100 c.1322G>A p.R441Q Chr.19: 45350690-45350690 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005962585 Chr.19: 45350719-45350723 on Build GRCh38
rs1005305271 Chr.19: 45350694-45350694 on Build GRCh38
rs752348371 Chr.19: 45350732-45350732 on Build GRCh38
rs1022533031 Chr.19: 45350838-45350838 on Build GRCh38
rs996365259 Chr.19: 45350799-45350809 on Build GRCh38
rs983813733 Chr.19: 45350749-45350750 on Build GRCh38
rs572326676 Chr.19: 45350814-45350814 on Build GRCh38
rs3916901 Chr.19: 45350758-45350758 on Build GRCh38
rs759624982 Chr.19: 45350711-45350711 on Build GRCh38
rs796392845 Chr.19: 45350698-45350699 on Build GRCh38
rs558846191 Chr.19: 45350813-45350813 on Build GRCh38
rs879789653 Chr.19: 45350810-45350813 on Build GRCh38
rs538786185 Chr.19: 45350807-45350807 on Build GRCh38
rs779895784 Chr.19: 45350686-45350686 on Build GRCh38
rs888097215 Chr.19: 45350764-45350764 on Build GRCh38
rs751392586 Chr.19: 45350729-45350729 on Build GRCh38
rs754802873 Chr.19: 45350698-45350699 on Build GRCh38
rs896034965 Chr.19: 45350687-45350687 on Build GRCh38
rs369702798 Chr.19: 45350813-45350813 on Build GRCh38
rs567336739 Chr.19: 45350812-45350813 on Build GRCh38
rs753627014 Chr.19: 45350695-45350729 on Build GRCh38
rs756865438 Chr.19: 45350783-45350783 on Build GRCh38
rs927163692 Chr.19: 45350854-45350854 on Build GRCh38
rs757858424 Chr.19: 45350738-45350738 on Build GRCh38
rs370566239 Chr.19: 45350730-45350730 on Build GRCh38
rs766373207 Chr.19: 45350753-45350753 on Build GRCh38
rs748349233 Chr.19: 45350719-45350720 on Build GRCh38
rs780520784 Chr.19: 45350691-45350691 on Build GRCh38
rs369131208 Chr.19: 45350689-45350689 on Build GRCh38
rs746423195 Chr.19: 45350785-45350785 on Build GRCh38
rs747581328 Chr.19: 45350769-45350770 on Build GRCh38
rs531834422 Chr.19: 45350713-45350713 on Build GRCh38
rs768889397 Chr.19: 45350797-45350797 on Build GRCh38
rs377547718 Chr.19: 45350714-45350714 on Build GRCh38
rs1023411331 Chr.19: 45350703-45350703 on Build GRCh38
rs758554856 Chr.19: 45350727-45350729 on Build GRCh38
rs747727725 Chr.19: 45350695-45350695 on Build GRCh38
rs551647275 Chr.19: 45350719-45350719 on Build GRCh38
rs199968742 Chr.19: 45350755-45350755 on Build GRCh38
rs767615912 Chr.19: 45350807-45350807 on Build GRCh38
rs978531123 Chr.19: 45350818-45350818 on Build GRCh38
rs771277800 Chr.19: 45350771-45350772 on Build GRCh38
rs758179169 Chr.19: 45350683-45350683 on Build GRCh38
rs371527303 Chr.19: 45350770-45350770 on Build GRCh38
rs776705760 Chr.19: 45350747-45350747 on Build GRCh38
rs958779446 Chr.19: 45350856-45350857 on Build GRCh38
rs900221244 Chr.19: 45350782-45350782 on Build GRCh38
rs968284825 Chr.19: 45350860-45350860 on Build GRCh38
rs776996317 Chr.19: 45350799-45350803 on Build GRCh38
rs373548688 Chr.19: 45350693-45350693 on Build GRCh38
rs776652733 Chr.19: 45350799-45350799 on Build GRCh38
rs766708678 Chr.19: 45350857-45350857 on Build GRCh38
rs367981132 Chr.19: 45350766-45350766 on Build GRCh38
rs553749571 Chr.19: 45350746-45350746 on Build GRCh38
rs1020132810 Chr.19: 45350814-45350815 on Build GRCh38
rs3916900 Chr.19: 45350765-45350765 on Build GRCh38
rs747576027 Chr.19: 45350750-45350751 on Build GRCh38
rs954552596 Chr.19: 45350803-45350808 on Build GRCh38
rs778409149 Chr.19: 45350698-45350698 on Build GRCh38
rs753426669 Chr.19: 45350774-45350774 on Build GRCh38
rs775015605 Chr.19: 45350792-45350792 on Build GRCh38
rs754674282 Chr.19: 45350690-45350690 on Build GRCh38
rs199586485 Chr.19: 45350720-45350720 on Build GRCh38
rs780290026 Chr.19: 45350796-45350796 on Build GRCh38
rs45562842 Chr.19: 45350805-45350805 on Build GRCh38
rs762568700 Chr.19: 45350865-45350865 on Build GRCh38
rs749751289 Chr.19: 45350737-45350737 on Build GRCh38
rs149624142 Chr.19: 45350733-45350733 on Build GRCh38
rs3916899 Chr.19: 45350787-45350787 on Build GRCh38
rs374098911 Chr.19: 45350724-45350724 on Build GRCh38
rs569089733 Chr.19: 45350696-45350696 on Build GRCh38
rs374426577 Chr.19: 45350736-45350736 on Build GRCh38
rs904917081 Chr.19: 45350776-45350776 on Build GRCh38
rs755845095 Chr.19: 45350768-45350768 on Build GRCh38
rs779104723 Chr.19: 45350740-45350740 on Build GRCh38
rs746209909 Chr.19: 45350741-45350741 on Build GRCh38
rs749876174 Chr.19: 45350699-45350699 on Build GRCh38
rs774787128 Chr.19: 45350706-45350706 on Build GRCh38
rs773883702 Chr.19: 45350682-45350683 on Build GRCh38
rs368867089 Chr.19: 45350743-45350743 on Build GRCh38
rs386809748 Chr.19: 45350807-45350813 on Build GRCh38
rs758788696 Chr.19: 45350791-45350791 on Build GRCh38
rs766105457 Chr.19: 45350725-45350725 on Build GRCh38
rs200512408 Chr.19: 45350745-45350745 on Build GRCh38
rs771354627 Chr.19: 45350704-45350704 on Build GRCh38

View Full Product Details