Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 745698-745957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN4
Entrez Gene ID
50628
Gene Name
gem nuclear organelle associated protein 4
Gene Aliases
HC56, HCAP1, HHRF-1, p97
UniGene
Hs.499620
Gene Chromosome Location
Chr.17: 744421-752868 on Build GRCh38
Transcripts
NM_015721.2 XM_005256667.4 XM_005256670.4 XM_011523910.2 XM_011523911.2 XM_011523912.2 XM_011523913.2 XM_017024709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449032 c.2137C>T p.Q713* Chr.17: 745906-745906 on Build GRCh38
4748341 c.2105G>A p.C702Y Chr.17: 745938-745938 on Build GRCh38
2797184 c.2187G>A p.A729A Chr.17: 745856-745856 on Build GRCh38
6271753 c.2271C>A p.S757S Chr.17: 745772-745772 on Build GRCh38
3521127 c.2266C>T p.L756L Chr.17: 745777-745777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371266453 Chr.17: 745912-745912 on Build GRCh38
rs757449752 Chr.17: 745897-745897 on Build GRCh38
rs369243436 Chr.17: 745901-745901 on Build GRCh38
rs528626826 Chr.17: 745796-745796 on Build GRCh38
rs371056751 Chr.17: 745847-745847 on Build GRCh38
rs762555788 Chr.17: 745856-745856 on Build GRCh38
rs550331694 Chr.17: 745850-745850 on Build GRCh38
rs753091395 Chr.17: 745761-745761 on Build GRCh38
rs377571129 Chr.17: 745773-745773 on Build GRCh38
rs775968214 Chr.17: 745923-745923 on Build GRCh38
rs759995757 Chr.17: 745832-745832 on Build GRCh38
rs188821606 Chr.17: 745846-745846 on Build GRCh38
rs751140134 Chr.17: 745878-745878 on Build GRCh38
rs201785298 Chr.17: 745770-745770 on Build GRCh38
rs765875076 Chr.17: 745863-745863 on Build GRCh38
rs912813860 Chr.17: 745883-745883 on Build GRCh38
rs201006353 Chr.17: 745903-745903 on Build GRCh38
rs760083435 Chr.17: 745742-745742 on Build GRCh38
rs771083325 Chr.17: 745880-745880 on Build GRCh38
rs963951883 Chr.17: 745735-745735 on Build GRCh38
rs749493927 Chr.17: 745895-745895 on Build GRCh38
rs772588769 Chr.17: 745837-745837 on Build GRCh38
rs769803112 Chr.17: 745762-745762 on Build GRCh38
rs34519093 Chr.17: 745924-745924 on Build GRCh38
rs1053610653 Chr.17: 745727-745727 on Build GRCh38
rs774827354 Chr.17: 745739-745739 on Build GRCh38
rs757918779 Chr.17: 745777-745777 on Build GRCh38
rs754909003 Chr.17: 745879-745879 on Build GRCh38
rs367588554 Chr.17: 745910-745910 on Build GRCh38
rs774453569 Chr.17: 745830-745830 on Build GRCh38
rs368956857 Chr.17: 745887-745887 on Build GRCh38
rs775770182 Chr.17: 745843-745843 on Build GRCh38
rs776892989 Chr.17: 745935-745935 on Build GRCh38
rs781088742 Chr.17: 745798-745798 on Build GRCh38
rs1062923 Chr.17: 745827-745827 on Build GRCh38
rs769841928 Chr.17: 745815-745815 on Build GRCh38
rs1014625499 Chr.17: 745819-745819 on Build GRCh38
rs369869667 Chr.17: 745779-745779 on Build GRCh38
rs780008339 Chr.17: 745899-745900 on Build GRCh38
rs374272851 Chr.17: 745848-745848 on Build GRCh38
rs747971249 Chr.17: 745805-745805 on Build GRCh38
rs762622001 Chr.17: 745937-745937 on Build GRCh38
rs201418426 Chr.17: 745825-745825 on Build GRCh38
rs8078660 Chr.17: 745797-745797 on Build GRCh38
rs866041208 Chr.17: 745852-745852 on Build GRCh38
rs769037135 Chr.17: 745933-745933 on Build GRCh38
rs767834385 Chr.17: 745746-745746 on Build GRCh38
rs372184984 Chr.17: 745888-745888 on Build GRCh38
rs746502473 Chr.17: 745900-745901 on Build GRCh38

View Full Product Details