Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27099269-27099540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGREF1
Entrez Gene ID
10669
Gene Name
cell growth regulator with EF-hand domain 1
Gene Aliases
CGR11
UniGene
Hs.159525
Gene Chromosome Location
Chr.2: 27099353-27119127 on Build GRCh38
Transcripts
NM_001166240.1 NM_001301324.1 NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_017004060.1 XM_017004061.1

Gene Symbol
KHK
Entrez Gene ID
3795
Gene Name
ketohexokinase
Gene Aliases
-
UniGene
Hs.567297
Gene Chromosome Location
Chr.2: 27086738-27100751 on Build GRCh38
Transcripts
NM_001166240.1 NM_001301324.1 NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_017004060.1 XM_017004061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3991239 c.733T>C p.F245L Chr.2: 27099499-27099499 on Build GRCh38
2913166 c.737C>T p.P246L Chr.2: 27099503-27099503 on Build GRCh38
6029783 c.641G>A p.R214H Chr.2: 27099272-27099272 on Build GRCh38
720661 c.694G>T p.A232S Chr.2: 27099460-27099460 on Build GRCh38
6835519 c.723C>G p.H241Q Chr.2: 27099489-27099489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567971261 Chr.2: 27099484-27099484 on Build GRCh38
rs534739226 Chr.2: 27099405-27099405 on Build GRCh38
rs779921285 Chr.2: 27099377-27099377 on Build GRCh38
rs139948811 Chr.2: 27099457-27099457 on Build GRCh38
rs886055890 Chr.2: 27099449-27099451 on Build GRCh38
rs979627678 Chr.2: 27099348-27099348 on Build GRCh38
rs201453373 Chr.2: 27099411-27099411 on Build GRCh38
rs371334415 Chr.2: 27099380-27099380 on Build GRCh38
rs764754040 Chr.2: 27099424-27099424 on Build GRCh38
rs751508802 Chr.2: 27099439-27099439 on Build GRCh38
rs767929279 Chr.2: 27099300-27099300 on Build GRCh38
rs768902999 Chr.2: 27099464-27099464 on Build GRCh38
rs754789918 Chr.2: 27099445-27099445 on Build GRCh38
rs926748212 Chr.2: 27099352-27099352 on Build GRCh38
rs758228868 Chr.2: 27099373-27099373 on Build GRCh38
rs763706345 Chr.2: 27099307-27099307 on Build GRCh38
rs533480314 Chr.2: 27099343-27099343 on Build GRCh38
rs527882092 Chr.2: 27099438-27099438 on Build GRCh38
rs549772974 Chr.2: 27099512-27099512 on Build GRCh38
rs374176560 Chr.2: 27099335-27099335 on Build GRCh38
rs776937292 Chr.2: 27099465-27099465 on Build GRCh38
rs148627840 Chr.2: 27099450-27099450 on Build GRCh38
rs199989985 Chr.2: 27099511-27099511 on Build GRCh38
rs992781636 Chr.2: 27099491-27099491 on Build GRCh38
rs776046472 Chr.2: 27099401-27099401 on Build GRCh38
rs374116984 Chr.2: 27099416-27099416 on Build GRCh38
rs764666583 Chr.2: 27099334-27099334 on Build GRCh38
rs554901179 Chr.2: 27099380-27099380 on Build GRCh38
rs201030559 Chr.2: 27099331-27099331 on Build GRCh38
rs575891124 Chr.2: 27099322-27099322 on Build GRCh38
rs771149929 Chr.2: 27099393-27099393 on Build GRCh38
rs776125990 Chr.2: 27099492-27099492 on Build GRCh38
rs760400912 Chr.2: 27099306-27099306 on Build GRCh38
rs144520325 Chr.2: 27099492-27099492 on Build GRCh38
rs761385266 Chr.2: 27099421-27099421 on Build GRCh38
rs749509415 Chr.2: 27099383-27099383 on Build GRCh38
rs752495277 Chr.2: 27099513-27099513 on Build GRCh38
rs371516742 Chr.2: 27099520-27099520 on Build GRCh38
rs772413859 Chr.2: 27099399-27099399 on Build GRCh38
rs745997707 Chr.2: 27099454-27099454 on Build GRCh38
rs377473990 Chr.2: 27099409-27099409 on Build GRCh38
rs142428157 Chr.2: 27099453-27099453 on Build GRCh38
rs772163501 Chr.2: 27099456-27099456 on Build GRCh38
rs757902561 Chr.2: 27099436-27099436 on Build GRCh38
rs751466422 Chr.2: 27099504-27099504 on Build GRCh38
rs781078287 Chr.2: 27099449-27099449 on Build GRCh38
rs878943136 Chr.2: 27099354-27099354 on Build GRCh38
rs761476412 Chr.2: 27099481-27099481 on Build GRCh38
rs747489398 Chr.2: 27099460-27099460 on Build GRCh38
rs374688541 Chr.2: 27099408-27099408 on Build GRCh38
rs965708116 Chr.2: 27099330-27099330 on Build GRCh38
rs368056714 Chr.2: 27099503-27099503 on Build GRCh38
rs968247460 Chr.2: 27099344-27099344 on Build GRCh38
rs897749184 Chr.2: 27099441-27099441 on Build GRCh38
rs959944601 Chr.2: 27099459-27099459 on Build GRCh38
rs772819437 Chr.2: 27099486-27099486 on Build GRCh38
rs749887208 Chr.2: 27099429-27099429 on Build GRCh38
rs369037689 Chr.2: 27099292-27099292 on Build GRCh38
rs1030915687 Chr.2: 27099385-27099385 on Build GRCh38
rs980080107 Chr.2: 27099440-27099440 on Build GRCh38
rs746979459 Chr.2: 27099396-27099397 on Build GRCh38
rs182367966 Chr.2: 27099379-27099379 on Build GRCh38
rs755955022 Chr.2: 27099514-27099514 on Build GRCh38
rs746413451 Chr.2: 27099394-27099394 on Build GRCh38
rs752967925 Chr.2: 27099303-27099303 on Build GRCh38
rs745546639 Chr.2: 27099445-27099447 on Build GRCh38

View Full Product Details