Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41715751-41715980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAST
Entrez Gene ID
2520
Gene Name
gastrin
Gene Aliases
GAS
UniGene
Hs.2681
Gene Chromosome Location
Chr.17: 41712326-41715969 on Build GRCh38
Transcripts
NM_000805.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130053 c.242A>G p.E81G Chr.17: 41715808-41715808 on Build GRCh38
6669520 c.244_246delGAA p.E85delE Chr.17: 41715810-41715812 on Build GRCh38
6843490 c.223A>G p.K75E Chr.17: 41715789-41715789 on Build GRCh38
1630191 c.289G>A p.A97T Chr.17: 41715855-41715855 on Build GRCh38
4536428 c.229G>A p.G77R Chr.17: 41715795-41715795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782023925 Chr.17: 41715820-41715820 on Build GRCh38
rs570450689 Chr.17: 41715828-41715828 on Build GRCh38
rs782698420 Chr.17: 41715955-41715955 on Build GRCh38
rs575117294 Chr.17: 41715858-41715858 on Build GRCh38
rs781902893 Chr.17: 41715850-41715850 on Build GRCh38
rs896374794 Chr.17: 41715892-41715892 on Build GRCh38
rs782224974 Chr.17: 41715801-41715801 on Build GRCh38
rs375174671 Chr.17: 41715832-41715832 on Build GRCh38
rs782345532 Chr.17: 41715919-41715919 on Build GRCh38
rs144173344 Chr.17: 41715781-41715781 on Build GRCh38
rs551846521 Chr.17: 41715827-41715827 on Build GRCh38
rs150206234 Chr.17: 41715771-41715771 on Build GRCh38
rs781950455 Chr.17: 41715800-41715800 on Build GRCh38
rs368684117 Chr.17: 41715920-41715920 on Build GRCh38
rs375354857 Chr.17: 41715882-41715882 on Build GRCh38
rs782538505 Chr.17: 41715945-41715945 on Build GRCh38
rs535385347 Chr.17: 41715830-41715830 on Build GRCh38
rs4290525 Chr.17: 41715774-41715774 on Build GRCh38
rs782576189 Chr.17: 41715887-41715887 on Build GRCh38
rs781866901 Chr.17: 41715809-41715810 on Build GRCh38
rs782188604 Chr.17: 41715906-41715906 on Build GRCh38
rs781979891 Chr.17: 41715921-41715921 on Build GRCh38
rs782170362 Chr.17: 41715822-41715822 on Build GRCh38
rs371500519 Chr.17: 41715782-41715782 on Build GRCh38
rs782768915 Chr.17: 41715846-41715846 on Build GRCh38
rs377220004 Chr.17: 41715770-41715770 on Build GRCh38
rs535647893 Chr.17: 41715927-41715927 on Build GRCh38
rs1012147954 Chr.17: 41715902-41715902 on Build GRCh38
rs782304560 Chr.17: 41715813-41715813 on Build GRCh38
rs782782523 Chr.17: 41715847-41715847 on Build GRCh38
rs782478288 Chr.17: 41715870-41715870 on Build GRCh38
rs782469519 Chr.17: 41715843-41715843 on Build GRCh38
rs782437955 Chr.17: 41715914-41715914 on Build GRCh38
rs782645115 Chr.17: 41715867-41715867 on Build GRCh38

View Full Product Details