Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 30728261-30728486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GADL1
Entrez Gene ID
339896
Gene Name
glutamate decarboxylase like 1
Gene Aliases
ADC, CSADC, HuADC, HuCSADC
UniGene
Hs.657052
Gene Chromosome Location
Chr.3: 30726009-30894661 on Build GRCh38
Transcripts
NM_207359.2 XM_017006297.1 XM_017006298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5708677 c.1546T>G p.L516V Chr.3: 30728262-30728262 on Build GRCh38
7125605 c.867G>A p.M289I Chr.3: 30728389-30728389 on Build GRCh38
4526673 c.1410G>A p.K470K Chr.3: 30728398-30728398 on Build GRCh38
4527168 c.1445G>A p.G482D Chr.3: 30728363-30728363 on Build GRCh38
3592349 c.1461G>A p.R487R Chr.3: 30728347-30728347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752508984 Chr.3: 30728413-30728413 on Build GRCh38
rs764024313 Chr.3: 30728428-30728428 on Build GRCh38
rs957194369 Chr.3: 30728412-30728412 on Build GRCh38
rs747341770 Chr.3: 30728364-30728364 on Build GRCh38
rs752670417 Chr.3: 30728341-30728341 on Build GRCh38
rs368666232 Chr.3: 30728408-30728408 on Build GRCh38
rs62636628 Chr.3: 30728327-30728327 on Build GRCh38
rs753717178 Chr.3: 30728434-30728434 on Build GRCh38
rs762017378 Chr.3: 30728383-30728383 on Build GRCh38
rs999902870 Chr.3: 30728320-30728320 on Build GRCh38
rs746250558 Chr.3: 30728298-30728298 on Build GRCh38
rs772320163 Chr.3: 30728301-30728301 on Build GRCh38
rs111590277 Chr.3: 30728340-30728340 on Build GRCh38
rs769312015 Chr.3: 30728371-30728371 on Build GRCh38
rs929541759 Chr.3: 30728432-30728432 on Build GRCh38
rs7617829 Chr.3: 30728456-30728456 on Build GRCh38
rs763699450 Chr.3: 30728460-30728460 on Build GRCh38
rs756657458 Chr.3: 30728349-30728349 on Build GRCh38
rs1004139181 Chr.3: 30728351-30728351 on Build GRCh38
rs148714643 Chr.3: 30728379-30728379 on Build GRCh38
rs201974837 Chr.3: 30728388-30728388 on Build GRCh38
rs776197454 Chr.3: 30728305-30728305 on Build GRCh38
rs745494042 Chr.3: 30728354-30728354 on Build GRCh38
rs750604147 Chr.3: 30728328-30728328 on Build GRCh38
rs899677985 Chr.3: 30728299-30728299 on Build GRCh38
rs144252169 Chr.3: 30728355-30728355 on Build GRCh38
rs770638457 Chr.3: 30728295-30728295 on Build GRCh38
rs112411488 Chr.3: 30728416-30728416 on Build GRCh38
rs755772521 Chr.3: 30728459-30728460 on Build GRCh38
rs376871595 Chr.3: 30728353-30728353 on Build GRCh38
rs867632772 Chr.3: 30728345-30728345 on Build GRCh38
rs777233618 Chr.3: 30728288-30728288 on Build GRCh38
rs755844299 Chr.3: 30728415-30728415 on Build GRCh38
rs973459694 Chr.3: 30728399-30728399 on Build GRCh38
rs967232899 Chr.3: 30728340-30728340 on Build GRCh38
rs909535132 Chr.3: 30728441-30728441 on Build GRCh38
rs777637585 Chr.3: 30728342-30728342 on Build GRCh38
rs779808482 Chr.3: 30728444-30728444 on Build GRCh38
rs998402965 Chr.3: 30728302-30728302 on Build GRCh38
rs764313488 Chr.3: 30728315-30728315 on Build GRCh38
rs756849041 Chr.3: 30728437-30728437 on Build GRCh38
rs12054099 Chr.3: 30728439-30728439 on Build GRCh38
rs986467165 Chr.3: 30728426-30728426 on Build GRCh38
rs765215969 Chr.3: 30728387-30728387 on Build GRCh38
rs761245778 Chr.3: 30728310-30728310 on Build GRCh38
rs992028065 Chr.3: 30728422-30728422 on Build GRCh38
rs758158852 Chr.3: 30728442-30728442 on Build GRCh38
rs754856307 Chr.3: 30728336-30728336 on Build GRCh38
rs755666784 Chr.3: 30728287-30728287 on Build GRCh38
rs776884931 Chr.3: 30728321-30728321 on Build GRCh38
rs370176334 Chr.3: 30728297-30728297 on Build GRCh38
rs775969862 Chr.3: 30728361-30728361 on Build GRCh38
rs769134341 Chr.3: 30728458-30728458 on Build GRCh38
rs867269290 Chr.3: 30728343-30728343 on Build GRCh38
rs372035518 Chr.3: 30728326-30728326 on Build GRCh38
rs748060157 Chr.3: 30728286-30728286 on Build GRCh38
rs748704610 Chr.3: 30728290-30728290 on Build GRCh38
rs958836362 Chr.3: 30728420-30728420 on Build GRCh38
rs62636627 Chr.3: 30728348-30728348 on Build GRCh38

View Full Product Details