Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59618531-59618780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT3
Entrez Gene ID
9245
Gene Name
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases
C2/4GnT, C24GNT, C2GNT2, C2GNTM, GNTM
UniGene
Hs.194710
Gene Chromosome Location
Chr.15: 59611783-59620011 on Build GRCh38
Transcripts
NM_004751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502818 c.411G>A p.M137I Chr.15: 59618649-59618649 on Build GRCh38
3502819 c.520G>A p.E174K Chr.15: 59618758-59618758 on Build GRCh38
3794276 c.372G>A p.L124L Chr.15: 59618610-59618610 on Build GRCh38
6502030 c.542C>A p.S181Y Chr.15: 59618780-59618780 on Build GRCh38
4492272 c.394C>T p.P132S Chr.15: 59618632-59618632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370387567 Chr.15: 59618561-59618561 on Build GRCh38
rs757226378 Chr.15: 59618706-59618706 on Build GRCh38
rs368540018 Chr.15: 59618556-59618556 on Build GRCh38
rs769563495 Chr.15: 59618610-59618610 on Build GRCh38
rs770418753 Chr.15: 59618646-59618646 on Build GRCh38
rs944242926 Chr.15: 59618676-59618676 on Build GRCh38
rs896399026 Chr.15: 59618747-59618747 on Build GRCh38
rs746705657 Chr.15: 59618733-59618733 on Build GRCh38
rs762187773 Chr.15: 59618576-59618576 on Build GRCh38
rs374984289 Chr.15: 59618658-59618658 on Build GRCh38
rs139614046 Chr.15: 59618689-59618689 on Build GRCh38
rs138908979 Chr.15: 59618575-59618575 on Build GRCh38
rs903552329 Chr.15: 59618727-59618727 on Build GRCh38
rs371933037 Chr.15: 59618732-59618732 on Build GRCh38
rs150011598 Chr.15: 59618717-59618717 on Build GRCh38
rs781493665 Chr.15: 59618697-59618697 on Build GRCh38
rs748004202 Chr.15: 59618608-59618608 on Build GRCh38
rs373801321 Chr.15: 59618565-59618565 on Build GRCh38
rs755937518 Chr.15: 59618698-59618698 on Build GRCh38
rs749201782 Chr.15: 59618702-59618702 on Build GRCh38
rs773992361 Chr.15: 59618553-59618553 on Build GRCh38
rs368222436 Chr.15: 59618703-59618703 on Build GRCh38
rs762250026 Chr.15: 59618666-59618666 on Build GRCh38
rs371624326 Chr.15: 59618643-59618643 on Build GRCh38
rs780346765 Chr.15: 59618597-59618597 on Build GRCh38
rs764421147 Chr.15: 59618574-59618574 on Build GRCh38
rs745406077 Chr.15: 59618720-59618720 on Build GRCh38
rs750935752 Chr.15: 59618679-59618679 on Build GRCh38
rs771239323 Chr.15: 59618654-59618654 on Build GRCh38
rs771565960 Chr.15: 59618721-59618721 on Build GRCh38
rs759157624 Chr.15: 59618648-59618648 on Build GRCh38
rs369412992 Chr.15: 59618738-59618738 on Build GRCh38
rs776810516 Chr.15: 59618665-59618665 on Build GRCh38
rs771745249 Chr.15: 59618649-59618649 on Build GRCh38
rs937679417 Chr.15: 59618728-59618728 on Build GRCh38
rs1055483480 Chr.15: 59618734-59618734 on Build GRCh38
rs750410596 Chr.15: 59618586-59618586 on Build GRCh38
rs780817051 Chr.15: 59618607-59618607 on Build GRCh38
rs773494709 Chr.15: 59618741-59618741 on Build GRCh38
rs777773712 Chr.15: 59618629-59618629 on Build GRCh38
rs755296336 Chr.15: 59618605-59618605 on Build GRCh38
rs777621588 Chr.15: 59618739-59618739 on Build GRCh38
rs148283440 Chr.15: 59618686-59618686 on Build GRCh38
rs751814891 Chr.15: 59618600-59618600 on Build GRCh38
rs765596866 Chr.15: 59618675-59618675 on Build GRCh38
rs758556052 Chr.15: 59618595-59618595 on Build GRCh38
rs763609698 Chr.15: 59618681-59618681 on Build GRCh38
rs765795133 Chr.15: 59618579-59618579 on Build GRCh38
rs755208383 Chr.15: 59618690-59618690 on Build GRCh38

View Full Product Details