Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76081743-76082016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC7
Entrez Gene ID
23265
Gene Name
exocyst complex component 7
Gene Aliases
2-5-3p, EX070, EXO70, EXOC1, Exo70p, YJL085W
UniGene
Hs.514496
Gene Chromosome Location
Chr.17: 76080993-76104221 on Build GRCh38
Transcripts
NM_001013839.3 NM_001145297.3 NM_001145298.3 NM_001145299.3 NM_001282313.1 NM_015219.4 XM_006721786.3 XM_006721787.3 XM_006721788.3 XM_006721789.3 XM_011524571.2 XM_017024397.1 NM_180990.3

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
UniGene
Hs.714919
Gene Chromosome Location
Chr.17: 76079182-76082804 on Build GRCh38
Transcripts
NM_001013839.3 NM_001145297.3 NM_001145298.3 NM_001145299.3 NM_001282313.1 NM_015219.4 XM_006721786.3 XM_006721787.3 XM_006721788.3 XM_006721789.3 XM_011524571.2 XM_017024397.1 NM_180990.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5768349 c.891C>T p.F297F Chr.17: 76081892-76081892 on Build GRCh38
1684531 c.926_927delTA p.I309fs*40 Chr.17: 76081927-76081928 on Build GRCh38
4069803 c.959G>A p.R320Q Chr.17: 76081960-76081960 on Build GRCh38
5768350 c.879C>T p.L293L Chr.17: 76081754-76081754 on Build GRCh38
349446 c.996C>A p.A332A Chr.17: 76081997-76081997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142424750 Chr.17: 76081905-76081905 on Build GRCh38
rs139224066 Chr.17: 76081979-76081979 on Build GRCh38
rs770932504 Chr.17: 76081867-76081867 on Build GRCh38
rs367895945 Chr.17: 76081892-76081892 on Build GRCh38
rs374914758 Chr.17: 76081871-76081871 on Build GRCh38
rs765160550 Chr.17: 76081879-76081880 on Build GRCh38
rs978006409 Chr.17: 76081801-76081801 on Build GRCh38
rs746784791 Chr.17: 76081847-76081847 on Build GRCh38
rs564705824 Chr.17: 76081982-76081982 on Build GRCh38
rs1049520943 Chr.17: 76081831-76081831 on Build GRCh38
rs112045875 Chr.17: 76081967-76081967 on Build GRCh38
rs756747853 Chr.17: 76081782-76081782 on Build GRCh38
rs766411667 Chr.17: 76081974-76081989 on Build GRCh38
rs779883597 Chr.17: 76081800-76081800 on Build GRCh38
rs750555414 Chr.17: 76081820-76081820 on Build GRCh38
rs925182032 Chr.17: 76081819-76081819 on Build GRCh38
rs749479934 Chr.17: 76081959-76081959 on Build GRCh38
rs749481557 Chr.17: 76081854-76081854 on Build GRCh38
rs749838124 Chr.17: 76081999-76081999 on Build GRCh38
rs776172135 Chr.17: 76081978-76081978 on Build GRCh38
rs773210527 Chr.17: 76081886-76081886 on Build GRCh38
rs576164907 Chr.17: 76081921-76081921 on Build GRCh38
rs761431638 Chr.17: 76081983-76081983 on Build GRCh38
rs758317028 Chr.17: 76081907-76081907 on Build GRCh38
rs371410673 Chr.17: 76081868-76081868 on Build GRCh38
rs374625380 Chr.17: 76081796-76081796 on Build GRCh38
rs545103310 Chr.17: 76081960-76081960 on Build GRCh38
rs751463301 Chr.17: 76081935-76081935 on Build GRCh38
rs371765385 Chr.17: 76081958-76081958 on Build GRCh38
rs761090443 Chr.17: 76081880-76081880 on Build GRCh38
rs761978685 Chr.17: 76081873-76081874 on Build GRCh38
rs762853986 Chr.17: 76081928-76081929 on Build GRCh38
rs763816558 Chr.17: 76081766-76081766 on Build GRCh38
rs367697538 Chr.17: 76081797-76081797 on Build GRCh38
rs776041658 Chr.17: 76081877-76081877 on Build GRCh38
rs745934049 Chr.17: 76081969-76081969 on Build GRCh38
rs111935487 Chr.17: 76081939-76081939 on Build GRCh38
rs754793577 Chr.17: 76081946-76081946 on Build GRCh38
rs764722869 Chr.17: 76081901-76081901 on Build GRCh38
rs776816026 Chr.17: 76081788-76081788 on Build GRCh38
rs753438847 Chr.17: 76081770-76081770 on Build GRCh38
rs772741101 Chr.17: 76081975-76081975 on Build GRCh38
rs757934380 Chr.17: 76081996-76081996 on Build GRCh38
rs371392278 Chr.17: 76081783-76081783 on Build GRCh38
rs770044149 Chr.17: 76081853-76081853 on Build GRCh38
rs753565753 Chr.17: 76081889-76081889 on Build GRCh38
rs772689861 Chr.17: 76081876-76081876 on Build GRCh38

View Full Product Details