Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81541082-81541353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
UniGene
Hs.313905
Gene Chromosome Location
Chr.17: 81539885-81552403 on Build GRCh38
Transcripts
NM_025161.5 NR_033338.1 XM_006722111.2 XM_006722112.3 XM_017025162.1 XM_017025163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415853 c.2470G>A p.D824N Chr.17: 81541353-81541353 on Build GRCh38
4667078 c.2476C>T p.R826C Chr.17: 81541347-81541347 on Build GRCh38
7246390 c.2512G>A p.E838K Chr.17: 81541311-81541311 on Build GRCh38
6415854 c.2017G>A p.D673N Chr.17: 81541353-81541353 on Build GRCh38
4667079 c.2023C>T p.R675C Chr.17: 81541347-81541347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925715402 Chr.17: 81541188-81541188 on Build GRCh38
rs148073755 Chr.17: 81541312-81541312 on Build GRCh38
rs751256452 Chr.17: 81541299-81541299 on Build GRCh38
rs568043485 Chr.17: 81541130-81541130 on Build GRCh38
rs781115159 Chr.17: 81541308-81541308 on Build GRCh38
rs748544057 Chr.17: 81541111-81541111 on Build GRCh38
rs945133173 Chr.17: 81541104-81541104 on Build GRCh38
rs541057203 Chr.17: 81541243-81541243 on Build GRCh38
rs374378717 Chr.17: 81541298-81541298 on Build GRCh38
rs559340182 Chr.17: 81541245-81541245 on Build GRCh38
rs1029706629 Chr.17: 81541165-81541165 on Build GRCh38
rs1042897491 Chr.17: 81541178-81541178 on Build GRCh38
rs916222691 Chr.17: 81541328-81541328 on Build GRCh38
rs78121521 Chr.17: 81541242-81541242 on Build GRCh38
rs1005904360 Chr.17: 81541147-81541147 on Build GRCh38
rs576566123 Chr.17: 81541203-81541203 on Build GRCh38
rs540152603 Chr.17: 81541184-81541184 on Build GRCh38
rs891209644 Chr.17: 81541254-81541254 on Build GRCh38
rs766943366 Chr.17: 81541290-81541290 on Build GRCh38
rs762099126 Chr.17: 81541263-81541263 on Build GRCh38
rs747709321 Chr.17: 81541116-81541116 on Build GRCh38
rs927689341 Chr.17: 81541223-81541223 on Build GRCh38
rs759185060 Chr.17: 81541302-81541302 on Build GRCh38
rs965373269 Chr.17: 81541143-81541143 on Build GRCh38
rs1029485964 Chr.17: 81541252-81541252 on Build GRCh38
rs1011791686 Chr.17: 81541284-81541284 on Build GRCh38
rs764123117 Chr.17: 81541331-81541331 on Build GRCh38
rs111228182 Chr.17: 81541202-81541202 on Build GRCh38
rs370590915 Chr.17: 81541311-81541311 on Build GRCh38
rs1006953304 Chr.17: 81541255-81541255 on Build GRCh38
rs1038713875 Chr.17: 81541244-81541244 on Build GRCh38
rs773622535 Chr.17: 81541283-81541283 on Build GRCh38
rs768785790 Chr.17: 81541320-81541320 on Build GRCh38
rs189067432 Chr.17: 81541128-81541128 on Build GRCh38
rs184096034 Chr.17: 81541159-81541159 on Build GRCh38
rs536959418 Chr.17: 81541187-81541187 on Build GRCh38
rs115362207 Chr.17: 81541248-81541248 on Build GRCh38
rs1027060594 Chr.17: 81541127-81541127 on Build GRCh38
rs181373937 Chr.17: 81541151-81541151 on Build GRCh38
rs955342946 Chr.17: 81541177-81541177 on Build GRCh38
rs768708470 Chr.17: 81541259-81541259 on Build GRCh38
rs1002357462 Chr.17: 81541235-81541235 on Build GRCh38
rs939005963 Chr.17: 81541122-81541122 on Build GRCh38
rs952787020 Chr.17: 81541127-81541129 on Build GRCh38
rs566403767 Chr.17: 81541164-81541164 on Build GRCh38
rs913375694 Chr.17: 81541197-81541197 on Build GRCh38
rs977801663 Chr.17: 81541221-81541221 on Build GRCh38
rs906159078 Chr.17: 81541117-81541117 on Build GRCh38
rs990836131 Chr.17: 81541195-81541195 on Build GRCh38
rs752101053 Chr.17: 81541291-81541291 on Build GRCh38
rs564258733 Chr.17: 81541262-81541262 on Build GRCh38
rs562164212 Chr.17: 81541193-81541193 on Build GRCh38
rs772327939 Chr.17: 81541332-81541332 on Build GRCh38
rs374838754 Chr.17: 81541258-81541258 on Build GRCh38
rs543896583 Chr.17: 81541229-81541229 on Build GRCh38
rs760772141 Chr.17: 81541321-81541321 on Build GRCh38
rs376222537 Chr.17: 81541206-81541206 on Build GRCh38
rs62076028 Chr.17: 81541237-81541237 on Build GRCh38
rs772057325 Chr.17: 81541309-81541309 on Build GRCh38
rs761146292 Chr.17: 81541289-81541289 on Build GRCh38
rs140994319 Chr.17: 81541315-81541315 on Build GRCh38
rs957626235 Chr.17: 81541194-81541194 on Build GRCh38
rs772421342 Chr.17: 81541265-81541265 on Build GRCh38

View Full Product Details