Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105524104-105524365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001243283.1 NM_001627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035935 c.202A>G p.I68V Chr.3: 105524316-105524316 on Build GRCh38
1182456 c.205G>A p.A69T Chr.3: 105524319-105524319 on Build GRCh38
3780616 c.212G>C p.R71T Chr.3: 105524326-105524326 on Build GRCh38
6504121 c.188G>A p.G63D Chr.3: 105524302-105524302 on Build GRCh38
4976109 c.243C>A p.D81E Chr.3: 105524357-105524357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774445203 Chr.3: 105524316-105524316 on Build GRCh38
rs369753930 Chr.3: 105524267-105524267 on Build GRCh38
rs941280732 Chr.3: 105524187-105524187 on Build GRCh38
rs1038185056 Chr.3: 105524209-105524209 on Build GRCh38
rs758659906 Chr.3: 105524326-105524326 on Build GRCh38
rs994043994 Chr.3: 105524226-105524226 on Build GRCh38
rs747372908 Chr.3: 105524333-105524333 on Build GRCh38
rs746416911 Chr.3: 105524344-105524344 on Build GRCh38
rs896993834 Chr.3: 105524216-105524216 on Build GRCh38
rs1050453377 Chr.3: 105524171-105524171 on Build GRCh38
rs751401355 Chr.3: 105524294-105524294 on Build GRCh38
rs375591933 Chr.3: 105524252-105524252 on Build GRCh38
rs185727605 Chr.3: 105524259-105524259 on Build GRCh38
rs552028173 Chr.3: 105524175-105524175 on Build GRCh38
rs867152890 Chr.3: 105524174-105524174 on Build GRCh38
rs530626148 Chr.3: 105524173-105524173 on Build GRCh38
rs373451274 Chr.3: 105524273-105524273 on Build GRCh38
rs778045642 Chr.3: 105524328-105524328 on Build GRCh38
rs895126374 Chr.3: 105524257-105524257 on Build GRCh38
rs750432365 Chr.3: 105524304-105524304 on Build GRCh38
rs779082795 Chr.3: 105524321-105524321 on Build GRCh38
rs777192728 Chr.3: 105524239-105524239 on Build GRCh38
rs1033977551 Chr.3: 105524296-105524296 on Build GRCh38
rs763734869 Chr.3: 105524283-105524283 on Build GRCh38
rs931966175 Chr.3: 105524168-105524168 on Build GRCh38
rs771346909 Chr.3: 105524341-105524341 on Build GRCh38
rs893995198 Chr.3: 105524147-105524147 on Build GRCh38
rs766553586 Chr.3: 105524297-105524297 on Build GRCh38
rs773823020 Chr.3: 105524146-105524146 on Build GRCh38
rs767548203 Chr.3: 105524298-105524298 on Build GRCh38
rs777102845 Chr.3: 105524343-105524343 on Build GRCh38
rs1003750522 Chr.3: 105524218-105524218 on Build GRCh38
rs528045605 Chr.3: 105524324-105524324 on Build GRCh38
rs764854205 Chr.3: 105524266-105524266 on Build GRCh38
rs1012255331 Chr.3: 105524317-105524317 on Build GRCh38

View Full Product Details