Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53951752-53952003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP4
Entrez Gene ID
652
Gene Name
bone morphogenetic protein 4
Gene Aliases
BMP2B, BMP2B1, MCOPS6, OFC11, ZYME
UniGene
Hs.68879
Gene Chromosome Location
Chr.14: 53949736-53956862 on Build GRCh38
Transcripts
NM_001202.4 NM_130850.3 XM_005268015.4 XM_017021603.1 XM_017021604.1 XM_017021605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4591319 c.340G>T p.A114S Chr.14: 53951883-53951883 on Build GRCh38
4972817 c.307G>A p.G103S Chr.14: 53951916-53951916 on Build GRCh38
6564508 c.280G>A p.E94K Chr.14: 53951943-53951943 on Build GRCh38
358845 c.251G>T p.R84L Chr.14: 53951972-53951972 on Build GRCh38
7357590 c.228T>A p.S76R Chr.14: 53951995-53951995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374418188 Chr.14: 53951796-53951796 on Build GRCh38
rs771098397 Chr.14: 53951823-53951823 on Build GRCh38
rs993370951 Chr.14: 53951808-53951808 on Build GRCh38
rs747238009 Chr.14: 53951883-53951883 on Build GRCh38
rs976229577 Chr.14: 53951853-53951853 on Build GRCh38
rs761051715 Chr.14: 53951860-53951860 on Build GRCh38
rs757845313 Chr.14: 53951809-53951809 on Build GRCh38
rs374781046 Chr.14: 53951833-53951833 on Build GRCh38
rs549252141 Chr.14: 53951885-53951885 on Build GRCh38
rs749321751 Chr.14: 53951817-53951817 on Build GRCh38
rs562412111 Chr.14: 53951802-53951802 on Build GRCh38
rs748028769 Chr.14: 53951893-53951893 on Build GRCh38
rs768455462 Chr.14: 53951829-53951829 on Build GRCh38
rs121912767 Chr.14: 53951951-53951951 on Build GRCh38
rs560378369 Chr.14: 53951844-53951844 on Build GRCh38
rs766091993 Chr.14: 53951856-53951856 on Build GRCh38
rs750017710 Chr.14: 53951774-53951774 on Build GRCh38
rs147931520 Chr.14: 53951959-53951959 on Build GRCh38
rs751697119 Chr.14: 53951963-53951963 on Build GRCh38
rs886050541 Chr.14: 53951935-53951935 on Build GRCh38
rs751266792 Chr.14: 53951857-53951857 on Build GRCh38
rs200488679 Chr.14: 53951958-53951958 on Build GRCh38
rs747861439 Chr.14: 53951983-53951983 on Build GRCh38
rs143687498 Chr.14: 53951878-53951878 on Build GRCh38
rs759247214 Chr.14: 53951911-53951911 on Build GRCh38
rs909451835 Chr.14: 53951875-53951875 on Build GRCh38
rs774069849 Chr.14: 53951908-53951908 on Build GRCh38
rs757977627 Chr.14: 53951778-53951778 on Build GRCh38
rs772152954 Chr.14: 53951824-53951824 on Build GRCh38
rs376960358 Chr.14: 53951861-53951861 on Build GRCh38
rs967937256 Chr.14: 53951872-53951872 on Build GRCh38
rs772578892 Chr.14: 53951837-53951837 on Build GRCh38
rs758526357 Chr.14: 53951877-53951877 on Build GRCh38
rs368426865 Chr.14: 53951841-53951841 on Build GRCh38
rs150718090 Chr.14: 53951791-53951792 on Build GRCh38
rs376124404 Chr.14: 53951811-53951811 on Build GRCh38
rs544919516 Chr.14: 53951830-53951830 on Build GRCh38
rs1011657927 Chr.14: 53951984-53951984 on Build GRCh38
rs991708313 Chr.14: 53951894-53951894 on Build GRCh38
rs766109101 Chr.14: 53951794-53951794 on Build GRCh38
rs770567411 Chr.14: 53951902-53951902 on Build GRCh38
rs765611781 Chr.14: 53951947-53951947 on Build GRCh38
rs746008809 Chr.14: 53951815-53951815 on Build GRCh38
rs377676306 Chr.14: 53951967-53951967 on Build GRCh38
rs746756115 Chr.14: 53951827-53951827 on Build GRCh38
rs757681410 Chr.14: 53951866-53951866 on Build GRCh38
rs778363960 Chr.14: 53951804-53951804 on Build GRCh38
rs202159001 Chr.14: 53951919-53951919 on Build GRCh38
rs868713706 Chr.14: 53951955-53951955 on Build GRCh38
rs121912765 Chr.14: 53951945-53951945 on Build GRCh38
rs371239780 Chr.14: 53951918-53951918 on Build GRCh38
rs527591537 Chr.14: 53951874-53951874 on Build GRCh38
rs749814916 Chr.14: 53951806-53951806 on Build GRCh38
rs750427266 Chr.14: 53951876-53951876 on Build GRCh38
rs754732494 Chr.14: 53951838-53951838 on Build GRCh38
rs754308298 Chr.14: 53951862-53951862 on Build GRCh38
rs773235106 Chr.14: 53951897-53951897 on Build GRCh38
rs781435238 Chr.14: 53951971-53951971 on Build GRCh38
rs72680532 Chr.14: 53951825-53951825 on Build GRCh38
rs919225259 Chr.14: 53951859-53951859 on Build GRCh38

View Full Product Details