Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 129106424-129106649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf33
Entrez Gene ID
132321
Gene Name
chromosome 4 open reading frame 33
Gene Aliases
-
UniGene
Hs.567679
Gene Chromosome Location
Chr.4: 129093674-129112688 on Build GRCh38
Transcripts
NM_001099783.1 NM_173487.2 XM_005262734.4 XM_017007722.1 XM_017007723.1 XM_017007724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667777 c.216G>T p.E72D Chr.4: 129106621-129106621 on Build GRCh38
329200 c.231T>A p.V77V Chr.4: 129106636-129106636 on Build GRCh38
5617202 c.201G>A p.L67L Chr.4: 129106606-129106606 on Build GRCh38
6235120 c.190G>A p.A64T Chr.4: 129106595-129106595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879537515 Chr.4: 129106535-129106535 on Build GRCh38
rs958095458 Chr.4: 129106498-129106498 on Build GRCh38
rs775156266 Chr.4: 129106546-129106546 on Build GRCh38
rs150207764 Chr.4: 129106594-129106594 on Build GRCh38
rs915285198 Chr.4: 129106526-129106529 on Build GRCh38
rs576800183 Chr.4: 129106482-129106482 on Build GRCh38
rs755910234 Chr.4: 129106597-129106597 on Build GRCh38
rs544058507 Chr.4: 129106502-129106502 on Build GRCh38
rs986197818 Chr.4: 129106476-129106476 on Build GRCh38
rs369388668 Chr.4: 129106559-129106559 on Build GRCh38
rs762220608 Chr.4: 129106569-129106569 on Build GRCh38
rs558214063 Chr.4: 129106455-129106455 on Build GRCh38
rs758542700 Chr.4: 129106595-129106595 on Build GRCh38
rs751849665 Chr.4: 129106596-129106596 on Build GRCh38
rs764456026 Chr.4: 129106564-129106564 on Build GRCh38
rs138803726 Chr.4: 129106608-129106608 on Build GRCh38
rs149388613 Chr.4: 129106610-129106610 on Build GRCh38
rs988793095 Chr.4: 129106521-129106521 on Build GRCh38
rs771683972 Chr.4: 129106538-129106538 on Build GRCh38
rs1032287193 Chr.4: 129106480-129106480 on Build GRCh38
rs778950276 Chr.4: 129106613-129106613 on Build GRCh38
rs914200720 Chr.4: 129106528-129106528 on Build GRCh38
rs757121662 Chr.4: 129106612-129106612 on Build GRCh38
rs926789878 Chr.4: 129106599-129106599 on Build GRCh38
rs765829874 Chr.4: 129106587-129106587 on Build GRCh38

View Full Product Details