Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25680065-25680294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
UniGene
Hs.638517
Gene Chromosome Location
Chr.15: 25672241-25865144 on Build GRCh38
Transcripts
NM_024490.3 XM_005268261.4 XM_011521826.2 XM_011521828.2 XM_011521829.2 XM_011521830.2 XM_017022436.1 XM_017022437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1678306 c.3789G>A p.W1263* Chr.15: 25680198-25680198 on Build GRCh38
4664130 c.3765C>A p.C1255* Chr.15: 25680222-25680222 on Build GRCh38
1678305 c.3808G>T p.G1270C Chr.15: 25680179-25680179 on Build GRCh38
1640198 c.3849C>T p.V1283V Chr.15: 25680138-25680138 on Build GRCh38
959986 c.3749C>T p.A1250V Chr.15: 25680238-25680238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748429470 Chr.15: 25680215-25680215 on Build GRCh38
rs746418207 Chr.15: 25680107-25680107 on Build GRCh38
rs866987134 Chr.15: 25680199-25680199 on Build GRCh38
rs778707138 Chr.15: 25680086-25680086 on Build GRCh38
rs756744281 Chr.15: 25680248-25680248 on Build GRCh38
rs778760652 Chr.15: 25680180-25680180 on Build GRCh38
rs994427100 Chr.15: 25680095-25680095 on Build GRCh38
rs149423703 Chr.15: 25680237-25680237 on Build GRCh38
rs766358448 Chr.15: 25680158-25680158 on Build GRCh38
rs1033065124 Chr.15: 25680143-25680143 on Build GRCh38
rs557178517 Chr.15: 25680116-25680116 on Build GRCh38
rs571098357 Chr.15: 25680266-25680266 on Build GRCh38
rs751146944 Chr.15: 25680244-25680244 on Build GRCh38
rs373211392 Chr.15: 25680111-25680111 on Build GRCh38
rs371380863 Chr.15: 25680220-25680220 on Build GRCh38
rs765870814 Chr.15: 25680205-25680205 on Build GRCh38
rs200589348 Chr.15: 25680226-25680226 on Build GRCh38
rs764554660 Chr.15: 25680100-25680100 on Build GRCh38
rs376938746 Chr.15: 25680182-25680182 on Build GRCh38
rs777367412 Chr.15: 25680187-25680187 on Build GRCh38
rs146387652 Chr.15: 25680238-25680238 on Build GRCh38
rs1025937225 Chr.15: 25680097-25680097 on Build GRCh38
rs138953004 Chr.15: 25680233-25680233 on Build GRCh38
rs759098238 Chr.15: 25680225-25680225 on Build GRCh38
rs528577942 Chr.15: 25680159-25680159 on Build GRCh38
rs377193909 Chr.15: 25680092-25680092 on Build GRCh38
rs115260856 Chr.15: 25680192-25680192 on Build GRCh38
rs751467738 Chr.15: 25680258-25680258 on Build GRCh38
rs752365063 Chr.15: 25680181-25680181 on Build GRCh38
rs543253900 Chr.15: 25680114-25680114 on Build GRCh38
rs116641809 Chr.15: 25680137-25680137 on Build GRCh38
rs546149030 Chr.15: 25680145-25680145 on Build GRCh38
rs756615990 Chr.15: 25680194-25680194 on Build GRCh38
rs937498012 Chr.15: 25680245-25680245 on Build GRCh38
rs758123629 Chr.15: 25680250-25680250 on Build GRCh38
rs777442267 Chr.15: 25680257-25680257 on Build GRCh38
rs760680989 Chr.15: 25680149-25680149 on Build GRCh38
rs981716503 Chr.15: 25680228-25680228 on Build GRCh38
rs780532261 Chr.15: 25680196-25680196 on Build GRCh38
rs111231177 Chr.15: 25680106-25680106 on Build GRCh38
rs752416936 Chr.15: 25680240-25680240 on Build GRCh38
rs768985513 Chr.15: 25680213-25680213 on Build GRCh38
rs754809069 Chr.15: 25680179-25680179 on Build GRCh38
rs201301127 Chr.15: 25680148-25680148 on Build GRCh38
rs749710242 Chr.15: 25680206-25680206 on Build GRCh38
rs372743966 Chr.15: 25680085-25680085 on Build GRCh38
rs185024491 Chr.15: 25680218-25680218 on Build GRCh38
rs761846602 Chr.15: 25680138-25680138 on Build GRCh38
rs774473987 Chr.15: 25680214-25680214 on Build GRCh38
rs763234901 Chr.15: 25680112-25680112 on Build GRCh38
rs957689135 Chr.15: 25680186-25680186 on Build GRCh38
rs772134207 Chr.15: 25680217-25680217 on Build GRCh38

View Full Product Details