Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36008449-36008718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE4
Entrez Gene ID
163183
Gene Name
spectrin repeat containing nuclear envelope family member 4
Gene Aliases
C19orf46, DFNB76, Nesp4
UniGene
Hs.436743
Gene Chromosome Location
Chr.19: 36003299-36008793 on Build GRCh38
Transcripts
NM_001039876.2 NM_001297735.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407431 c.61G>T p.G21* Chr.19: 36008621-36008621 on Build GRCh38
4154181 c.21G>T p.L7L Chr.19: 36008661-36008661 on Build GRCh38
1393027 c.105C>T p.P35P Chr.19: 36008577-36008577 on Build GRCh38
4752824 c.112G>T p.G38* Chr.19: 36008570-36008570 on Build GRCh38
1393028 c.94A>C p.T32P Chr.19: 36008588-36008588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201479405 Chr.19: 36008568-36008568 on Build GRCh38
rs758855710 Chr.19: 36008567-36008567 on Build GRCh38
rs769881264 Chr.19: 36008638-36008638 on Build GRCh38
rs746066870 Chr.19: 36008571-36008571 on Build GRCh38
rs761782441 Chr.19: 36008672-36008672 on Build GRCh38
rs764476302 Chr.19: 36008636-36008636 on Build GRCh38
rs748488209 Chr.19: 36008565-36008565 on Build GRCh38
rs1017783692 Chr.19: 36008697-36008697 on Build GRCh38
rs200751555 Chr.19: 36008577-36008577 on Build GRCh38
rs371943624 Chr.19: 36008541-36008541 on Build GRCh38
rs774065121 Chr.19: 36008663-36008663 on Build GRCh38
rs565009900 Chr.19: 36008587-36008587 on Build GRCh38
rs775716919 Chr.19: 36008572-36008572 on Build GRCh38
rs112235977 Chr.19: 36008510-36008510 on Build GRCh38
rs755883439 Chr.19: 36008557-36008557 on Build GRCh38
rs772750921 Chr.19: 36008509-36008509 on Build GRCh38
rs370082413 Chr.19: 36008660-36008660 on Build GRCh38
rs778068502 Chr.19: 36008595-36008595 on Build GRCh38
rs980905445 Chr.19: 36008640-36008640 on Build GRCh38
rs749513822 Chr.19: 36008656-36008656 on Build GRCh38
rs762846272 Chr.19: 36008534-36008534 on Build GRCh38
rs368026319 Chr.19: 36008518-36008518 on Build GRCh38
rs765002656 Chr.19: 36008525-36008525 on Build GRCh38
rs866074467 Chr.19: 36008579-36008579 on Build GRCh38
rs775312221 Chr.19: 36008529-36008529 on Build GRCh38
rs912874557 Chr.19: 36008696-36008696 on Build GRCh38
rs773309896 Chr.19: 36008680-36008680 on Build GRCh38
rs747476231 Chr.19: 36008569-36008569 on Build GRCh38
rs376125930 Chr.19: 36008691-36008691 on Build GRCh38
rs767035394 Chr.19: 36008582-36008582 on Build GRCh38
rs928159872 Chr.19: 36008643-36008643 on Build GRCh38
rs138787817 Chr.19: 36008586-36008586 on Build GRCh38
rs757752683 Chr.19: 36008605-36008605 on Build GRCh38
rs976457204 Chr.19: 36008507-36008507 on Build GRCh38
rs758800042 Chr.19: 36008559-36008560 on Build GRCh38
rs547543775 Chr.19: 36008655-36008655 on Build GRCh38
rs532471769 Chr.19: 36008598-36008598 on Build GRCh38
rs764049093 Chr.19: 36008535-36008535 on Build GRCh38
rs1018253553 Chr.19: 36008491-36008491 on Build GRCh38
rs760205161 Chr.19: 36008690-36008690 on Build GRCh38
rs373148223 Chr.19: 36008675-36008675 on Build GRCh38
rs759819586 Chr.19: 36008522-36008522 on Build GRCh38
rs749498016 Chr.19: 36008613-36008613 on Build GRCh38
rs965007530 Chr.19: 36008495-36008495 on Build GRCh38
rs200926652 Chr.19: 36008585-36008585 on Build GRCh38
rs747534534 Chr.19: 36008600-36008600 on Build GRCh38
rs771041135 Chr.19: 36008570-36008570 on Build GRCh38
rs762684679 Chr.19: 36008573-36008573 on Build GRCh38
rs779393935 Chr.19: 36008560-36008560 on Build GRCh38
rs376639796 Chr.19: 36008544-36008544 on Build GRCh38
rs751470589 Chr.19: 36008538-36008538 on Build GRCh38
rs916455445 Chr.19: 36008584-36008584 on Build GRCh38
rs781299860 Chr.19: 36008615-36008615 on Build GRCh38
rs774276326 Chr.19: 36008576-36008576 on Build GRCh38
rs766938134 Chr.19: 36008545-36008545 on Build GRCh38
rs750079185 Chr.19: 36008552-36008552 on Build GRCh38
rs939638065 Chr.19: 36008499-36008499 on Build GRCh38
rs1006420994 Chr.19: 36008476-36008476 on Build GRCh38
rs763806745 Chr.19: 36008574-36008574 on Build GRCh38
rs745908281 Chr.19: 36008624-36008624 on Build GRCh38
rs770930638 Chr.19: 36008516-36008516 on Build GRCh38

View Full Product Details