Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168944664-168944925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB11
Entrez Gene ID
8647
Gene Name
ATP binding cassette subfamily B member 11
Gene Aliases
ABC16, BRIC2, BSEP, PFIC-2, PFIC2, PGY4, SPGP
UniGene
Hs.658439
Gene Chromosome Location
Chr.2: 168915468-169031396 on Build GRCh38
Transcripts
NM_003742.2 XM_006712817.3 XM_011512077.2 XM_011512078.2 XM_011512080.2 XM_011512081.2 XM_017005165.1 XM_017005166.1 XM_017005167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009122 c.2494C>T p.R832C Chr.2: 168944721-168944721 on Build GRCh38
4816802 c.2550C>T p.L850L Chr.2: 168944665-168944665 on Build GRCh38
366342 c.2543A>T p.D848V Chr.2: 168944672-168944672 on Build GRCh38
6528514 c.2478C>G p.L826L Chr.2: 168944737-168944737 on Build GRCh38
5465174 c.2547C>T p.D849D Chr.2: 168944668-168944668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747065554 Chr.2: 168944847-168944847 on Build GRCh38
rs773596715 Chr.2: 168944743-168944743 on Build GRCh38
rs763478813 Chr.2: 168944903-168944903 on Build GRCh38
rs747266011 Chr.2: 168944726-168944726 on Build GRCh38
rs777570649 Chr.2: 168944713-168944713 on Build GRCh38
rs767146905 Chr.2: 168944778-168944778 on Build GRCh38
rs934431901 Chr.2: 168944794-168944794 on Build GRCh38
rs367874361 Chr.2: 168944692-168944692 on Build GRCh38
rs769056572 Chr.2: 168944856-168944856 on Build GRCh38
rs756323541 Chr.2: 168944727-168944727 on Build GRCh38
rs777104186 Chr.2: 168944736-168944736 on Build GRCh38
rs768970711 Chr.2: 168944732-168944732 on Build GRCh38
rs762069217 Chr.2: 168944737-168944737 on Build GRCh38
rs776712250 Chr.2: 168944866-168944866 on Build GRCh38
rs780323926 Chr.2: 168944828-168944828 on Build GRCh38
rs866565512 Chr.2: 168944757-168944757 on Build GRCh38
rs377240498 Chr.2: 168944688-168944688 on Build GRCh38
rs747862823 Chr.2: 168944699-168944699 on Build GRCh38
rs368092083 Chr.2: 168944899-168944899 on Build GRCh38
rs778002575 Chr.2: 168944822-168944822 on Build GRCh38
rs770066051 Chr.2: 168944882-168944882 on Build GRCh38
rs369231927 Chr.2: 168944695-168944695 on Build GRCh38
rs376959410 Chr.2: 168944775-168944775 on Build GRCh38
rs374548469 Chr.2: 168944762-168944762 on Build GRCh38
rs376255350 Chr.2: 168944720-168944720 on Build GRCh38
rs765892854 Chr.2: 168944760-168944760 on Build GRCh38
rs11568373 Chr.2: 168944893-168944893 on Build GRCh38
rs200903127 Chr.2: 168944821-168944821 on Build GRCh38
rs749207092 Chr.2: 168944813-168944813 on Build GRCh38
rs760018261 Chr.2: 168944784-168944784 on Build GRCh38
rs748335505 Chr.2: 168944879-168944879 on Build GRCh38
rs764214936 Chr.2: 168944808-168944808 on Build GRCh38
rs752606585 Chr.2: 168944787-168944787 on Build GRCh38
rs775663263 Chr.2: 168944725-168944725 on Build GRCh38
rs1043182223 Chr.2: 168944765-168944765 on Build GRCh38
rs763389560 Chr.2: 168944759-168944759 on Build GRCh38
rs765838062 Chr.2: 168944739-168944739 on Build GRCh38
rs1017537324 Chr.2: 168944832-168944832 on Build GRCh38
rs199967668 Chr.2: 168944799-168944799 on Build GRCh38
rs372757355 Chr.2: 168944700-168944700 on Build GRCh38
rs754572557 Chr.2: 168944694-168944694 on Build GRCh38
rs772294884 Chr.2: 168944721-168944721 on Build GRCh38
rs749186418 Chr.2: 168944716-168944716 on Build GRCh38

View Full Product Details