Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43977981-43978248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT3
Entrez Gene ID
56894
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases
LPAAT-GAMMA1, LPAAT3
UniGene
Hs.248785
Gene Chromosome Location
Chr.21: 43865160-43987594 on Build GRCh38
Transcripts
NM_001037553.1 NM_020132.4 XM_005261160.4 XM_006724029.3 XM_006724030.3 XM_006724031.3 XM_011529660.2 XM_011529661.2 XM_011529662.2 XM_011529663.2 XM_011529664.2 XM_011529665.2 XM_017028408.1 XM_017028409.1 XM_017028410.1 XM_017028411.1 XM_017028412.1 XM_017028413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941532 c.788T>C p.I263T Chr.21: 43978066-43978066 on Build GRCh38
4926672 c.834C>G p.Y278* Chr.21: 43978112-43978112 on Build GRCh38
5392995 c.770G>A p.R257K Chr.21: 43978048-43978048 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758135046 Chr.21: 43978152-43978152 on Build GRCh38
rs112377435 Chr.21: 43978199-43978199 on Build GRCh38
rs747939954 Chr.21: 43978060-43978060 on Build GRCh38
rs1022874929 Chr.21: 43978196-43978196 on Build GRCh38
rs777569532 Chr.21: 43978154-43978154 on Build GRCh38
rs757220851 Chr.21: 43978157-43978157 on Build GRCh38
rs753302460 Chr.21: 43978012-43978012 on Build GRCh38
rs372147252 Chr.21: 43978156-43978156 on Build GRCh38
rs763293403 Chr.21: 43978124-43978124 on Build GRCh38
rs746902461 Chr.21: 43978068-43978068 on Build GRCh38
rs554919809 Chr.21: 43978028-43978028 on Build GRCh38
rs1052130801 Chr.21: 43978183-43978183 on Build GRCh38
rs779480600 Chr.21: 43978006-43978006 on Build GRCh38
rs535119285 Chr.21: 43978201-43978201 on Build GRCh38
rs969975301 Chr.21: 43978203-43978203 on Build GRCh38
rs753507089 Chr.21: 43978020-43978020 on Build GRCh38
rs755634515 Chr.21: 43978069-43978069 on Build GRCh38
rs776804701 Chr.21: 43978070-43978070 on Build GRCh38
rs114513677 Chr.21: 43978204-43978204 on Build GRCh38
rs778503601 Chr.21: 43978056-43978056 on Build GRCh38
rs762331844 Chr.21: 43978135-43978135 on Build GRCh38
rs774734123 Chr.21: 43978134-43978134 on Build GRCh38
rs775834879 Chr.21: 43978112-43978112 on Build GRCh38
rs1026988009 Chr.21: 43978215-43978215 on Build GRCh38
rs770183276 Chr.21: 43978083-43978083 on Build GRCh38
rs753382243 Chr.21: 43978140-43978140 on Build GRCh38
rs199981101 Chr.21: 43978147-43978147 on Build GRCh38
rs781246419 Chr.21: 43978171-43978171 on Build GRCh38
rs374817231 Chr.21: 43978146-43978146 on Build GRCh38
rs917477491 Chr.21: 43978074-43978074 on Build GRCh38
rs565879993 Chr.21: 43978218-43978218 on Build GRCh38
rs777642168 Chr.21: 43978067-43978067 on Build GRCh38
rs951532049 Chr.21: 43978089-43978089 on Build GRCh38
rs921577648 Chr.21: 43978165-43978165 on Build GRCh38
rs771891429 Chr.21: 43978065-43978065 on Build GRCh38
rs1041523075 Chr.21: 43978210-43978210 on Build GRCh38
rs1034750409 Chr.21: 43978008-43978008 on Build GRCh38
rs895635960 Chr.21: 43978182-43978182 on Build GRCh38
rs189929741 Chr.21: 43978186-43978186 on Build GRCh38
rs759869743 Chr.21: 43978079-43978079 on Build GRCh38
rs764450949 Chr.21: 43978133-43978133 on Build GRCh38
rs370413568 Chr.21: 43978037-43978037 on Build GRCh38
rs765901362 Chr.21: 43978136-43978136 on Build GRCh38
rs754459878 Chr.21: 43978141-43978141 on Build GRCh38

View Full Product Details