Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207022644-207022869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf116
Entrez Gene ID
79098
Gene Name
chromosome 1 open reading frame 116
Gene Aliases
SARG
UniGene
Hs.32417
Gene Chromosome Location
Chr.1: 207018521-207032761 on Build GRCh38
Transcripts
NM_001083924.1 NM_023938.5 XM_005273259.1 XM_006711530.1 XM_011509973.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1626782 c.1003C>T p.L335L Chr.1: 207022761-207022761 on Build GRCh38
6024869 c.1075G>A p.D359N Chr.1: 207022689-207022689 on Build GRCh38
7358213 c.1073A>C p.Q358P Chr.1: 207022691-207022691 on Build GRCh38
4456630 c.1016C>T p.S339L Chr.1: 207022748-207022748 on Build GRCh38
6029635 c.969C>T p.S323S Chr.1: 207022795-207022795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772692630 Chr.1: 207022699-207022699 on Build GRCh38
rs770202598 Chr.1: 207022662-207022662 on Build GRCh38
rs761385462 Chr.1: 207022690-207022690 on Build GRCh38
rs914745648 Chr.1: 207022737-207022737 on Build GRCh38
rs779255398 Chr.1: 207022741-207022741 on Build GRCh38
rs375385107 Chr.1: 207022745-207022745 on Build GRCh38
rs35487307 Chr.1: 207022683-207022683 on Build GRCh38
rs190635695 Chr.1: 207022722-207022722 on Build GRCh38
rs1054991201 Chr.1: 207022716-207022716 on Build GRCh38
rs923576946 Chr.1: 207022759-207022759 on Build GRCh38
rs750613399 Chr.1: 207022814-207022814 on Build GRCh38
rs149898526 Chr.1: 207022727-207022727 on Build GRCh38
rs371156517 Chr.1: 207022730-207022730 on Build GRCh38
rs756324918 Chr.1: 207022822-207022822 on Build GRCh38
rs749559696 Chr.1: 207022711-207022721 on Build GRCh38
rs760298740 Chr.1: 207022705-207022705 on Build GRCh38
rs111307067 Chr.1: 207022669-207022669 on Build GRCh38
rs769153212 Chr.1: 207022761-207022761 on Build GRCh38
rs779458236 Chr.1: 207022833-207022833 on Build GRCh38
rs772874675 Chr.1: 207022698-207022699 on Build GRCh38
rs772472350 Chr.1: 207022844-207022844 on Build GRCh38
rs201485765 Chr.1: 207022794-207022794 on Build GRCh38
rs779188511 Chr.1: 207022764-207022764 on Build GRCh38
rs756269498 Chr.1: 207022750-207022750 on Build GRCh38
rs376328889 Chr.1: 207022724-207022724 on Build GRCh38
rs1056603056 Chr.1: 207022781-207022781 on Build GRCh38
rs760077787 Chr.1: 207022778-207022780 on Build GRCh38
rs760080872 Chr.1: 207022792-207022792 on Build GRCh38
rs771704465 Chr.1: 207022790-207022790 on Build GRCh38
rs749764904 Chr.1: 207022760-207022760 on Build GRCh38
rs956361647 Chr.1: 207022797-207022797 on Build GRCh38
rs765325005 Chr.1: 207022796-207022796 on Build GRCh38
rs753873351 Chr.1: 207022831-207022831 on Build GRCh38
rs762885342 Chr.1: 207022810-207022810 on Build GRCh38
rs749596824 Chr.1: 207022672-207022672 on Build GRCh38
rs767834591 Chr.1: 207022811-207022811 on Build GRCh38
rs547704570 Chr.1: 207022791-207022791 on Build GRCh38
rs200569642 Chr.1: 207022725-207022725 on Build GRCh38
rs748481037 Chr.1: 207022838-207022838 on Build GRCh38
rs774727060 Chr.1: 207022689-207022689 on Build GRCh38
rs139119607 Chr.1: 207022691-207022691 on Build GRCh38
rs781628755 Chr.1: 207022735-207022735 on Build GRCh38
rs771333520 Chr.1: 207022752-207022754 on Build GRCh38
rs145735004 Chr.1: 207022828-207022828 on Build GRCh38
rs757539856 Chr.1: 207022728-207022728 on Build GRCh38
rs755460908 Chr.1: 207022832-207022832 on Build GRCh38
rs752536090 Chr.1: 207022805-207022805 on Build GRCh38
rs748566878 Chr.1: 207022776-207022776 on Build GRCh38
rs977713611 Chr.1: 207022746-207022746 on Build GRCh38
rs774648306 Chr.1: 207022754-207022755 on Build GRCh38
rs770424183 Chr.1: 207022793-207022793 on Build GRCh38
rs780022265 Chr.1: 207022751-207022751 on Build GRCh38

View Full Product Details