Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19656369-19656606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3A2
Entrez Gene ID
224
Gene Name
aldehyde dehydrogenase 3 family member A2
Gene Aliases
ALDH10, FALDH, SLS
UniGene
Hs.499886
Gene Chromosome Location
Chr.17: 19648136-19677596 on Build GRCh38
Transcripts
NM_000382.2 NM_001031806.1 XM_011523732.1 XM_011523733.1 XM_017024355.1 XM_017024356.1 XM_017024357.1 XM_017024358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5452848 c.529C>T p.R177* Chr.17: 19656423-19656423 on Build GRCh38
3819016 c.587C>T p.A196V Chr.17: 19656481-19656481 on Build GRCh38
4064643 c.649G>A p.D217N Chr.17: 19656543-19656543 on Build GRCh38
7183933 c.565G>T p.V189F Chr.17: 19656459-19656459 on Build GRCh38
6346535 c.493G>T p.G165C Chr.17: 19656387-19656387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747414379 Chr.17: 19656466-19656466 on Build GRCh38
rs777261766 Chr.17: 19656484-19656484 on Build GRCh38
rs145709210 Chr.17: 19656564-19656564 on Build GRCh38
rs786205501 Chr.17: 19656517-19656517 on Build GRCh38
rs748735361 Chr.17: 19656488-19656488 on Build GRCh38
rs772967175 Chr.17: 19656464-19656465 on Build GRCh38
rs72547561 Chr.17: 19656423-19656423 on Build GRCh38
rs387906254 Chr.17: 19656415-19656415 on Build GRCh38
rs772822042 Chr.17: 19656540-19656540 on Build GRCh38
rs375800329 Chr.17: 19656524-19656524 on Build GRCh38
rs569611052 Chr.17: 19656571-19656571 on Build GRCh38
rs886052687 Chr.17: 19656574-19656574 on Build GRCh38
rs886041826 Chr.17: 19656468-19656469 on Build GRCh38
rs768202306 Chr.17: 19656520-19656520 on Build GRCh38
rs571845978 Chr.17: 19656414-19656414 on Build GRCh38
rs904950394 Chr.17: 19656446-19656446 on Build GRCh38
rs771277873 Chr.17: 19656479-19656479 on Build GRCh38
rs762531887 Chr.17: 19656406-19656406 on Build GRCh38
rs115977487 Chr.17: 19656457-19656457 on Build GRCh38
rs142587791 Chr.17: 19656537-19656537 on Build GRCh38
rs780052067 Chr.17: 19656438-19656438 on Build GRCh38
rs72547565 Chr.17: 19656572-19656572 on Build GRCh38
rs1009610342 Chr.17: 19656451-19656451 on Build GRCh38
rs371478620 Chr.17: 19656425-19656425 on Build GRCh38
rs769354565 Chr.17: 19656431-19656431 on Build GRCh38
rs140649688 Chr.17: 19656533-19656533 on Build GRCh38
rs370609228 Chr.17: 19656407-19656407 on Build GRCh38
rs944597535 Chr.17: 19656422-19656422 on Build GRCh38
rs967751381 Chr.17: 19656562-19656562 on Build GRCh38
rs72547564 Chr.17: 19656535-19656535 on Build GRCh38
rs72547563 Chr.17: 19656448-19656448 on Build GRCh38
rs766220815 Chr.17: 19656555-19656555 on Build GRCh38
rs755954696 Chr.17: 19656433-19656433 on Build GRCh38
rs867007123 Chr.17: 19656463-19656463 on Build GRCh38
rs1057516851 Chr.17: 19656471-19656471 on Build GRCh38
rs145431941 Chr.17: 19656424-19656424 on Build GRCh38
rs61757681 Chr.17: 19656458-19656458 on Build GRCh38
rs772457181 Chr.17: 19656460-19656460 on Build GRCh38
rs377139218 Chr.17: 19656465-19656465 on Build GRCh38
rs760519248 Chr.17: 19656553-19656553 on Build GRCh38
rs72547562 Chr.17: 19656445-19656445 on Build GRCh38
rs763732078 Chr.17: 19656412-19656412 on Build GRCh38
rs752850716 Chr.17: 19656578-19656578 on Build GRCh38
rs1049260316 Chr.17: 19656442-19656442 on Build GRCh38

View Full Product Details