Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51246656-51246915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAZAP2
Entrez Gene ID
9802
Gene Name
DAZ associated protein 2
Gene Aliases
PRTB
UniGene
Hs.369761
Gene Chromosome Location
Chr.12: 51238724-51246717 on Build GRCh38
Transcripts
NM_001136269.1 NM_001031628.1 NM_001033873.1

Gene Symbol
SMAGP
Entrez Gene ID
57228
Gene Name
small cell adhesion glycoprotein
Gene Aliases
hSMAGP
UniGene
Hs.652389
Gene Chromosome Location
Chr.12: 51245349-51270418 on Build GRCh38
Transcripts
NM_001136269.1 NM_001031628.1 NM_001033873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259619 c.115+7G>A p.? Chr.12: 51246744-51246744 on Build GRCh38
2009251 c.37G>T p.E13* Chr.12: 51246829-51246829 on Build GRCh38
6251916 c.52C>A p.P18T Chr.12: 51246814-51246814 on Build GRCh38
5640854 c.58T>G p.L20V Chr.12: 51246808-51246808 on Build GRCh38
1586559 c.60A>G p.L20L Chr.12: 51246806-51246806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180683045 Chr.12: 51246866-51246866 on Build GRCh38
rs761786839 Chr.12: 51246813-51246813 on Build GRCh38
rs1047943285 Chr.12: 51246728-51246728 on Build GRCh38
rs756404521 Chr.12: 51246725-51246725 on Build GRCh38
rs768182205 Chr.12: 51246744-51246745 on Build GRCh38
rs200401466 Chr.12: 51246767-51246767 on Build GRCh38
rs202077012 Chr.12: 51246877-51246877 on Build GRCh38
rs764431864 Chr.12: 51246696-51246696 on Build GRCh38
rs906642929 Chr.12: 51246873-51246873 on Build GRCh38
rs756563779 Chr.12: 51246844-51246844 on Build GRCh38
rs759419748 Chr.12: 51246778-51246778 on Build GRCh38
rs772997248 Chr.12: 51246760-51246760 on Build GRCh38
rs933502674 Chr.12: 51246688-51246688 on Build GRCh38
rs766883286 Chr.12: 51246845-51246845 on Build GRCh38
rs191455226 Chr.12: 51246706-51246706 on Build GRCh38
rs755399536 Chr.12: 51246857-51246857 on Build GRCh38
rs754568910 Chr.12: 51246745-51246745 on Build GRCh38
rs749677392 Chr.12: 51246736-51246736 on Build GRCh38
rs375988755 Chr.12: 51246704-51246704 on Build GRCh38
rs149526777 Chr.12: 51246888-51246888 on Build GRCh38
rs377338392 Chr.12: 51246757-51246757 on Build GRCh38
rs542086691 Chr.12: 51246747-51246747 on Build GRCh38
rs770801758 Chr.12: 51246769-51246769 on Build GRCh38
rs571966168 Chr.12: 51246811-51246811 on Build GRCh38
rs761104626 Chr.12: 51246824-51246824 on Build GRCh38
rs778519940 Chr.12: 51246858-51246858 on Build GRCh38
rs368812714 Chr.12: 51246794-51246794 on Build GRCh38
rs1021802210 Chr.12: 51246756-51246756 on Build GRCh38
rs957939870 Chr.12: 51246891-51246891 on Build GRCh38
rs376273085 Chr.12: 51246827-51246829 on Build GRCh38
rs965483694 Chr.12: 51246833-51246833 on Build GRCh38
rs540626685 Chr.12: 51246819-51246819 on Build GRCh38
rs754132797 Chr.12: 51246846-51246846 on Build GRCh38
rs755736203 Chr.12: 51246770-51246770 on Build GRCh38
rs763873810 Chr.12: 51246820-51246820 on Build GRCh38
rs999284329 Chr.12: 51246888-51246889 on Build GRCh38
rs767905283 Chr.12: 51246713-51246713 on Build GRCh38
rs750996847 Chr.12: 51246719-51246719 on Build GRCh38
rs370142414 Chr.12: 51246735-51246735 on Build GRCh38
rs764726983 Chr.12: 51246874-51246875 on Build GRCh38
rs1033306933 Chr.12: 51246698-51246698 on Build GRCh38
rs764346286 Chr.12: 51246790-51246790 on Build GRCh38

View Full Product Details