Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17588417-17588648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF10L
Entrez Gene ID
55160
Gene Name
Rho guanine nucleotide exchange factor 10 like
Gene Aliases
GrinchGEF
UniGene
Hs.443460
Gene Chromosome Location
Chr.1: 17520197-17697875 on Build GRCh38
Transcripts
NM_001011722.2 NM_001319837.1 NM_018125.3 NR_137287.1 NR_137288.1 XM_005245923.2 XM_005245925.2 XM_006710728.1 XM_006710729.1 XM_006710731.1 XM_011541691.2 XM_011541692.2 XM_011541693.2 XM_011541695.2 XM_011541696.2 XM_017001617.1 XM_017001619.1 XM_017001620.1 XM_017001621.1 XM_017001622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4025584 c.226C>A p.P76T Chr.1: 17588448-17588448 on Build GRCh38
4771205 c.233C>T p.P78L Chr.1: 17588455-17588455 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753803365 Chr.1: 17588471-17588471 on Build GRCh38
rs900393825 Chr.1: 17588556-17588556 on Build GRCh38
rs758573562 Chr.1: 17588484-17588484 on Build GRCh38
rs770460733 Chr.1: 17588456-17588456 on Build GRCh38
rs748558269 Chr.1: 17588509-17588509 on Build GRCh38
rs948373481 Chr.1: 17588561-17588561 on Build GRCh38
rs750431217 Chr.1: 17588482-17588482 on Build GRCh38
rs1054046149 Chr.1: 17588568-17588568 on Build GRCh38
rs765226704 Chr.1: 17588473-17588473 on Build GRCh38
rs186542657 Chr.1: 17588508-17588508 on Build GRCh38
rs773674715 Chr.1: 17588459-17588459 on Build GRCh38
rs116178304 Chr.1: 17588571-17588571 on Build GRCh38
rs551290009 Chr.1: 17588619-17588619 on Build GRCh38
rs780401691 Chr.1: 17588496-17588496 on Build GRCh38
rs1010386641 Chr.1: 17588589-17588589 on Build GRCh38
rs767114186 Chr.1: 17588466-17588466 on Build GRCh38
rs146072588 Chr.1: 17588472-17588472 on Build GRCh38
rs773797677 Chr.1: 17588515-17588515 on Build GRCh38
rs760436961 Chr.1: 17588470-17588470 on Build GRCh38
rs372972068 Chr.1: 17588529-17588529 on Build GRCh38
rs548939948 Chr.1: 17588455-17588455 on Build GRCh38
rs759049180 Chr.1: 17588462-17588462 on Build GRCh38
rs190925482 Chr.1: 17588530-17588530 on Build GRCh38
rs944532384 Chr.1: 17588527-17588527 on Build GRCh38
rs909599599 Chr.1: 17588573-17588573 on Build GRCh38
rs990814355 Chr.1: 17588493-17588493 on Build GRCh38
rs369264523 Chr.1: 17588525-17588525 on Build GRCh38
rs747274176 Chr.1: 17588497-17588497 on Build GRCh38
rs1020001303 Chr.1: 17588615-17588615 on Build GRCh38
rs941115063 Chr.1: 17588578-17588578 on Build GRCh38
rs978558564 Chr.1: 17588476-17588476 on Build GRCh38
rs770129253 Chr.1: 17588510-17588510 on Build GRCh38
rs930705781 Chr.1: 17588509-17588509 on Build GRCh38

View Full Product Details