Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18059424-18059693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH1
Entrez Gene ID
427
Gene Name
N-acylsphingosine amidohydrolase 1
Gene Aliases
AC, ACDase, ASAH, PHP, PHP32, SMAPME
UniGene
Hs.527412
Gene Chromosome Location
Chr.8: 18056299-18084998 on Build GRCh38
Transcripts
NM_001127505.2 NM_004315.5 NM_177924.4 XM_005273504.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291618 c.870A>C p.E290D Chr.8: 18059619-18059619 on Build GRCh38
6536474 c.941G>C p.R314T Chr.8: 18059596-18059596 on Build GRCh38
4512207 c.895C>T p.L299L Chr.8: 18059642-18059642 on Build GRCh38
3412897 c.922G>A p.D308N Chr.8: 18059460-18059460 on Build GRCh38
1097632 c.945G>T p.K315N Chr.8: 18059592-18059592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776590059 Chr.8: 18059617-18059617 on Build GRCh38
rs768737924 Chr.8: 18059613-18059613 on Build GRCh38
rs550035024 Chr.8: 18059536-18059536 on Build GRCh38
rs73581678 Chr.8: 18059500-18059500 on Build GRCh38
rs781646539 Chr.8: 18059565-18059565 on Build GRCh38
rs201442214 Chr.8: 18059525-18059525 on Build GRCh38
rs1038751008 Chr.8: 18059600-18059600 on Build GRCh38
rs689586 Chr.8: 18059488-18059488 on Build GRCh38
rs377398635 Chr.8: 18059527-18059527 on Build GRCh38
rs779310797 Chr.8: 18059485-18059485 on Build GRCh38
rs750455734 Chr.8: 18059642-18059642 on Build GRCh38
rs547575890 Chr.8: 18059477-18059477 on Build GRCh38
rs780542522 Chr.8: 18059461-18059461 on Build GRCh38
rs753916004 Chr.8: 18059524-18059524 on Build GRCh38
rs372742726 Chr.8: 18059515-18059515 on Build GRCh38
rs139585444 Chr.8: 18059454-18059454 on Build GRCh38
rs895669204 Chr.8: 18059656-18059656 on Build GRCh38
rs770536091 Chr.8: 18059494-18059494 on Build GRCh38
rs780454465 Chr.8: 18059547-18059547 on Build GRCh38
rs201082114 Chr.8: 18059563-18059563 on Build GRCh38
rs371154344 Chr.8: 18059632-18059632 on Build GRCh38
rs1010033653 Chr.8: 18059667-18059667 on Build GRCh38
rs747457090 Chr.8: 18059466-18059466 on Build GRCh38
rs751966694 Chr.8: 18059650-18059650 on Build GRCh38
rs149712271 Chr.8: 18059585-18059585 on Build GRCh38
rs760525887 Chr.8: 18059607-18059607 on Build GRCh38
rs750591004 Chr.8: 18059537-18059537 on Build GRCh38
rs758747892 Chr.8: 18059460-18059460 on Build GRCh38
rs774017376 Chr.8: 18059495-18059495 on Build GRCh38
rs369238481 Chr.8: 18059514-18059514 on Build GRCh38
rs397509415 Chr.8: 18059568-18059568 on Build GRCh38
rs748797618 Chr.8: 18059484-18059484 on Build GRCh38
rs1037765355 Chr.8: 18059594-18059594 on Build GRCh38
rs781668260 Chr.8: 18059663-18059663 on Build GRCh38
rs794729663 Chr.8: 18059639-18059639 on Build GRCh38
rs746288619 Chr.8: 18059499-18059499 on Build GRCh38
rs751913390 Chr.8: 18059552-18059552 on Build GRCh38
rs755283315 Chr.8: 18059652-18059652 on Build GRCh38
rs771847002 Chr.8: 18059603-18059603 on Build GRCh38
rs780767410 Chr.8: 18059521-18059521 on Build GRCh38
rs761801571 Chr.8: 18059630-18059630 on Build GRCh38
rs937056036 Chr.8: 18059634-18059634 on Build GRCh38
rs758703043 Chr.8: 18059541-18059541 on Build GRCh38
rs745596489 Chr.8: 18059602-18059602 on Build GRCh38
rs770411519 Chr.8: 18059580-18059580 on Build GRCh38
rs200718467 Chr.8: 18059501-18059501 on Build GRCh38
rs759257092 Chr.8: 18059498-18059498 on Build GRCh38
rs78267388 Chr.8: 18059579-18059579 on Build GRCh38
rs190788527 Chr.8: 18059646-18059646 on Build GRCh38
rs530331314 Chr.8: 18059520-18059520 on Build GRCh38
rs200565354 Chr.8: 18059586-18059586 on Build GRCh38
rs763224305 Chr.8: 18059645-18059645 on Build GRCh38
rs964761076 Chr.8: 18059519-18059519 on Build GRCh38
rs185687182 Chr.8: 18059468-18059468 on Build GRCh38

View Full Product Details