Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169387022-169387253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLZF1
Entrez Gene ID
8548
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
GOLGIN-45, JEM-1, JEM-1s, JEM1
UniGene
Hs.130746
Gene Chromosome Location
Chr.1: 169367879-169396544 on Build GRCh38
Transcripts
NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078498 c.1147G>A p.E383K Chr.1: 169387126-169387126 on Build GRCh38
6122306 c.1085C>T p.S362L Chr.1: 169387064-169387064 on Build GRCh38
4458014 c.1069C>T p.P357S Chr.1: 169387048-169387048 on Build GRCh38
6059173 c.1102G>T p.A368S Chr.1: 169387081-169387081 on Build GRCh38
7154697 c.1142G>C p.R381T Chr.1: 169387121-169387121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144655565 Chr.1: 169387071-169387071 on Build GRCh38
rs757473451 Chr.1: 169387125-169387125 on Build GRCh38
rs764515306 Chr.1: 169387207-169387207 on Build GRCh38
rs767962455 Chr.1: 169387102-169387102 on Build GRCh38
rs764427263 Chr.1: 169387117-169387117 on Build GRCh38
rs754141029 Chr.1: 169387124-169387124 on Build GRCh38
rs776031512 Chr.1: 169387200-169387200 on Build GRCh38
rs756533987 Chr.1: 169387115-169387115 on Build GRCh38
rs765905577 Chr.1: 169387190-169387190 on Build GRCh38
rs11544428 Chr.1: 169387111-169387111 on Build GRCh38
rs753168847 Chr.1: 169387103-169387103 on Build GRCh38
rs148163554 Chr.1: 169387089-169387089 on Build GRCh38
rs74383380 Chr.1: 169387060-169387060 on Build GRCh38
rs550591205 Chr.1: 169387177-169387177 on Build GRCh38
rs200837980 Chr.1: 169387151-169387151 on Build GRCh38
rs746930700 Chr.1: 169387158-169387158 on Build GRCh38
rs769356495 Chr.1: 169387180-169387180 on Build GRCh38
rs761181111 Chr.1: 169387205-169387205 on Build GRCh38
rs373643357 Chr.1: 169387154-169387154 on Build GRCh38
rs551974736 Chr.1: 169387210-169387210 on Build GRCh38
rs776574218 Chr.1: 169387160-169387160 on Build GRCh38
rs768568248 Chr.1: 169387063-169387063 on Build GRCh38
rs762480944 Chr.1: 169387097-169387097 on Build GRCh38
rs116192263 Chr.1: 169387189-169387189 on Build GRCh38
rs1044112526 Chr.1: 169387193-169387193 on Build GRCh38
rs779153670 Chr.1: 169387142-169387142 on Build GRCh38
rs746276207 Chr.1: 169387216-169387216 on Build GRCh38
rs761571132 Chr.1: 169387076-169387076 on Build GRCh38
rs138679316 Chr.1: 169387084-169387084 on Build GRCh38
rs189043990 Chr.1: 169387188-169387188 on Build GRCh38
rs768658214 Chr.1: 169387159-169387159 on Build GRCh38
rs757591575 Chr.1: 169387225-169387225 on Build GRCh38

View Full Product Details