Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73432157-73432430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
UniGene
Hs.184720
Gene Chromosome Location
Chr.2: 73385758-73609919 on Build GRCh38
Transcripts
NM_015120.4

SNP Detail

SNP ID Chromosome Location
rs149239794 Chr.2: 73432310-73432310 on Build GRCh38
rs755499539 Chr.2: 73432312-73432312 on Build GRCh38
rs749794703 Chr.2: 73432245-73432245 on Build GRCh38
rs993181191 Chr.2: 73432352-73432352 on Build GRCh38
rs760775500 Chr.2: 73432266-73432266 on Build GRCh38
rs370873335 Chr.2: 73432212-73432212 on Build GRCh38
rs772485366 Chr.2: 73432260-73432260 on Build GRCh38
rs572922738 Chr.2: 73432195-73432195 on Build GRCh38
rs544918897 Chr.2: 73432357-73432357 on Build GRCh38
rs377283762 Chr.2: 73432198-73432198 on Build GRCh38
rs748453467 Chr.2: 73432323-73432323 on Build GRCh38
rs947794687 Chr.2: 73432302-73432302 on Build GRCh38
rs200933935 Chr.2: 73432295-73432295 on Build GRCh38
rs886056299 Chr.2: 73432199-73432199 on Build GRCh38
rs774631882 Chr.2: 73432252-73432252 on Build GRCh38
rs373469023 Chr.2: 73432193-73432193 on Build GRCh38
rs551157480 Chr.2: 73432343-73432343 on Build GRCh38
rs769308689 Chr.2: 73432238-73432238 on Build GRCh38
rs200454461 Chr.2: 73432279-73432279 on Build GRCh38
rs17434013 Chr.2: 73432328-73432328 on Build GRCh38
rs762837795 Chr.2: 73432277-73432277 on Build GRCh38
rs745317947 Chr.2: 73432217-73432217 on Build GRCh38
rs761600950 Chr.2: 73432319-73432319 on Build GRCh38
rs201990757 Chr.2: 73432259-73432259 on Build GRCh38
rs540547127 Chr.2: 73432211-73432211 on Build GRCh38
rs779412612 Chr.2: 73432322-73432322 on Build GRCh38
rs774335945 Chr.2: 73432202-73432202 on Build GRCh38
rs758407495 Chr.2: 73432252-73432265 on Build GRCh38
rs771010537 Chr.2: 73432338-73432338 on Build GRCh38
rs775455907 Chr.2: 73432275-73432275 on Build GRCh38
rs780562456 Chr.2: 73432241-73432241 on Build GRCh38
rs757050699 Chr.2: 73432281-73432281 on Build GRCh38
rs879046601 Chr.2: 73432201-73432201 on Build GRCh38
rs773569879 Chr.2: 73432262-73432262 on Build GRCh38
rs938822642 Chr.2: 73432349-73432349 on Build GRCh38
rs374677818 Chr.2: 73432329-73432329 on Build GRCh38
rs755613962 Chr.2: 73432296-73432296 on Build GRCh38
rs766390643 Chr.2: 73432270-73432270 on Build GRCh38
rs376610564 Chr.2: 73432306-73432306 on Build GRCh38
rs777458464 Chr.2: 73432204-73432204 on Build GRCh38
rs767100210 Chr.2: 73432284-73432284 on Build GRCh38
rs763926547 Chr.2: 73432278-73432278 on Build GRCh38
rs935279576 Chr.2: 73432240-73432240 on Build GRCh38
rs989088490 Chr.2: 73432251-73432251 on Build GRCh38
rs747264158 Chr.2: 73432335-73432335 on Build GRCh38
rs766524523 Chr.2: 73432328-73432328 on Build GRCh38

View Full Product Details