Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26336166-26336399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1L
Entrez Gene ID
55057
Gene Name
absent in melanoma 1-like
Gene Aliases
CRYBG2
UniGene
Hs.128738
Gene Chromosome Location
Chr.1: 26321859-26354130 on Build GRCh38
Transcripts
NM_001039775.3 XM_005245918.2 XM_011541672.1 XM_011541673.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143721 c.1425G>T p.E475D Chr.1: 26336362-26336362 on Build GRCh38
4791113 c.1469C>A p.P490H Chr.1: 26336188-26336188 on Build GRCh38
3934645 c.940C>T p.Q314* Chr.1: 26336204-26336204 on Build GRCh38
4791112 c.956C>A p.P319H Chr.1: 26336188-26336188 on Build GRCh38
4143720 c.912G>T p.E304D Chr.1: 26336362-26336362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752320496 Chr.1: 26336294-26336294 on Build GRCh38
rs374452979 Chr.1: 26336235-26336235 on Build GRCh38
rs764362233 Chr.1: 26336230-26336230 on Build GRCh38
rs540474304 Chr.1: 26336288-26336288 on Build GRCh38
rs777790139 Chr.1: 26336252-26336252 on Build GRCh38
rs554803698 Chr.1: 26336253-26336253 on Build GRCh38
rs908958249 Chr.1: 26336232-26336232 on Build GRCh38
rs760845135 Chr.1: 26336375-26336375 on Build GRCh38
rs760655355 Chr.1: 26336229-26336229 on Build GRCh38
rs1002523911 Chr.1: 26336231-26336231 on Build GRCh38
rs558884185 Chr.1: 26336288-26336288 on Build GRCh38
rs761978192 Chr.1: 26336244-26336244 on Build GRCh38
rs755547998 Chr.1: 26336296-26336296 on Build GRCh38
rs775651908 Chr.1: 26336217-26336217 on Build GRCh38
rs755966991 Chr.1: 26336228-26336229 on Build GRCh38
rs962279002 Chr.1: 26336242-26336242 on Build GRCh38
rs746160149 Chr.1: 26336358-26336358 on Build GRCh38
rs768697094 Chr.1: 26336376-26336376 on Build GRCh38
rs7517559 Chr.1: 26336283-26336283 on Build GRCh38
rs900372500 Chr.1: 26336267-26336267 on Build GRCh38
rs376253645 Chr.1: 26336316-26336316 on Build GRCh38
rs772391205 Chr.1: 26336199-26336199 on Build GRCh38
rs766197855 Chr.1: 26336247-26336247 on Build GRCh38
rs968553090 Chr.1: 26336367-26336367 on Build GRCh38
rs760120484 Chr.1: 26336191-26336191 on Build GRCh38
rs1020932546 Chr.1: 26336336-26336336 on Build GRCh38
rs370660970 Chr.1: 26336319-26336319 on Build GRCh38
rs771014750 Chr.1: 26336350-26336351 on Build GRCh38
rs1000987861 Chr.1: 26336370-26336370 on Build GRCh38
rs777618874 Chr.1: 26336305-26336305 on Build GRCh38
rs906881252 Chr.1: 26336224-26336224 on Build GRCh38
rs779423566 Chr.1: 26336328-26336328 on Build GRCh38
rs149208276 Chr.1: 26336368-26336368 on Build GRCh38
rs891680837 Chr.1: 26336298-26336298 on Build GRCh38
rs772690353 Chr.1: 26336364-26336364 on Build GRCh38
rs754959740 Chr.1: 26336254-26336254 on Build GRCh38
rs767107659 Chr.1: 26336300-26336300 on Build GRCh38
rs997303401 Chr.1: 26336275-26336275 on Build GRCh38

View Full Product Details