Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122742569-122742812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS12
Entrez Gene ID
166379
Gene Name
Bardet-Biedl syndrome 12
Gene Aliases
C4orf24
UniGene
Hs.400698
Gene Chromosome Location
Chr.4: 122700437-122744943 on Build GRCh38
Transcripts
NM_001178007.1 NM_152618.2 XM_011531680.2 XM_017007831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6166073 c.698A>T p.H233L Chr.4: 122742590-122742590 on Build GRCh38
5995386 c.677G>A p.C226Y Chr.4: 122742569-122742569 on Build GRCh38
1131281 c.697C>A p.H233N Chr.4: 122742589-122742589 on Build GRCh38
3945952 c.753A>G p.G251G Chr.4: 122742645-122742645 on Build GRCh38
7357325 c.748G>A p.D250N Chr.4: 122742640-122742640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751181744 Chr.4: 122742787-122742787 on Build GRCh38
rs758191621 Chr.4: 122742637-122742637 on Build GRCh38
rs148604293 Chr.4: 122742677-122742677 on Build GRCh38
rs974173261 Chr.4: 122742681-122742681 on Build GRCh38
rs121918328 Chr.4: 122742757-122742757 on Build GRCh38
rs769485563 Chr.4: 122742682-122742682 on Build GRCh38
rs766149148 Chr.4: 122742774-122742774 on Build GRCh38
rs981575853 Chr.4: 122742771-122742771 on Build GRCh38
rs773045310 Chr.4: 122742683-122742683 on Build GRCh38
rs201227132 Chr.4: 122742715-122742715 on Build GRCh38
rs865920726 Chr.4: 122742670-122742671 on Build GRCh38
rs150040166 Chr.4: 122742679-122742679 on Build GRCh38
rs915644000 Chr.4: 122742701-122742701 on Build GRCh38
rs778226928 Chr.4: 122742626-122742626 on Build GRCh38
rs996402604 Chr.4: 122742733-122742733 on Build GRCh38
rs774510452 Chr.4: 122742702-122742702 on Build GRCh38
rs34094714 Chr.4: 122742723-122742724 on Build GRCh38
rs756441486 Chr.4: 122742622-122742622 on Build GRCh38
rs778624524 Chr.4: 122742679-122742680 on Build GRCh38
rs751682549 Chr.4: 122742600-122742600 on Build GRCh38
rs375150883 Chr.4: 122742727-122742727 on Build GRCh38
rs745504524 Chr.4: 122742750-122742752 on Build GRCh38
rs17006082 Chr.4: 122742606-122742606 on Build GRCh38
rs185341959 Chr.4: 122742657-122742657 on Build GRCh38
rs757672620 Chr.4: 122742765-122742765 on Build GRCh38
rs948467559 Chr.4: 122742716-122742716 on Build GRCh38
rs764693618 Chr.4: 122742746-122742746 on Build GRCh38
rs754242636 Chr.4: 122742628-122742628 on Build GRCh38
rs140537219 Chr.4: 122742611-122742611 on Build GRCh38
rs754293872 Chr.4: 122742755-122742755 on Build GRCh38
rs762709515 Chr.4: 122742690-122742690 on Build GRCh38
rs768162160 Chr.4: 122742668-122742668 on Build GRCh38
rs746565072 Chr.4: 122742667-122742667 on Build GRCh38
rs759567833 Chr.4: 122742705-122742705 on Build GRCh38
rs761302293 Chr.4: 122742734-122742734 on Build GRCh38
rs771195621 Chr.4: 122742694-122742694 on Build GRCh38

View Full Product Details