Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72823957-72824226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG16L2
Entrez Gene ID
89849
Gene Name
autophagy related 16 like 2
Gene Aliases
ATG16B, WDR80
UniGene
Hs.653186
Gene Chromosome Location
Chr.11: 72814406-72843740 on Build GRCh38
Transcripts
NM_001318766.1 NM_033388.1 XM_005274376.4 XM_006718732.2 XM_006718733.3 XM_006718734.2 XM_011545332.1 XM_011545333.1 XM_011545334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3453123 c.875A>G p.K292R Chr.11: 72824110-72824110 on Build GRCh38
7090313 c.844C>A p.L282M Chr.11: 72824079-72824079 on Build GRCh38
5739371 c.848C>T p.T283M Chr.11: 72824083-72824083 on Build GRCh38
6224285 c.849G>A p.T283T Chr.11: 72824084-72824084 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs934217236 Chr.11: 72824201-72824201 on Build GRCh38
rs548428200 Chr.11: 72824184-72824184 on Build GRCh38
rs766083733 Chr.11: 72824164-72824164 on Build GRCh38
rs749977076 Chr.11: 72824018-72824018 on Build GRCh38
rs762042459 Chr.11: 72824070-72824070 on Build GRCh38
rs767551802 Chr.11: 72824122-72824125 on Build GRCh38
rs749316023 Chr.11: 72824141-72824141 on Build GRCh38
rs760297673 Chr.11: 72824163-72824163 on Build GRCh38
rs758316776 Chr.11: 72824046-72824046 on Build GRCh38
rs753432918 Chr.11: 72824167-72824167 on Build GRCh38
rs967483695 Chr.11: 72824005-72824005 on Build GRCh38
rs79145554 Chr.11: 72824122-72824122 on Build GRCh38
rs199982763 Chr.11: 72824155-72824155 on Build GRCh38
rs745976106 Chr.11: 72824047-72824047 on Build GRCh38
rs769823638 Chr.11: 72824049-72824049 on Build GRCh38
rs200822228 Chr.11: 72824129-72824129 on Build GRCh38
rs748965358 Chr.11: 72824084-72824084 on Build GRCh38
rs756241191 Chr.11: 72824140-72824141 on Build GRCh38
rs754974392 Chr.11: 72824094-72824095 on Build GRCh38
rs756981735 Chr.11: 72824015-72824015 on Build GRCh38
rs372293223 Chr.11: 72824078-72824078 on Build GRCh38
rs775543374 Chr.11: 72824053-72824053 on Build GRCh38
rs769179882 Chr.11: 72824146-72824146 on Build GRCh38
rs565219118 Chr.11: 72824036-72824036 on Build GRCh38
rs896109077 Chr.11: 72824017-72824017 on Build GRCh38
rs773617764 Chr.11: 72824157-72824157 on Build GRCh38
rs956656729 Chr.11: 72824151-72824153 on Build GRCh38
rs970386752 Chr.11: 72824086-72824086 on Build GRCh38
rs112951181 Chr.11: 72824038-72824038 on Build GRCh38
rs752543316 Chr.11: 72824107-72824107 on Build GRCh38
rs769972686 Chr.11: 72824136-72824136 on Build GRCh38
rs774719029 Chr.11: 72824064-72824064 on Build GRCh38
rs1041773030 Chr.11: 72824202-72824202 on Build GRCh38
rs768243020 Chr.11: 72824185-72824190 on Build GRCh38
rs780150270 Chr.11: 72824021-72824021 on Build GRCh38
rs763491240 Chr.11: 72824060-72824060 on Build GRCh38
rs749292068 Chr.11: 72824025-72824025 on Build GRCh38
rs566703479 Chr.11: 72824205-72824205 on Build GRCh38
rs573021635 Chr.11: 72824029-72824029 on Build GRCh38
rs780009142 Chr.11: 72824094-72824094 on Build GRCh38
rs372312861 Chr.11: 72824147-72824147 on Build GRCh38
rs368710471 Chr.11: 72824131-72824131 on Build GRCh38
rs370361758 Chr.11: 72824063-72824063 on Build GRCh38
rs998162044 Chr.11: 72823993-72823993 on Build GRCh38
rs201827763 Chr.11: 72824034-72824034 on Build GRCh38
rs562772338 Chr.11: 72824154-72824154 on Build GRCh38
rs1057301808 Chr.11: 72824001-72824001 on Build GRCh38
rs749982182 Chr.11: 72824081-72824081 on Build GRCh38
rs752823580 Chr.11: 72824126-72824127 on Build GRCh38
rs776653026 Chr.11: 72824039-72824039 on Build GRCh38
rs1020277109 Chr.11: 72823997-72823997 on Build GRCh38
rs780155191 Chr.11: 72824138-72824138 on Build GRCh38
rs778956881 Chr.11: 72824092-72824092 on Build GRCh38
rs767027658 Chr.11: 72824016-72824016 on Build GRCh38
rs754991850 Chr.11: 72824090-72824090 on Build GRCh38
rs758071790 Chr.11: 72824114-72824114 on Build GRCh38
rs375195570 Chr.11: 72824020-72824020 on Build GRCh38
rs139269726 Chr.11: 72824083-72824083 on Build GRCh38

View Full Product Details