Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 64581627-64581900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHLHE22
Entrez Gene ID
27319
Gene Name
basic helix-loop-helix family member e22
Gene Aliases
BHLHB5, Beta3, CAGL85, TNRC20
UniGene
Hs.388788
Gene Chromosome Location
Chr.8: 64580238-64583634 on Build GRCh38
Transcripts
NM_152414.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3716075 c.891G>A p.A297A Chr.8: 64581681-64581681 on Build GRCh38
6392758 c.854C>T p.T285M Chr.8: 64581644-64581644 on Build GRCh38
3779303 c.991G>A p.A331T Chr.8: 64581781-64581781 on Build GRCh38
4628770 c.896C>T p.A299V Chr.8: 64581686-64581686 on Build GRCh38
3650163 c.1074C>T p.S358S Chr.8: 64581864-64581864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045892167 Chr.8: 64581831-64581831 on Build GRCh38
rs775317700 Chr.8: 64581660-64581660 on Build GRCh38
rs764064764 Chr.8: 64581714-64581714 on Build GRCh38
rs562585263 Chr.8: 64581681-64581681 on Build GRCh38
rs757685270 Chr.8: 64581718-64581718 on Build GRCh38
rs773983223 Chr.8: 64581880-64581880 on Build GRCh38
rs920864420 Chr.8: 64581761-64581761 on Build GRCh38
rs753992551 Chr.8: 64581716-64581716 on Build GRCh38
rs866854588 Chr.8: 64581700-64581700 on Build GRCh38
rs34169687 Chr.8: 64581782-64581782 on Build GRCh38
rs765776454 Chr.8: 64581826-64581826 on Build GRCh38
rs769298111 Chr.8: 64581858-64581858 on Build GRCh38
rs779589370 Chr.8: 64581719-64581719 on Build GRCh38
rs759032356 Chr.8: 64581833-64581833 on Build GRCh38
rs764428609 Chr.8: 64581814-64581814 on Build GRCh38
rs750781448 Chr.8: 64581720-64581720 on Build GRCh38
rs753814211 Chr.8: 64581818-64581818 on Build GRCh38
rs768064913 Chr.8: 64581669-64581669 on Build GRCh38
rs780918012 Chr.8: 64581850-64581850 on Build GRCh38
rs543941490 Chr.8: 64581648-64581648 on Build GRCh38
rs750981140 Chr.8: 64581832-64581832 on Build GRCh38
rs754563189 Chr.8: 64581846-64581846 on Build GRCh38
rs779796027 Chr.8: 64581735-64581735 on Build GRCh38
rs761653987 Chr.8: 64581675-64581675 on Build GRCh38
rs769669417 Chr.8: 64581783-64581783 on Build GRCh38
rs34233614 Chr.8: 64581738-64581738 on Build GRCh38
rs762758678 Chr.8: 64581696-64581696 on Build GRCh38
rs780563329 Chr.8: 64581834-64581834 on Build GRCh38
rs770721497 Chr.8: 64581805-64581805 on Build GRCh38
rs908026012 Chr.8: 64581819-64581819 on Build GRCh38
rs749205922 Chr.8: 64581870-64581870 on Build GRCh38
rs776265176 Chr.8: 64581673-64581673 on Build GRCh38
rs36035514 Chr.8: 64581646-64581646 on Build GRCh38
rs34144912 Chr.8: 64581711-64581711 on Build GRCh38
rs34641263 Chr.8: 64581825-64581825 on Build GRCh38
rs747571849 Chr.8: 64581852-64581852 on Build GRCh38
rs1018947663 Chr.8: 64581649-64581649 on Build GRCh38
rs142170160 Chr.8: 64581693-64581693 on Build GRCh38
rs34155586 Chr.8: 64581804-64581804 on Build GRCh38
rs35800845 Chr.8: 64581706-64581706 on Build GRCh38
rs371051368 Chr.8: 64581824-64581824 on Build GRCh38
rs760799564 Chr.8: 64581809-64581809 on Build GRCh38
rs747740432 Chr.8: 64581779-64581779 on Build GRCh38
rs767686453 Chr.8: 64581701-64581701 on Build GRCh38
rs746461876 Chr.8: 64581665-64581665 on Build GRCh38
rs758839125 Chr.8: 64581732-64581732 on Build GRCh38
rs771806180 Chr.8: 64581654-64581654 on Build GRCh38
rs201511746 Chr.8: 64581845-64581845 on Build GRCh38
rs768423797 Chr.8: 64581745-64581745 on Build GRCh38
rs79001543 Chr.8: 64581876-64581876 on Build GRCh38
rs937001665 Chr.8: 64581855-64581855 on Build GRCh38
rs1049640253 Chr.8: 64581802-64581802 on Build GRCh38
rs528823279 Chr.8: 64581847-64581847 on Build GRCh38
rs180818429 Chr.8: 64581869-64581869 on Build GRCh38
rs546421536 Chr.8: 64581741-64581741 on Build GRCh38
rs780535265 Chr.8: 64581756-64581756 on Build GRCh38
rs774257929 Chr.8: 64581806-64581806 on Build GRCh38
rs35928427 Chr.8: 64581663-64581663 on Build GRCh38
rs35315171 Chr.8: 64581828-64581828 on Build GRCh38
rs773163934 Chr.8: 64581801-64581801 on Build GRCh38

View Full Product Details