Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112162416-112162687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD4
Entrez Gene ID
283450
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Gene Aliases
C12orf51, POTAGE
UniGene
Hs.530943
Gene Chromosome Location
Chr.12: 112160188-112382092 on Build GRCh38
Transcripts
NM_001109662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991648 c.11932A>G p.I3978V Chr.12: 112162446-112162446 on Build GRCh38
935166 c.11950C>T p.R3984C Chr.12: 112162428-112162428 on Build GRCh38
272404 c.11956G>A p.A3986T Chr.12: 112162422-112162422 on Build GRCh38
4885723 c.11865C>A p.D3955E Chr.12: 112162513-112162513 on Build GRCh38
6505345 c.11911C>T p.P3971S Chr.12: 112162467-112162467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780844632 Chr.12: 112162510-112162510 on Build GRCh38
rs751035258 Chr.12: 112162507-112162507 on Build GRCh38
rs972440909 Chr.12: 112162609-112162609 on Build GRCh38
rs73207617 Chr.12: 112162621-112162621 on Build GRCh38
rs755020522 Chr.12: 112162541-112162541 on Build GRCh38
rs746373458 Chr.12: 112162565-112162565 on Build GRCh38
rs779610403 Chr.12: 112162498-112162498 on Build GRCh38
rs779195250 Chr.12: 112162562-112162562 on Build GRCh38
rs922376478 Chr.12: 112162617-112162617 on Build GRCh38
rs191600493 Chr.12: 112162634-112162634 on Build GRCh38
rs754564804 Chr.12: 112162508-112162508 on Build GRCh38
rs749614514 Chr.12: 112162543-112162543 on Build GRCh38
rs750348704 Chr.12: 112162438-112162438 on Build GRCh38
rs1043881301 Chr.12: 112162663-112162663 on Build GRCh38
rs927961683 Chr.12: 112162536-112162536 on Build GRCh38
rs1011006127 Chr.12: 112162657-112162657 on Build GRCh38
rs535569640 Chr.12: 112162440-112162440 on Build GRCh38
rs771256812 Chr.12: 112162546-112162546 on Build GRCh38
rs781115259 Chr.12: 112162599-112162599 on Build GRCh38
rs985530202 Chr.12: 112162660-112162660 on Build GRCh38

View Full Product Details