Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74573003-74573256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000275.1 XM_017000276.1 XM_017000277.1 XM_017000278.1 XM_017000279.1 XM_017000280.1 XM_017000281.1 XM_017000282.1 XM_017000283.1 XM_017000284.1 XM_017000285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358263 c.2579G>T p.G860V Chr.1: 74573131-74573131 on Build GRCh38
4908539 c.2675C>T p.A892V Chr.1: 74573035-74573035 on Build GRCh38
6127697 c.2617G>C p.D873H Chr.1: 74573093-74573093 on Build GRCh38
6064393 c.2592A>G p.A864A Chr.1: 74573118-74573118 on Build GRCh38
309648 c.2696T>A p.L899* Chr.1: 74573014-74573014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549850597 Chr.1: 74573078-74573078 on Build GRCh38
rs914638032 Chr.1: 74573049-74573049 on Build GRCh38
rs779107144 Chr.1: 74573129-74573129 on Build GRCh38
rs776725780 Chr.1: 74573146-74573146 on Build GRCh38
rs112583366 Chr.1: 74573063-74573063 on Build GRCh38
rs760378727 Chr.1: 74573171-74573171 on Build GRCh38
rs116636724 Chr.1: 74573152-74573152 on Build GRCh38
rs767224975 Chr.1: 74573092-74573092 on Build GRCh38
rs149957800 Chr.1: 74573030-74573030 on Build GRCh38
rs779581893 Chr.1: 74573113-74573113 on Build GRCh38
rs765967118 Chr.1: 74573183-74573183 on Build GRCh38
rs757462593 Chr.1: 74573208-74573208 on Build GRCh38
rs750090566 Chr.1: 74573102-74573102 on Build GRCh38
rs761705838 Chr.1: 74573087-74573087 on Build GRCh38
rs772182365 Chr.1: 74573228-74573228 on Build GRCh38
rs773184825 Chr.1: 74573169-74573169 on Build GRCh38
rs60711359 Chr.1: 74573196-74573196 on Build GRCh38
rs779315737 Chr.1: 74573131-74573131 on Build GRCh38
rs138902428 Chr.1: 74573149-74573149 on Build GRCh38
rs747225871 Chr.1: 74573140-74573140 on Build GRCh38
rs545646275 Chr.1: 74573025-74573025 on Build GRCh38
rs776195150 Chr.1: 74573145-74573145 on Build GRCh38
rs781275566 Chr.1: 74573216-74573216 on Build GRCh38
rs753344414 Chr.1: 74573189-74573189 on Build GRCh38
rs770983302 Chr.1: 74573076-74573076 on Build GRCh38
rs1003535105 Chr.1: 74573088-74573088 on Build GRCh38
rs555839913 Chr.1: 74573220-74573220 on Build GRCh38
rs147806023 Chr.1: 74573054-74573054 on Build GRCh38
rs895625895 Chr.1: 74573071-74573071 on Build GRCh38
rs1016185785 Chr.1: 74573148-74573148 on Build GRCh38
rs547343006 Chr.1: 74573147-74573147 on Build GRCh38
rs867665343 Chr.1: 74573197-74573197 on Build GRCh38
rs753189414 Chr.1: 74573200-74573200 on Build GRCh38
rs747397482 Chr.1: 74573207-74573207 on Build GRCh38
rs778539431 Chr.1: 74573186-74573186 on Build GRCh38
rs762927326 Chr.1: 74573082-74573082 on Build GRCh38
rs373173004 Chr.1: 74573070-74573070 on Build GRCh38
rs751330822 Chr.1: 74573086-74573086 on Build GRCh38
rs150485650 Chr.1: 74573138-74573138 on Build GRCh38
rs537105429 Chr.1: 74573219-74573219 on Build GRCh38
rs761236621 Chr.1: 74573194-74573195 on Build GRCh38
rs755460357 Chr.1: 74573128-74573128 on Build GRCh38
rs749799744 Chr.1: 74573026-74573026 on Build GRCh38
rs923534263 Chr.1: 74573161-74573161 on Build GRCh38
rs200840954 Chr.1: 74573195-74573195 on Build GRCh38
rs199985207 Chr.1: 74573117-74573117 on Build GRCh38
rs772238066 Chr.1: 74573059-74573059 on Build GRCh38
rs374973689 Chr.1: 74573223-74573223 on Build GRCh38
rs767419093 Chr.1: 74573160-74573160 on Build GRCh38
rs35151101 Chr.1: 74573224-74573225 on Build GRCh38
rs763962348 Chr.1: 74573084-74573084 on Build GRCh38
rs780597999 Chr.1: 74573126-74573126 on Build GRCh38
rs755899755 Chr.1: 74573108-74573108 on Build GRCh38
rs748071177 Chr.1: 74573224-74573224 on Build GRCh38
rs778078353 Chr.1: 74573206-74573206 on Build GRCh38
rs143993406 Chr.1: 74573051-74573051 on Build GRCh38
rs772438085 Chr.1: 74573134-74573134 on Build GRCh38
rs773090775 Chr.1: 74573230-74573230 on Build GRCh38
rs372137519 Chr.1: 74573145-74573145 on Build GRCh38
rs779310921 Chr.1: 74573031-74573031 on Build GRCh38
rs778167980 Chr.1: 74573137-74573137 on Build GRCh38
rs894153165 Chr.1: 74573132-74573132 on Build GRCh38
rs535750867 Chr.1: 74573133-74573133 on Build GRCh38

View Full Product Details