Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63630889-63631156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL3
Entrez Gene ID
25923
Gene Name
atlastin GTPase 3
Gene Aliases
HSN1F
UniGene
Hs.356719
Gene Chromosome Location
Chr.11: 63624082-63671974 on Build GRCh38
Transcripts
NM_001290048.1 NM_015459.4 XM_006718493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356845 c.1603G>T p.A535S Chr.11: 63631057-63631057 on Build GRCh38
6487696 c.1564C>G p.Q522E Chr.11: 63631096-63631096 on Build GRCh38
6358103 c.1466A>G p.Y489C Chr.11: 63631113-63631113 on Build GRCh38
6362468 c.1536G>T p.E512D Chr.11: 63631043-63631043 on Build GRCh38
3451303 c.1489C>T p.R497C Chr.11: 63631090-63631090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373239913 Chr.11: 63631090-63631090 on Build GRCh38
rs183951324 Chr.11: 63631057-63631057 on Build GRCh38
rs192770663 Chr.11: 63630984-63630984 on Build GRCh38
rs148102000 Chr.11: 63631024-63631026 on Build GRCh38
rs887938036 Chr.11: 63630963-63630963 on Build GRCh38
rs202227458 Chr.11: 63631079-63631079 on Build GRCh38
rs112847445 Chr.11: 63631078-63631078 on Build GRCh38
rs753236635 Chr.11: 63631051-63631051 on Build GRCh38
rs375456455 Chr.11: 63631029-63631031 on Build GRCh38
rs370704908 Chr.11: 63631127-63631127 on Build GRCh38
rs997195989 Chr.11: 63631022-63631022 on Build GRCh38
rs1036600969 Chr.11: 63630994-63630994 on Build GRCh38
rs1028638609 Chr.11: 63631053-63631053 on Build GRCh38
rs988042420 Chr.11: 63631107-63631107 on Build GRCh38
rs1005530746 Chr.11: 63631072-63631072 on Build GRCh38
rs946357359 Chr.11: 63630932-63630932 on Build GRCh38
rs773136140 Chr.11: 63631128-63631128 on Build GRCh38
rs956743740 Chr.11: 63631104-63631104 on Build GRCh38
rs377140030 Chr.11: 63631121-63631121 on Build GRCh38
rs748101985 Chr.11: 63631111-63631111 on Build GRCh38
rs771573931 Chr.11: 63631002-63631002 on Build GRCh38
rs200026815 Chr.11: 63631058-63631058 on Build GRCh38
rs1002281378 Chr.11: 63631094-63631094 on Build GRCh38
rs760115626 Chr.11: 63631079-63631080 on Build GRCh38
rs767450095 Chr.11: 63631005-63631005 on Build GRCh38
rs913560093 Chr.11: 63630922-63630922 on Build GRCh38
rs369705829 Chr.11: 63631030-63631030 on Build GRCh38
rs769323613 Chr.11: 63631060-63631060 on Build GRCh38
rs940994445 Chr.11: 63630983-63630983 on Build GRCh38
rs375726105 Chr.11: 63631011-63631011 on Build GRCh38
rs753832259 Chr.11: 63631027-63631027 on Build GRCh38
rs910325856 Chr.11: 63630968-63630968 on Build GRCh38
rs766690357 Chr.11: 63631026-63631026 on Build GRCh38
rs768233696 Chr.11: 63631086-63631086 on Build GRCh38
rs779242170 Chr.11: 63631040-63631040 on Build GRCh38
rs979170555 Chr.11: 63630940-63630940 on Build GRCh38
rs528048400 Chr.11: 63631056-63631056 on Build GRCh38
rs750605896 Chr.11: 63631006-63631006 on Build GRCh38

View Full Product Details