Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129829148-129829383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf78
Entrez Gene ID
51759
Gene Name
chromosome 9 open reading frame 78
Gene Aliases
HCA59, HSPC220, bA409K20.3
UniGene
Hs.278429
Gene Chromosome Location
Chr.9: 129827285-129835293 on Build GRCh38
Transcripts
NM_016520.2 XM_011518780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596412 c.724_730delGAGCCCA p.E242fs*5 Chr.9: 129829253-129829259 on Build GRCh38
6455039 c.762G>A p.E254E Chr.9: 129829221-129829221 on Build GRCh38
3654831 c.685C>T p.H229Y Chr.9: 129829298-129829298 on Build GRCh38
1460438 c.723_725delAGA p.E242delE Chr.9: 129829258-129829260 on Build GRCh38
1105882 c.778C>T p.R260W Chr.9: 129829205-129829205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773890545 Chr.9: 129829352-129829352 on Build GRCh38
rs777087140 Chr.9: 129829189-129829189 on Build GRCh38
rs761952121 Chr.9: 129829255-129829255 on Build GRCh38
rs767660346 Chr.9: 129829342-129829342 on Build GRCh38
rs139248631 Chr.9: 129829282-129829282 on Build GRCh38
rs773375419 Chr.9: 129829330-129829330 on Build GRCh38
rs373452546 Chr.9: 129829284-129829284 on Build GRCh38
rs374249652 Chr.9: 129829185-129829185 on Build GRCh38
rs769525413 Chr.9: 129829225-129829225 on Build GRCh38
rs745707856 Chr.9: 129829212-129829212 on Build GRCh38
rs758042858 Chr.9: 129829283-129829283 on Build GRCh38
rs756733770 Chr.9: 129829298-129829298 on Build GRCh38
rs199630059 Chr.9: 129829273-129829273 on Build GRCh38
rs750944606 Chr.9: 129829274-129829274 on Build GRCh38
rs140049867 Chr.9: 129829205-129829205 on Build GRCh38
rs772308905 Chr.9: 129829329-129829329 on Build GRCh38
rs376381202 Chr.9: 129829325-129829325 on Build GRCh38
rs199549692 Chr.9: 129829174-129829174 on Build GRCh38
rs768829061 Chr.9: 129829334-129829334 on Build GRCh38
rs748285215 Chr.9: 129829307-129829307 on Build GRCh38
rs201417433 Chr.9: 129829192-129829192 on Build GRCh38
rs967599854 Chr.9: 129829259-129829259 on Build GRCh38
rs763016070 Chr.9: 129829245-129829245 on Build GRCh38
rs200595056 Chr.9: 129829251-129829251 on Build GRCh38
rs910607987 Chr.9: 129829351-129829351 on Build GRCh38
rs977601405 Chr.9: 129829260-129829260 on Build GRCh38
rs756571102 Chr.9: 129829275-129829275 on Build GRCh38
rs1007823265 Chr.9: 129829317-129829317 on Build GRCh38
rs746537716 Chr.9: 129829286-129829286 on Build GRCh38
rs3818553 Chr.9: 129829230-129829230 on Build GRCh38
rs200922853 Chr.9: 129829289-129829289 on Build GRCh38
rs764250406 Chr.9: 129829246-129829246 on Build GRCh38
rs769579469 Chr.9: 129829312-129829312 on Build GRCh38
rs761852463 Chr.9: 129829337-129829337 on Build GRCh38
rs372860206 Chr.9: 129829182-129829182 on Build GRCh38
rs766811260 Chr.9: 129829281-129829281 on Build GRCh38
rs774602738 Chr.9: 129829335-129829335 on Build GRCh38
rs764968548 Chr.9: 129829263-129829265 on Build GRCh38
rs376310519 Chr.9: 129829184-129829184 on Build GRCh38
rs771646091 Chr.9: 129829186-129829186 on Build GRCh38
rs770840294 Chr.9: 129829195-129829195 on Build GRCh38
rs750888164 Chr.9: 129829173-129829173 on Build GRCh38

View Full Product Details