Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 121112584-121112833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D32
Entrez Gene ID
221322
Gene Name
TBC1 domain family member 32
Gene Aliases
BROMI, C6orf170, C6orf171, bA301B7.2, bA57L9.1, dJ310J6.1
UniGene
Hs.121396
Gene Chromosome Location
Chr.6: 121079494-121335382 on Build GRCh38
Transcripts
NM_152730.5 NR_104452.1 XM_005266861.2 XM_011535569.1 XM_011535570.1 XM_011535571.2 XM_011535574.1 XM_011535580.2 XM_017010397.1 XM_017010398.1 XM_017010399.1 XM_017010400.1 XM_017010401.1 XM_017010404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797431 c.3178C>T p.Q1060* Chr.6: 121112651-121112651 on Build GRCh38
6797432 c.3176T>C p.L1059P Chr.6: 121112653-121112653 on Build GRCh38
6797433 c.3177G>C p.L1059L Chr.6: 121112652-121112652 on Build GRCh38
6529928 c.3180A>G p.Q1060Q Chr.6: 121112649-121112649 on Build GRCh38
6567556 c.3183G>A p.G1061G Chr.6: 121112646-121112646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054333876 Chr.6: 121112645-121112645 on Build GRCh38
rs765216265 Chr.6: 121112689-121112689 on Build GRCh38
rs781461289 Chr.6: 121112616-121112616 on Build GRCh38
rs919574998 Chr.6: 121112687-121112687 on Build GRCh38
rs777791020 Chr.6: 121112628-121112628 on Build GRCh38
rs768299017 Chr.6: 121112655-121112655 on Build GRCh38
rs775173488 Chr.6: 121112652-121112652 on Build GRCh38
rs888076868 Chr.6: 121112773-121112773 on Build GRCh38
rs746242313 Chr.6: 121112644-121112644 on Build GRCh38
rs375903914 Chr.6: 121112777-121112777 on Build GRCh38
rs776219743 Chr.6: 121112676-121112676 on Build GRCh38
rs748226334 Chr.6: 121112617-121112617 on Build GRCh38
rs199671235 Chr.6: 121112647-121112647 on Build GRCh38
rs982939087 Chr.6: 121112729-121112729 on Build GRCh38
rs762785589 Chr.6: 121112700-121112700 on Build GRCh38
rs943879946 Chr.6: 121112764-121112764 on Build GRCh38
rs760360569 Chr.6: 121112653-121112653 on Build GRCh38
rs756157845 Chr.6: 121112620-121112620 on Build GRCh38
rs548490962 Chr.6: 121112634-121112634 on Build GRCh38
rs912276191 Chr.6: 121112730-121112730 on Build GRCh38
rs371460758 Chr.6: 121112632-121112632 on Build GRCh38
rs367648371 Chr.6: 121112692-121112692 on Build GRCh38
rs761845055 Chr.6: 121112683-121112683 on Build GRCh38
rs375947439 Chr.6: 121112639-121112639 on Build GRCh38

View Full Product Details