Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26463375-26463630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A1
Entrez Gene ID
11120
Gene Name
butyrophilin subfamily 2 member A1
Gene Aliases
BK14H9.1, BT2.1, BTF1, BTN2.1, DJ3E1.1
UniGene
Hs.159028
Gene Chromosome Location
Chr.6: 26457904-26476621 on Build GRCh38
Transcripts
NM_001197233.2 NM_001197234.2 NM_007049.4 NM_078476.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613502 c.649A>G p.M217V Chr.6: 26463462-26463462 on Build GRCh38
6613501 c.649A>G p.M217V Chr.6: 26463462-26463462 on Build GRCh38
6485939 c.651G>A p.M217I Chr.6: 26463464-26463464 on Build GRCh38
6485938 c.651G>A p.M217I Chr.6: 26463464-26463464 on Build GRCh38
5809249 c.661A>T p.I221F Chr.6: 26463474-26463474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775554453 Chr.6: 26463416-26463416 on Build GRCh38
rs757153020 Chr.6: 26463472-26463473 on Build GRCh38
rs368095730 Chr.6: 26463539-26463539 on Build GRCh38
rs571328001 Chr.6: 26463403-26463403 on Build GRCh38
rs371521117 Chr.6: 26463463-26463463 on Build GRCh38
rs142538560 Chr.6: 26463503-26463503 on Build GRCh38
rs374138534 Chr.6: 26463407-26463407 on Build GRCh38
rs13195509 Chr.6: 26463432-26463432 on Build GRCh38
rs767895715 Chr.6: 26463448-26463448 on Build GRCh38
rs756808547 Chr.6: 26463566-26463566 on Build GRCh38
rs763157667 Chr.6: 26463425-26463425 on Build GRCh38
rs950767630 Chr.6: 26463572-26463572 on Build GRCh38
rs750733328 Chr.6: 26463455-26463455 on Build GRCh38
rs72500818 Chr.6: 26463550-26463550 on Build GRCh38
rs746933028 Chr.6: 26463396-26463396 on Build GRCh38
rs552507552 Chr.6: 26463494-26463494 on Build GRCh38
rs371634109 Chr.6: 26463398-26463398 on Build GRCh38
rs756120881 Chr.6: 26463576-26463577 on Build GRCh38
rs753374732 Chr.6: 26463540-26463540 on Build GRCh38
rs138727076 Chr.6: 26463464-26463464 on Build GRCh38
rs920767570 Chr.6: 26463422-26463422 on Build GRCh38
rs375240672 Chr.6: 26463427-26463427 on Build GRCh38
rs559126711 Chr.6: 26463594-26463594 on Build GRCh38
rs777364003 Chr.6: 26463487-26463487 on Build GRCh38
rs770952034 Chr.6: 26463492-26463492 on Build GRCh38
rs890529838 Chr.6: 26463583-26463584 on Build GRCh38
rs535843474 Chr.6: 26463491-26463491 on Build GRCh38
rs182643948 Chr.6: 26463560-26463560 on Build GRCh38
rs750861051 Chr.6: 26463553-26463553 on Build GRCh38
rs747753643 Chr.6: 26463476-26463476 on Build GRCh38
rs750498544 Chr.6: 26463506-26463509 on Build GRCh38
rs751068433 Chr.6: 26463564-26463564 on Build GRCh38
rs752073488 Chr.6: 26463428-26463428 on Build GRCh38
rs992880895 Chr.6: 26463586-26463586 on Build GRCh38
rs773676526 Chr.6: 26463596-26463596 on Build GRCh38
rs764163602 Chr.6: 26463530-26463530 on Build GRCh38
rs757988172 Chr.6: 26463484-26463484 on Build GRCh38
rs900465450 Chr.6: 26463609-26463609 on Build GRCh38
rs768058892 Chr.6: 26463551-26463551 on Build GRCh38
rs375972503 Chr.6: 26463559-26463559 on Build GRCh38
rs779689829 Chr.6: 26463535-26463535 on Build GRCh38
rs781310146 Chr.6: 26463481-26463482 on Build GRCh38
rs144358349 Chr.6: 26463408-26463408 on Build GRCh38
rs781321601 Chr.6: 26463573-26463573 on Build GRCh38
rs766642614 Chr.6: 26463554-26463554 on Build GRCh38
rs776502299 Chr.6: 26463404-26463404 on Build GRCh38
rs777461374 Chr.6: 26463563-26463563 on Build GRCh38
rs199893249 Chr.6: 26463413-26463413 on Build GRCh38
rs775838040 Chr.6: 26463501-26463501 on Build GRCh38
rs935921091 Chr.6: 26463542-26463542 on Build GRCh38
rs754672073 Chr.6: 26463595-26463595 on Build GRCh38
rs770867844 Chr.6: 26463401-26463401 on Build GRCh38
rs760144826 Chr.6: 26463398-26463404 on Build GRCh38

View Full Product Details