Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123328061-123328286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2
Entrez Gene ID
29028
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
ANCCA, CT137, PRO2000
UniGene
Hs.370834
Gene Chromosome Location
Chr.8: 123319850-123396992 on Build GRCh38
Transcripts
NM_014109.3 XM_011516994.2 XM_011516996.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274063 c.3808T>C p.C1270R Chr.8: 123328250-123328250 on Build GRCh38
749491 c.3792G>C p.L1264F Chr.8: 123328266-123328266 on Build GRCh38
77304 c.3823T>A p.S1275T Chr.8: 123328235-123328235 on Build GRCh38
6179570 c.3854A>T p.E1285V Chr.8: 123328204-123328204 on Build GRCh38
4915055 c.3850G>A p.D1284N Chr.8: 123328208-123328208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893405937 Chr.8: 123328234-123328234 on Build GRCh38
rs777397334 Chr.8: 123328222-123328222 on Build GRCh38
rs772510886 Chr.8: 123328186-123328187 on Build GRCh38
rs978489225 Chr.8: 123328105-123328105 on Build GRCh38
rs767834213 Chr.8: 123328173-123328173 on Build GRCh38
rs770943105 Chr.8: 123328233-123328233 on Build GRCh38
rs545358720 Chr.8: 123328099-123328099 on Build GRCh38
rs1051653132 Chr.8: 123328178-123328178 on Build GRCh38
rs527682440 Chr.8: 123328186-123328186 on Build GRCh38
rs368107495 Chr.8: 123328185-123328185 on Build GRCh38
rs746901456 Chr.8: 123328223-123328223 on Build GRCh38
rs778550385 Chr.8: 123328216-123328216 on Build GRCh38
rs776527575 Chr.8: 123328243-123328243 on Build GRCh38
rs752845367 Chr.8: 123328131-123328131 on Build GRCh38
rs922422211 Chr.8: 123328165-123328165 on Build GRCh38
rs757828918 Chr.8: 123328147-123328147 on Build GRCh38
rs556393688 Chr.8: 123328138-123328138 on Build GRCh38
rs762247864 Chr.8: 123328153-123328154 on Build GRCh38
rs369321666 Chr.8: 123328217-123328217 on Build GRCh38
rs745680844 Chr.8: 123328246-123328246 on Build GRCh38
rs79894171 Chr.8: 123328221-123328221 on Build GRCh38
rs3758122 Chr.8: 123328219-123328219 on Build GRCh38
rs1040784133 Chr.8: 123328145-123328145 on Build GRCh38

View Full Product Details