Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228102169-228102406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
UniGene
Hs.445952
Gene Chromosome Location
Chr.1: 228100727-228103462 on Build GRCh38
Transcripts
NM_024319.3 NR_130975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295953 c.404G>C p.R135P Chr.1: 228102353-228102353 on Build GRCh38
4028855 c.382G>A p.V128M Chr.1: 228102375-228102375 on Build GRCh38
7358300 c.425A>G p.K142R Chr.1: 228102332-228102332 on Build GRCh38
6009843 c.439G>T p.V147L Chr.1: 228102318-228102318 on Build GRCh38
6124798 c.437C>G p.S146C Chr.1: 228102320-228102320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369194534 Chr.1: 228102324-228102324 on Build GRCh38
rs956940702 Chr.1: 228102191-228102191 on Build GRCh38
rs773858150 Chr.1: 228102281-228102281 on Build GRCh38
rs768292808 Chr.1: 228102370-228102370 on Build GRCh38
rs947909071 Chr.1: 228102301-228102301 on Build GRCh38
rs578158660 Chr.1: 228102223-228102223 on Build GRCh38
rs758317136 Chr.1: 228102215-228102215 on Build GRCh38
rs1037634609 Chr.1: 228102288-228102288 on Build GRCh38
rs1030007746 Chr.1: 228102305-228102305 on Build GRCh38
rs982769714 Chr.1: 228102262-228102262 on Build GRCh38
rs1039400723 Chr.1: 228102218-228102218 on Build GRCh38
rs778552072 Chr.1: 228102375-228102375 on Build GRCh38
rs777713372 Chr.1: 228102261-228102261 on Build GRCh38
rs777130616 Chr.1: 228102290-228102290 on Build GRCh38
rs781000351 Chr.1: 228102339-228102339 on Build GRCh38
rs763126994 Chr.1: 228102321-228102321 on Build GRCh38
rs779475958 Chr.1: 228102271-228102271 on Build GRCh38
rs528084635 Chr.1: 228102295-228102295 on Build GRCh38
rs746911412 Chr.1: 228102264-228102264 on Build GRCh38
rs145743393 Chr.1: 228102350-228102350 on Build GRCh38
rs768239498 Chr.1: 228102279-228102279 on Build GRCh38
rs1011133799 Chr.1: 228102198-228102198 on Build GRCh38
rs752792503 Chr.1: 228102207-228102207 on Build GRCh38
rs758155115 Chr.1: 228102285-228102292 on Build GRCh38
rs756850969 Chr.1: 228102204-228102205 on Build GRCh38
rs757053740 Chr.1: 228102265-228102265 on Build GRCh38
rs942727658 Chr.1: 228102354-228102354 on Build GRCh38
rs765301504 Chr.1: 228102204-228102204 on Build GRCh38
rs755215152 Chr.1: 228102203-228102203 on Build GRCh38
rs61825155 Chr.1: 228102332-228102332 on Build GRCh38
rs901749116 Chr.1: 228102219-228102219 on Build GRCh38
rs762065505 Chr.1: 228102270-228102271 on Build GRCh38
rs558352129 Chr.1: 228102192-228102192 on Build GRCh38
rs750207588 Chr.1: 228102344-228102344 on Build GRCh38
rs754125957 Chr.1: 228102199-228102199 on Build GRCh38
rs780516497 Chr.1: 228102317-228102320 on Build GRCh38
rs753286442 Chr.1: 228102199-228102200 on Build GRCh38
rs920409852 Chr.1: 228102292-228102292 on Build GRCh38
rs1011907889 Chr.1: 228102272-228102272 on Build GRCh38
rs902280027 Chr.1: 228102248-228102248 on Build GRCh38
rs906248662 Chr.1: 228102328-228102328 on Build GRCh38
rs955546434 Chr.1: 228102258-228102258 on Build GRCh38
rs922747896 Chr.1: 228102255-228102255 on Build GRCh38
rs769453961 Chr.1: 228102269-228102269 on Build GRCh38
rs2295995 Chr.1: 228102275-228102275 on Build GRCh38
rs752814685 Chr.1: 228102304-228102304 on Build GRCh38
rs540925037 Chr.1: 228102232-228102232 on Build GRCh38
rs755813107 Chr.1: 228102349-228102349 on Build GRCh38
rs780910425 Chr.1: 228102266-228102266 on Build GRCh38
rs765476215 Chr.1: 228102298-228102298 on Build GRCh38
rs975448763 Chr.1: 228102233-228102233 on Build GRCh38
rs761205135 Chr.1: 228102282-228102282 on Build GRCh38
rs771479442 Chr.1: 228102286-228102286 on Build GRCh38
rs899673663 Chr.1: 228102213-228102213 on Build GRCh38
rs373950653 Chr.1: 228102353-228102353 on Build GRCh38
rs764775944 Chr.1: 228102260-228102261 on Build GRCh38

View Full Product Details