Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17743730-17743971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BID
Entrez Gene ID
637
Gene Name
BH3 interacting domain death agonist
Gene Aliases
FP497
UniGene
Hs.517145
Gene Chromosome Location
Chr.22: 17734140-17774665 on Build GRCh38
Transcripts
NM_001196.3 NM_001244567.1 NM_001244569.1 NM_001244570.1 NM_001244572.1 NM_197966.2 NM_197967.2 XM_017028906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116387 c.329G>C p.S110T Chr.22: 17743835-17743835 on Build GRCh38
3785603 c.201C>T p.S67S Chr.22: 17743825-17743825 on Build GRCh38
3964075 c.223+6C>T p.? Chr.22: 17743797-17743797 on Build GRCh38
3964073 c.223+7G>T p.? Chr.22: 17743796-17743796 on Build GRCh38
3708124 c.190A>T p.S64C Chr.22: 17743836-17743836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189923680 Chr.22: 17743942-17743942 on Build GRCh38
rs752491643 Chr.22: 17743775-17743775 on Build GRCh38
rs772763406 Chr.22: 17743887-17743887 on Build GRCh38
rs978578139 Chr.22: 17743786-17743786 on Build GRCh38
rs747016844 Chr.22: 17743899-17743899 on Build GRCh38
rs776679558 Chr.22: 17743905-17743905 on Build GRCh38
rs563580878 Chr.22: 17743764-17743764 on Build GRCh38
rs750006117 Chr.22: 17743852-17743852 on Build GRCh38
rs547029088 Chr.22: 17743864-17743864 on Build GRCh38
rs200409062 Chr.22: 17743912-17743912 on Build GRCh38
rs768886503 Chr.22: 17743835-17743836 on Build GRCh38
rs529450978 Chr.22: 17743784-17743784 on Build GRCh38
rs374324393 Chr.22: 17743796-17743796 on Build GRCh38
rs760132280 Chr.22: 17743925-17743925 on Build GRCh38
rs761738989 Chr.22: 17743918-17743918 on Build GRCh38
rs767379821 Chr.22: 17743922-17743922 on Build GRCh38
rs755255176 Chr.22: 17743869-17743869 on Build GRCh38
rs748577760 Chr.22: 17743881-17743881 on Build GRCh38
rs371839796 Chr.22: 17743793-17743793 on Build GRCh38
rs984226517 Chr.22: 17743802-17743802 on Build GRCh38
rs771344517 Chr.22: 17743838-17743838 on Build GRCh38
rs11542850 Chr.22: 17743878-17743878 on Build GRCh38
rs61757628 Chr.22: 17743824-17743824 on Build GRCh38
rs779372119 Chr.22: 17743872-17743872 on Build GRCh38
rs372024838 Chr.22: 17743911-17743911 on Build GRCh38
rs749551829 Chr.22: 17743819-17743819 on Build GRCh38
rs754705127 Chr.22: 17743936-17743936 on Build GRCh38
rs922447901 Chr.22: 17743854-17743854 on Build GRCh38
rs753398808 Chr.22: 17743933-17743933 on Build GRCh38
rs938056280 Chr.22: 17743801-17743801 on Build GRCh38
rs753553004 Chr.22: 17743868-17743868 on Build GRCh38
rs756092459 Chr.22: 17743841-17743841 on Build GRCh38
rs738094 Chr.22: 17743755-17743755 on Build GRCh38
rs2072392 Chr.22: 17743846-17743846 on Build GRCh38
rs201438109 Chr.22: 17743835-17743835 on Build GRCh38
rs952377551 Chr.22: 17743798-17743798 on Build GRCh38
rs2072393 Chr.22: 17743762-17743762 on Build GRCh38
rs759844569 Chr.22: 17743910-17743910 on Build GRCh38
rs760965442 Chr.22: 17743837-17743837 on Build GRCh38
rs764590275 Chr.22: 17743926-17743926 on Build GRCh38
rs777994982 Chr.22: 17743889-17743889 on Build GRCh38
rs769081918 Chr.22: 17743823-17743823 on Build GRCh38
rs767725706 Chr.22: 17743753-17743753 on Build GRCh38
rs199617244 Chr.22: 17743923-17743923 on Build GRCh38
rs906261272 Chr.22: 17743771-17743771 on Build GRCh38
rs780397060 Chr.22: 17743797-17743797 on Build GRCh38
rs773469812 Chr.22: 17743836-17743836 on Build GRCh38
rs879161693 Chr.22: 17743856-17743856 on Build GRCh38
rs543334573 Chr.22: 17743792-17743792 on Build GRCh38
rs749846664 Chr.22: 17743798-17743798 on Build GRCh38
rs908249052 Chr.22: 17743939-17743939 on Build GRCh38
rs765102403 Chr.22: 17743765-17743765 on Build GRCh38
rs748717824 Chr.22: 17743831-17743831 on Build GRCh38
rs150608132 Chr.22: 17743900-17743900 on Build GRCh38
rs377243261 Chr.22: 17743838-17743839 on Build GRCh38
rs755629886 Chr.22: 17743853-17743853 on Build GRCh38

View Full Product Details