Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57250350-57250575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2BP
Entrez Gene ID
23568
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
Gene Aliases
BART, BART1, RP66
UniGene
Hs.632873
Gene Chromosome Location
Chr.16: 57245126-57253635 on Build GRCh38
Transcripts
NM_012106.3 XM_011522977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6226936 c.294-9C>T p.? Chr.16: 57250402-57250402 on Build GRCh38
971816 c.367G>T p.E123* Chr.16: 57250484-57250484 on Build GRCh38
6144741 c.386G>A p.R129K Chr.16: 57250503-57250503 on Build GRCh38
5623461 c.318T>G p.G106G Chr.16: 57250435-57250435 on Build GRCh38
5971830 c.301A>G p.K101E Chr.16: 57250418-57250418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372794991 Chr.16: 57250475-57250475 on Build GRCh38
rs374438905 Chr.16: 57250493-57250493 on Build GRCh38
rs758528608 Chr.16: 57250448-57250448 on Build GRCh38
rs1031519012 Chr.16: 57250374-57250374 on Build GRCh38
rs762805709 Chr.16: 57250401-57250401 on Build GRCh38
rs759227898 Chr.16: 57250399-57250399 on Build GRCh38
rs754081066 Chr.16: 57250474-57250474 on Build GRCh38
rs748674247 Chr.16: 57250525-57250525 on Build GRCh38
rs780317166 Chr.16: 57250378-57250378 on Build GRCh38
rs183238176 Chr.16: 57250464-57250464 on Build GRCh38
rs761681643 Chr.16: 57250437-57250437 on Build GRCh38
rs775105436 Chr.16: 57250420-57250420 on Build GRCh38
rs151111778 Chr.16: 57250429-57250429 on Build GRCh38
rs1047657310 Chr.16: 57250416-57250416 on Build GRCh38
rs374945556 Chr.16: 57250403-57250403 on Build GRCh38
rs1057111225 Chr.16: 57250445-57250445 on Build GRCh38
rs770859200 Chr.16: 57250519-57250521 on Build GRCh38
rs765259582 Chr.16: 57250402-57250402 on Build GRCh38
rs772389298 Chr.16: 57250532-57250532 on Build GRCh38
rs750251288 Chr.16: 57250444-57250444 on Build GRCh38
rs201339545 Chr.16: 57250435-57250435 on Build GRCh38
rs370623214 Chr.16: 57250379-57250379 on Build GRCh38
rs767350325 Chr.16: 57250440-57250440 on Build GRCh38
rs777916280 Chr.16: 57250545-57250545 on Build GRCh38
rs143902334 Chr.16: 57250430-57250430 on Build GRCh38
rs759373737 Chr.16: 57250396-57250396 on Build GRCh38
rs887254680 Chr.16: 57250439-57250439 on Build GRCh38
rs768035815 Chr.16: 57250377-57250377 on Build GRCh38
rs780186728 Chr.16: 57250454-57250454 on Build GRCh38
rs368436972 Chr.16: 57250385-57250385 on Build GRCh38
rs772506259 Chr.16: 57250389-57250389 on Build GRCh38
rs778332995 Chr.16: 57250534-57250534 on Build GRCh38
rs894569070 Chr.16: 57250500-57250500 on Build GRCh38

View Full Product Details