Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116393380-116393621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027646.1

Gene Symbol
ATP1A1-AS1
Entrez Gene ID
84852
Gene Name
ATP1A1 antisense RNA 1
Gene Aliases
ATP1A1OS, C1orf203
UniGene
Hs.664752
Gene Chromosome Location
Chr.1: 116392865-116418622 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3740562 c.1542C>T p.I514I Chr.1: 116393605-116393605 on Build GRCh38
1172010 c.1511T>C p.L504P Chr.1: 116393574-116393574 on Build GRCh38
3801397 c.1522A>C p.K508Q Chr.1: 116393585-116393585 on Build GRCh38
423544 c.1553G>A p.C518Y Chr.1: 116393616-116393616 on Build GRCh38
3801398 c.1534G>A p.E512K Chr.1: 116393597-116393597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968592287 Chr.1: 116393491-116393491 on Build GRCh38
rs11540953 Chr.1: 116393560-116393560 on Build GRCh38
rs139336321 Chr.1: 116393446-116393446 on Build GRCh38
rs372221522 Chr.1: 116393554-116393554 on Build GRCh38
rs772726777 Chr.1: 116393480-116393480 on Build GRCh38
rs767656830 Chr.1: 116393580-116393580 on Build GRCh38
rs759865265 Chr.1: 116393499-116393499 on Build GRCh38
rs1020133696 Chr.1: 116393450-116393450 on Build GRCh38
rs367957404 Chr.1: 116393590-116393590 on Build GRCh38
rs752825876 Chr.1: 116393509-116393509 on Build GRCh38
rs534034408 Chr.1: 116393519-116393519 on Build GRCh38
rs775427851 Chr.1: 116393548-116393563 on Build GRCh38
rs760952426 Chr.1: 116393487-116393487 on Build GRCh38
rs903586824 Chr.1: 116393449-116393449 on Build GRCh38
rs776551345 Chr.1: 116393489-116393489 on Build GRCh38
rs768214820 Chr.1: 116393568-116393568 on Build GRCh38
rs764054683 Chr.1: 116393523-116393523 on Build GRCh38
rs771333251 Chr.1: 116393575-116393575 on Build GRCh38
rs773813096 Chr.1: 116393485-116393485 on Build GRCh38
rs751451407 Chr.1: 116393549-116393549 on Build GRCh38
rs141400264 Chr.1: 116393559-116393559 on Build GRCh38
rs1004153266 Chr.1: 116393566-116393566 on Build GRCh38
rs144872101 Chr.1: 116393572-116393572 on Build GRCh38
rs962820956 Chr.1: 116393578-116393578 on Build GRCh38
rs748836037 Chr.1: 116393563-116393563 on Build GRCh38
rs570274541 Chr.1: 116393436-116393436 on Build GRCh38
rs765411583 Chr.1: 116393501-116393501 on Build GRCh38
rs759588556 Chr.1: 116393510-116393510 on Build GRCh38
rs778154659 Chr.1: 116393570-116393570 on Build GRCh38
rs548753797 Chr.1: 116393415-116393415 on Build GRCh38
rs779802423 Chr.1: 116393561-116393561 on Build GRCh38
rs200995911 Chr.1: 116393522-116393522 on Build GRCh38
rs944994318 Chr.1: 116393425-116393425 on Build GRCh38
rs150035947 Chr.1: 116393498-116393498 on Build GRCh38
rs1010040437 Chr.1: 116393447-116393447 on Build GRCh38
rs1033172330 Chr.1: 116393477-116393477 on Build GRCh38

View Full Product Details