Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102876821-102877094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AS3MT
Entrez Gene ID
57412
Gene Name
arsenite methyltransferase
Gene Aliases
CYT19
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102869453-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2059545 c.571G>T p.E191* Chr.10: 102876996-102876996 on Build GRCh38
5369984 c.545A>T p.Y182F Chr.10: 102876970-102876970 on Build GRCh38
6855300 c.600A>C p.K200N Chr.10: 102877025-102877025 on Build GRCh38
6605406 c.563C>T p.T188M Chr.10: 102876988-102876988 on Build GRCh38
914269 c.583G>T p.E195* Chr.10: 102877008-102877008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777704958 Chr.10: 102877072-102877072 on Build GRCh38
rs1004089439 Chr.10: 102876880-102876880 on Build GRCh38
rs774767962 Chr.10: 102877023-102877023 on Build GRCh38
rs948619969 Chr.10: 102876867-102876867 on Build GRCh38
rs747221003 Chr.10: 102876969-102876969 on Build GRCh38
rs775763434 Chr.10: 102877023-102877024 on Build GRCh38
rs377312367 Chr.10: 102876992-102876992 on Build GRCh38
rs763450550 Chr.10: 102877016-102877016 on Build GRCh38
rs370022454 Chr.10: 102877034-102877034 on Build GRCh38
rs758468005 Chr.10: 102877071-102877071 on Build GRCh38
rs745981957 Chr.10: 102876988-102876988 on Build GRCh38
rs753331760 Chr.10: 102876934-102876934 on Build GRCh38
rs373776846 Chr.10: 102877044-102877044 on Build GRCh38
rs752623913 Chr.10: 102877070-102877070 on Build GRCh38
rs758201841 Chr.10: 102876963-102876963 on Build GRCh38
rs576986384 Chr.10: 102876981-102876981 on Build GRCh38
rs376202918 Chr.10: 102877048-102877048 on Build GRCh38
rs376758155 Chr.10: 102876906-102876906 on Build GRCh38
rs565707676 Chr.10: 102876927-102876927 on Build GRCh38
rs946259068 Chr.10: 102876991-102876991 on Build GRCh38
rs556211745 Chr.10: 102877018-102877018 on Build GRCh38
rs778669383 Chr.10: 102876951-102876951 on Build GRCh38
rs753552796 Chr.10: 102877037-102877037 on Build GRCh38
rs868176114 Chr.10: 102877030-102877030 on Build GRCh38
rs894203244 Chr.10: 102876865-102876865 on Build GRCh38
rs374230034 Chr.10: 102876980-102876980 on Build GRCh38
rs17883232 Chr.10: 102876944-102876944 on Build GRCh38
rs1048695845 Chr.10: 102876876-102876876 on Build GRCh38
rs3740393 Chr.10: 102876898-102876898 on Build GRCh38
rs558829089 Chr.10: 102876968-102876968 on Build GRCh38
rs775931783 Chr.10: 102876997-102876997 on Build GRCh38
rs190534777 Chr.10: 102876889-102876889 on Build GRCh38
rs888246986 Chr.10: 102876878-102876878 on Build GRCh38
rs748128749 Chr.10: 102876955-102876955 on Build GRCh38
rs755784439 Chr.10: 102876907-102876907 on Build GRCh38

View Full Product Details