Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66340547-66340820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRMS1
Entrez Gene ID
25855
Gene Name
breast cancer metastasis suppressor 1
Gene Aliases
-
UniGene
Hs.100426
Gene Chromosome Location
Chr.11: 66337333-66345111 on Build GRCh38
Transcripts
NM_001024957.1 NM_015399.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1322241 c.503A>C p.E168A Chr.11: 66340806-66340806 on Build GRCh38
1322242 c.503A>C p.E168A Chr.11: 66340806-66340806 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774560947 Chr.11: 66340613-66340613 on Build GRCh38
rs1019704759 Chr.11: 66340621-66340621 on Build GRCh38
rs564839726 Chr.11: 66340587-66340587 on Build GRCh38
rs17850564 Chr.11: 66340784-66340784 on Build GRCh38
rs1010117479 Chr.11: 66340714-66340714 on Build GRCh38
rs549457089 Chr.11: 66340715-66340715 on Build GRCh38
rs756661112 Chr.11: 66340749-66340749 on Build GRCh38
rs569385160 Chr.11: 66340716-66340716 on Build GRCh38
rs200297342 Chr.11: 66340776-66340776 on Build GRCh38
rs529599194 Chr.11: 66340712-66340712 on Build GRCh38
rs908136889 Chr.11: 66340789-66340789 on Build GRCh38
rs1008772825 Chr.11: 66340612-66340612 on Build GRCh38
rs367980384 Chr.11: 66340766-66340766 on Build GRCh38
rs527247978 Chr.11: 66340622-66340622 on Build GRCh38
rs199827924 Chr.11: 66340761-66340761 on Build GRCh38
rs368698243 Chr.11: 66340768-66340768 on Build GRCh38
rs893054806 Chr.11: 66340668-66340668 on Build GRCh38
rs551815579 Chr.11: 66340736-66340736 on Build GRCh38
rs1032670760 Chr.11: 66340644-66340644 on Build GRCh38
rs951595921 Chr.11: 66340592-66340592 on Build GRCh38
rs560897571 Chr.11: 66340669-66340669 on Build GRCh38
rs770936650 Chr.11: 66340783-66340783 on Build GRCh38
rs762805402 Chr.11: 66340788-66340788 on Build GRCh38
rs1035085497 Chr.11: 66340756-66340756 on Build GRCh38
rs995783107 Chr.11: 66340639-66340639 on Build GRCh38
rs763829379 Chr.11: 66340725-66340725 on Build GRCh38
rs748352609 Chr.11: 66340767-66340767 on Build GRCh38
rs200013073 Chr.11: 66340797-66340797 on Build GRCh38
rs758164326 Chr.11: 66340724-66340724 on Build GRCh38
rs751370154 Chr.11: 66340798-66340798 on Build GRCh38
rs371270195 Chr.11: 66340748-66340748 on Build GRCh38
rs971227168 Chr.11: 66340734-66340734 on Build GRCh38
rs575951728 Chr.11: 66340774-66340774 on Build GRCh38
rs964159065 Chr.11: 66340631-66340631 on Build GRCh38
rs749788627 Chr.11: 66340765-66340765 on Build GRCh38
rs547441291 Chr.11: 66340666-66340666 on Build GRCh38
rs538062418 Chr.11: 66340735-66340735 on Build GRCh38

View Full Product Details