Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 82317927-82318180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP1L
Entrez Gene ID
92270
Gene Name
ATPase H+ transporting accessory protein 1 like
Gene Aliases
-
UniGene
Hs.729805
Gene Chromosome Location
Chr.5: 82278645-82344294 on Build GRCh38
Transcripts
NM_001017971.2 XM_011543719.2 XM_011543720.2 XM_011543723.2 XM_017010025.1 XM_017010026.1 XM_017010027.1 XM_017010028.1 XM_017010029.1 XM_017010030.1 XM_017010031.1 XM_017010032.1 XM_017010033.1 XM_017010034.1 XM_017010035.1 XM_017010036.1 XM_017010037.1 XM_017010039.1 XM_017010040.1 XM_017010041.1 XM_017010042.1 XM_017010043.1 XM_017010044.1 XM_017010045.1 XM_017010046.1 XM_017010047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1070767 c.424G>T p.A142S Chr.5: 82318049-82318049 on Build GRCh38
7194887 c.465G>C p.M155I Chr.5: 82318090-82318090 on Build GRCh38
4788148 c.424G>A p.A142T Chr.5: 82318049-82318049 on Build GRCh38
3975647 c.483G>A p.L161L Chr.5: 82318108-82318108 on Build GRCh38
6608003 c.450G>T p.L150L Chr.5: 82318075-82318075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371506768 Chr.5: 82318048-82318048 on Build GRCh38
rs745780740 Chr.5: 82318092-82318121 on Build GRCh38
rs750580645 Chr.5: 82318156-82318156 on Build GRCh38
rs546492707 Chr.5: 82318121-82318121 on Build GRCh38
rs746704637 Chr.5: 82318122-82318122 on Build GRCh38
rs144403166 Chr.5: 82318062-82318062 on Build GRCh38
rs778112304 Chr.5: 82317965-82317965 on Build GRCh38
rs142800418 Chr.5: 82317962-82317962 on Build GRCh38
rs764558806 Chr.5: 82318144-82318144 on Build GRCh38
rs771863968 Chr.5: 82318096-82318096 on Build GRCh38
rs369271171 Chr.5: 82318013-82318013 on Build GRCh38
rs776129355 Chr.5: 82318136-82318136 on Build GRCh38
rs767614630 Chr.5: 82318149-82318149 on Build GRCh38
rs760108631 Chr.5: 82318044-82318044 on Build GRCh38
rs770479126 Chr.5: 82318026-82318026 on Build GRCh38
rs567976526 Chr.5: 82318135-82318135 on Build GRCh38
rs201177049 Chr.5: 82318025-82318025 on Build GRCh38
rs772833883 Chr.5: 82318021-82318021 on Build GRCh38
rs781078854 Chr.5: 82317975-82317975 on Build GRCh38
rs1037241137 Chr.5: 82318126-82318126 on Build GRCh38
rs143947906 Chr.5: 82318140-82318140 on Build GRCh38
rs529723863 Chr.5: 82317968-82317968 on Build GRCh38
rs747904138 Chr.5: 82318019-82318019 on Build GRCh38
rs747992660 Chr.5: 82318093-82318093 on Build GRCh38
rs771776158 Chr.5: 82318020-82318020 on Build GRCh38
rs375164846 Chr.5: 82317999-82317999 on Build GRCh38
rs941292535 Chr.5: 82318108-82318108 on Build GRCh38
rs370223240 Chr.5: 82317956-82317956 on Build GRCh38
rs550926078 Chr.5: 82318146-82318146 on Build GRCh38
rs146594152 Chr.5: 82318078-82318078 on Build GRCh38
rs141342982 Chr.5: 82318130-82318130 on Build GRCh38
rs778726660 Chr.5: 82318018-82318018 on Build GRCh38
rs139291392 Chr.5: 82318051-82318051 on Build GRCh38
rs376603706 Chr.5: 82318049-82318049 on Build GRCh38
rs770439334 Chr.5: 82317963-82317963 on Build GRCh38
rs75677180 Chr.5: 82318047-82318047 on Build GRCh38
rs201256534 Chr.5: 82318043-82318043 on Build GRCh38
rs770491899 Chr.5: 82318145-82318145 on Build GRCh38
rs372250186 Chr.5: 82317972-82317972 on Build GRCh38
rs553279470 Chr.5: 82318003-82318003 on Build GRCh38
rs781367326 Chr.5: 82318064-82318064 on Build GRCh38
rs745337152 Chr.5: 82318131-82318131 on Build GRCh38
rs373541683 Chr.5: 82318138-82318138 on Build GRCh38
rs549556265 Chr.5: 82318125-82318125 on Build GRCh38
rs541773997 Chr.5: 82317976-82317976 on Build GRCh38
rs746383032 Chr.5: 82318079-82318079 on Build GRCh38
rs369765030 Chr.5: 82318063-82318063 on Build GRCh38
rs200466976 Chr.5: 82318085-82318085 on Build GRCh38
rs932784520 Chr.5: 82318045-82318045 on Build GRCh38

View Full Product Details